Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 130624764-130625310 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENOX2
Entrez Gene ID
10495
Gene Name
ecto-NOX disulfide-thiol exchanger 2
Gene Aliases
APK1, COVA1, tNOX
UniGene
Hs.171458
Gene Chromosome Location
Chr.X: 130622330-130903317 on Build GRCh38
Transcripts
NM_001281736.1 NM_006375.3 NM_182314.2 XM_005262354.3 XM_011531244.2 XM_011531245.2 XM_011531247.2 XM_011531249.2 XM_011531251.2 XM_017029226.1 XM_017029227.1 XM_017029228.1 XM_017029229.1

SNP Detail

SNP ID Chromosome Location
rs759803895 Chr.X: 130625277-130625277 on Build GRCh38
rs929720177 Chr.X: 130625066-130625066 on Build GRCh38
rs1801048 Chr.X: 130625212-130625212 on Build GRCh38
rs768546280 Chr.X: 130624835-130624835 on Build GRCh38
rs776613918 Chr.X: 130624841-130624841 on Build GRCh38
rs754313461 Chr.X: 130625256-130625256 on Build GRCh38
rs780325863 Chr.X: 130624959-130624959 on Build GRCh38
rs896625086 Chr.X: 130624861-130624861 on Build GRCh38
rs902914216 Chr.X: 130624985-130624985 on Build GRCh38
rs955204245 Chr.X: 130625149-130625149 on Build GRCh38
rs953492884 Chr.X: 130624834-130624834 on Build GRCh38
rs1032670212 Chr.X: 130625037-130625037 on Build GRCh38
rs1014434827 Chr.X: 130624878-130624878 on Build GRCh38
rs942204743 Chr.X: 130624859-130624859 on Build GRCh38
rs996726896 Chr.X: 130625094-130625094 on Build GRCh38
rs976532375 Chr.X: 130624806-130624806 on Build GRCh38
rs139135250 Chr.X: 130624825-130624825 on Build GRCh38
rs1028166004 Chr.X: 130625113-130625113 on Build GRCh38
rs372658934 Chr.X: 130624842-130624842 on Build GRCh38
rs749535074 Chr.X: 130624964-130624964 on Build GRCh38
rs1045883789 Chr.X: 130625128-130625128 on Build GRCh38
rs779113648 Chr.X: 130625129-130625129 on Build GRCh38
rs1001589253 Chr.X: 130625274-130625274 on Build GRCh38
rs750884527 Chr.X: 130624788-130624788 on Build GRCh38
rs906907485 Chr.X: 130625142-130625142 on Build GRCh38
rs1036545361 Chr.X: 130624998-130624998 on Build GRCh38
rs1045462928 Chr.X: 130624916-130624920 on Build GRCh38
rs754217515 Chr.X: 130625273-130625273 on Build GRCh38
rs987306933 Chr.X: 130625226-130625226 on Build GRCh38
rs768745270 Chr.X: 130625002-130625002 on Build GRCh38
rs957313005 Chr.X: 130625074-130625074 on Build GRCh38
rs1055746048 Chr.X: 130625279-130625279 on Build GRCh38

View Full Product Details