Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 131084136-131084640 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP36
Entrez Gene ID
158763
Gene Name
Rho GTPase activating protein 36
Gene Aliases
-
UniGene
Hs.22905
Gene Chromosome Location
Chr.X: 131058242-131089885 on Build GRCh38
Transcripts
NM_001282607.1 NM_144967.3 XM_006724729.1 XM_011531280.1

SNP Detail

SNP ID Chromosome Location
rs748003647 Chr.X: 131084247-131084247 on Build GRCh38
rs993054055 Chr.X: 131084220-131084220 on Build GRCh38
rs774401636 Chr.X: 131084411-131084412 on Build GRCh38
rs912010620 Chr.X: 131084576-131084576 on Build GRCh38
rs767581133 Chr.X: 131084193-131084193 on Build GRCh38
rs777596367 Chr.X: 131084258-131084258 on Build GRCh38
rs750411009 Chr.X: 131084602-131084602 on Build GRCh38
rs746622736 Chr.X: 131084273-131084273 on Build GRCh38
rs765302887 Chr.X: 131084444-131084444 on Build GRCh38
rs1040982512 Chr.X: 131084514-131084514 on Build GRCh38
rs375987655 Chr.X: 131084221-131084221 on Build GRCh38
rs925970620 Chr.X: 131084462-131084462 on Build GRCh38
rs775947738 Chr.X: 131084364-131084364 on Build GRCh38
rs368655030 Chr.X: 131084606-131084606 on Build GRCh38
rs9306826 Chr.X: 131084477-131084477 on Build GRCh38
rs778740792 Chr.X: 131084337-131084337 on Build GRCh38
rs996757932 Chr.X: 131084541-131084541 on Build GRCh38
rs1008028681 Chr.X: 131084503-131084503 on Build GRCh38
rs778977366 Chr.X: 131084229-131084229 on Build GRCh38
rs780684919 Chr.X: 131084302-131084302 on Build GRCh38
rs758232536 Chr.X: 131084253-131084253 on Build GRCh38
rs916130102 Chr.X: 131084290-131084290 on Build GRCh38
rs1025193472 Chr.X: 131084303-131084303 on Build GRCh38
rs774573780 Chr.X: 131084389-131084389 on Build GRCh38
rs957677166 Chr.X: 131084502-131084502 on Build GRCh38
rs370581558 Chr.X: 131084380-131084380 on Build GRCh38
rs777651830 Chr.X: 131084577-131084577 on Build GRCh38
rs544523095 Chr.X: 131084501-131084501 on Build GRCh38
rs750576860 Chr.X: 131084419-131084419 on Build GRCh38
rs752638035 Chr.X: 131084459-131084459 on Build GRCh38
rs948726991 Chr.X: 131084310-131084310 on Build GRCh38
rs763893915 Chr.X: 131084356-131084356 on Build GRCh38
rs761767794 Chr.X: 131084594-131084594 on Build GRCh38
rs745365369 Chr.X: 131084354-131084354 on Build GRCh38
rs767326463 Chr.X: 131084539-131084539 on Build GRCh38
rs867673869 Chr.X: 131084410-131084411 on Build GRCh38
rs763340310 Chr.X: 131084379-131084379 on Build GRCh38
rs770360336 Chr.X: 131084296-131084296 on Build GRCh38
rs373976010 Chr.X: 131084422-131084422 on Build GRCh38
rs756999878 Chr.X: 131084607-131084607 on Build GRCh38
rs370207587 Chr.X: 131084168-131084168 on Build GRCh38
rs758287542 Chr.X: 131084575-131084575 on Build GRCh38
rs989167574 Chr.X: 131084509-131084509 on Build GRCh38
rs1053193817 Chr.X: 131084486-131084486 on Build GRCh38
rs767631022 Chr.X: 131084417-131084417 on Build GRCh38
rs757307582 Chr.X: 131084187-131084187 on Build GRCh38
rs753725785 Chr.X: 131084197-131084197 on Build GRCh38
rs755012866 Chr.X: 131084223-131084223 on Build GRCh38
rs373627286 Chr.X: 131084169-131084169 on Build GRCh38
rs762032019 Chr.X: 131084409-131084409 on Build GRCh38
rs999440923 Chr.X: 131084493-131084493 on Build GRCh38
rs751741640 Chr.X: 131084184-131084184 on Build GRCh38

View Full Product Details