Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 132078463-132078969 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRMD7
Entrez Gene ID
90167
Gene Name
FERM domain containing 7
Gene Aliases
NYS, NYS1, XIPAN
UniGene
Hs.170776
Gene Chromosome Location
Chr.X: 132076986-132128022 on Build GRCh38
Transcripts
NM_001306193.1 NM_194277.2 XM_017029947.1 XM_017029948.1 XM_017029949.1

SNP Detail

SNP ID Chromosome Location
rs910445814 Chr.X: 132078525-132078525 on Build GRCh38
rs377679619 Chr.X: 132078523-132078523 on Build GRCh38
rs370911852 Chr.X: 132078538-132078538 on Build GRCh38
rs6637934 Chr.X: 132078614-132078614 on Build GRCh38
rs771555307 Chr.X: 132078796-132078796 on Build GRCh38
rs770281308 Chr.X: 132078945-132078945 on Build GRCh38
rs887254851 Chr.X: 132078622-132078622 on Build GRCh38
rs901917911 Chr.X: 132078906-132078906 on Build GRCh38
rs894711989 Chr.X: 132078719-132078719 on Build GRCh38
rs776876410 Chr.X: 132078943-132078943 on Build GRCh38
rs760039580 Chr.X: 132078944-132078944 on Build GRCh38
rs750373967 Chr.X: 132078887-132078887 on Build GRCh38
rs149411634 Chr.X: 132078801-132078801 on Build GRCh38
rs763195387 Chr.X: 132078870-132078870 on Build GRCh38
rs777016184 Chr.X: 132078835-132078835 on Build GRCh38
rs759686216 Chr.X: 132078841-132078841 on Build GRCh38
rs200152758 Chr.X: 132078630-132078630 on Build GRCh38
rs376220211 Chr.X: 132078907-132078907 on Build GRCh38
rs756025701 Chr.X: 132078890-132078890 on Build GRCh38
rs1013206878 Chr.X: 132078742-132078742 on Build GRCh38
rs745594201 Chr.X: 132078527-132078527 on Build GRCh38
rs1021674049 Chr.X: 132078790-132078790 on Build GRCh38
rs867962651 Chr.X: 132078581-132078581 on Build GRCh38
rs143451711 Chr.X: 132078543-132078543 on Build GRCh38
rs748910947 Chr.X: 132078698-132078698 on Build GRCh38
rs781339757 Chr.X: 132078646-132078646 on Build GRCh38
rs199717232 Chr.X: 132078486-132078486 on Build GRCh38
rs767944826 Chr.X: 132078551-132078551 on Build GRCh38
rs146341306 Chr.X: 132078588-132078588 on Build GRCh38
rs745817146 Chr.X: 132078653-132078653 on Build GRCh38
rs772267583 Chr.X: 132078695-132078697 on Build GRCh38
rs1043935466 Chr.X: 132078718-132078718 on Build GRCh38
rs7051368 Chr.X: 132078916-132078916 on Build GRCh38
rs755141737 Chr.X: 132078928-132078928 on Build GRCh38
rs61751728 Chr.X: 132078817-132078817 on Build GRCh38
rs747502543 Chr.X: 132078936-132078936 on Build GRCh38
rs766736966 Chr.X: 132078592-132078592 on Build GRCh38
rs181962233 Chr.X: 132078544-132078544 on Build GRCh38
rs200838126 Chr.X: 132078524-132078524 on Build GRCh38
rs771403134 Chr.X: 132078939-132078939 on Build GRCh38
rs192322553 Chr.X: 132078512-132078512 on Build GRCh38
rs1035849739 Chr.X: 132078688-132078688 on Build GRCh38
rs5977623 Chr.X: 132078484-132078484 on Build GRCh38
rs925472063 Chr.X: 132078750-132078750 on Build GRCh38
rs771620923 Chr.X: 132078934-132078934 on Build GRCh38
rs199576118 Chr.X: 132078536-132078536 on Build GRCh38
rs755898813 Chr.X: 132078663-132078663 on Build GRCh38
rs761002093 Chr.X: 132078590-132078590 on Build GRCh38
rs766452194 Chr.X: 132078487-132078487 on Build GRCh38
rs185946711 Chr.X: 132078854-132078854 on Build GRCh38
rs528032957 Chr.X: 132078549-132078549 on Build GRCh38
rs773855252 Chr.X: 132078775-132078775 on Build GRCh38
rs868605726 Chr.X: 132078582-132078582 on Build GRCh38
rs771224602 Chr.X: 132078834-132078834 on Build GRCh38
rs765439361 Chr.X: 132078615-132078615 on Build GRCh38
rs756312864 Chr.X: 132078759-132078759 on Build GRCh38
rs773577568 Chr.X: 132078587-132078587 on Build GRCh38
rs753601085 Chr.X: 132078895-132078895 on Build GRCh38
rs968426085 Chr.X: 132078872-132078872 on Build GRCh38
rs764235664 Chr.X: 132078871-132078871 on Build GRCh38
rs752957017 Chr.X: 132078863-132078863 on Build GRCh38
rs768253456 Chr.X: 132078774-132078774 on Build GRCh38
rs943531530 Chr.X: 132078818-132078818 on Build GRCh38
rs755054807 Chr.X: 132078491-132078491 on Build GRCh38
rs1039567662 Chr.X: 132078897-132078897 on Build GRCh38
rs772282277 Chr.X: 132078852-132078852 on Build GRCh38
rs779741935 Chr.X: 132078891-132078891 on Build GRCh38
rs746965522 Chr.X: 132078517-132078517 on Build GRCh38
rs754058324 Chr.X: 132078601-132078601 on Build GRCh38
rs138801659 Chr.X: 132078643-132078643 on Build GRCh38
rs779874421 Chr.X: 132078679-132078679 on Build GRCh38
rs754076102 Chr.X: 132078490-132078490 on Build GRCh38
rs777290427 Chr.X: 132078935-132078935 on Build GRCh38

View Full Product Details