Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 134991377-134991884 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMIM10
Entrez Gene ID
644538
Gene Name
small integral membrane protein 10
Gene Aliases
CXorf69
UniGene
Hs.42239
Gene Chromosome Location
Chr.X: 134990938-134992473 on Build GRCh38
Transcripts
NM_001163438.1

SNP Detail

SNP ID Chromosome Location
rs374435572 Chr.X: 134991825-134991825 on Build GRCh38
rs771250589 Chr.X: 134991737-134991737 on Build GRCh38
rs746006610 Chr.X: 134991805-134991805 on Build GRCh38
rs937989150 Chr.X: 134991547-134991547 on Build GRCh38
rs1055147417 Chr.X: 134991599-134991599 on Build GRCh38
rs1000713221 Chr.X: 134991831-134991831 on Build GRCh38
rs748828256 Chr.X: 134991566-134991566 on Build GRCh38
rs149284005 Chr.X: 134991502-134991502 on Build GRCh38
rs11459085 Chr.X: 134991729-134991730 on Build GRCh38
rs143441716 Chr.X: 134991662-134991662 on Build GRCh38
rs1039762923 Chr.X: 134991632-134991632 on Build GRCh38
rs867227492 Chr.X: 134991679-134991679 on Build GRCh38
rs755663451 Chr.X: 134991462-134991462 on Build GRCh38
rs758888451 Chr.X: 134991651-134991652 on Build GRCh38
rs1050802809 Chr.X: 134991663-134991663 on Build GRCh38
rs1038825628 Chr.X: 134991468-134991468 on Build GRCh38
rs751448400 Chr.X: 134991440-134991440 on Build GRCh38
rs921045487 Chr.X: 134991635-134991635 on Build GRCh38
rs914804140 Chr.X: 134991571-134991571 on Build GRCh38
rs866772823 Chr.X: 134991731-134991731 on Build GRCh38
rs866258326 Chr.X: 134991730-134991730 on Build GRCh38
rs369157958 Chr.X: 134991717-134991718 on Build GRCh38
rs764694514 Chr.X: 134991442-134991442 on Build GRCh38
rs983106848 Chr.X: 134991740-134991740 on Build GRCh38
rs981601282 Chr.X: 134991398-134991398 on Build GRCh38
rs956181222 Chr.X: 134991405-134991405 on Build GRCh38
rs11739 Chr.X: 134991445-134991445 on Build GRCh38
rs946436119 Chr.X: 134991614-134991614 on Build GRCh38
rs1013481295 Chr.X: 134991724-134991725 on Build GRCh38
rs764076022 Chr.X: 134991401-134991401 on Build GRCh38
rs908748166 Chr.X: 134991431-134991431 on Build GRCh38
rs776865455 Chr.X: 134991741-134991741 on Build GRCh38
rs867738668 Chr.X: 134991717-134991717 on Build GRCh38

View Full Product Details