Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 134992104-134992612 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMIM10
Entrez Gene ID
644538
Gene Name
small integral membrane protein 10
Gene Aliases
CXorf69
UniGene
Hs.42239
Gene Chromosome Location
Chr.X: 134990938-134992473 on Build GRCh38
Transcripts
NM_001163438.1

SNP Detail

SNP ID Chromosome Location
rs112489209 Chr.X: 134992505-134992505 on Build GRCh38
rs201293806 Chr.X: 134992497-134992498 on Build GRCh38
rs143707742 Chr.X: 134992344-134992344 on Build GRCh38
rs891866972 Chr.X: 134992507-134992507 on Build GRCh38
rs67444257 Chr.X: 134992506-134992507 on Build GRCh38
rs372120042 Chr.X: 134992498-134992498 on Build GRCh38
rs2498769 Chr.X: 134992364-134992364 on Build GRCh38
rs1021904681 Chr.X: 134992173-134992173 on Build GRCh38
rs917957126 Chr.X: 134992240-134992242 on Build GRCh38
rs769955086 Chr.X: 134992562-134992562 on Build GRCh38
rs756376275 Chr.X: 134992504-134992504 on Build GRCh38
rs745948789 Chr.X: 134992495-134992496 on Build GRCh38
rs1011768781 Chr.X: 134992566-134992566 on Build GRCh38
rs73572720 Chr.X: 134992338-134992338 on Build GRCh38
rs1022228309 Chr.X: 134992573-134992573 on Build GRCh38
rs1044293823 Chr.X: 134992482-134992482 on Build GRCh38
rs898436990 Chr.X: 134992390-134992390 on Build GRCh38
rs368875123 Chr.X: 134992314-134992314 on Build GRCh38
rs750105087 Chr.X: 134992292-134992292 on Build GRCh38
rs35608661 Chr.X: 134992415-134992415 on Build GRCh38
rs112436853 Chr.X: 134992505-134992506 on Build GRCh38
rs949298248 Chr.X: 134992289-134992289 on Build GRCh38
rs920943347 Chr.X: 134992216-134992216 on Build GRCh38
rs757618898 Chr.X: 134992499-134992499 on Build GRCh38
rs986550289 Chr.X: 134992241-134992241 on Build GRCh38
rs200774969 Chr.X: 134992495-134992495 on Build GRCh38
rs948665674 Chr.X: 134992434-134992434 on Build GRCh38
rs767364072 Chr.X: 134992267-134992267 on Build GRCh38
rs974998385 Chr.X: 134992209-134992209 on Build GRCh38
rs921706958 Chr.X: 134992525-134992525 on Build GRCh38
rs974289140 Chr.X: 134992196-134992196 on Build GRCh38
rs1031748749 Chr.X: 134992506-134992506 on Build GRCh38
rs933610017 Chr.X: 134992217-134992217 on Build GRCh38
rs796770707 Chr.X: 134992497-134992498 on Build GRCh38
rs965311920 Chr.X: 134992184-134992184 on Build GRCh38
rs189594999 Chr.X: 134992497-134992497 on Build GRCh38
rs746228091 Chr.X: 134992539-134992539 on Build GRCh38
rs938139754 Chr.X: 134992218-134992218 on Build GRCh38
rs137941120 Chr.X: 134992146-134992146 on Build GRCh38

View Full Product Details