Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 136209638-136210137 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FHL1
Entrez Gene ID
2273
Gene Name
four and a half LIM domains 1
Gene Aliases
FHL-1, FHL1A, FHL1B, FLH1A, KYOT, RBMX1A, RBMX1B, SLIM, SLIM-1, SLIM1, SLIMMER, XMPMA
UniGene
Hs.435369
Gene Chromosome Location
Chr.X: 136146702-136211359 on Build GRCh38
Transcripts
NM_001159699.1 NM_001159700.1 NM_001159701.1 NM_001159702.2 NM_001159703.1 NM_001159704.1 NM_001167819.1 NM_001449.4 NR_027621.1 XM_006724743.2 XM_006724744.2 XM_006724745.3 XM_006724746.2 XM_006724747.2 XM_011531316.1 XM_017029357.1

SNP Detail

SNP ID Chromosome Location
rs1017189624 Chr.X: 136209755-136209755 on Build GRCh38
rs151315725 Chr.X: 136210005-136210005 on Build GRCh38
rs1039991254 Chr.X: 136210002-136210002 on Build GRCh38
rs368428875 Chr.X: 136209863-136209863 on Build GRCh38
rs888816161 Chr.X: 136209687-136209687 on Build GRCh38
rs777561065 Chr.X: 136209838-136209839 on Build GRCh38
rs141231353 Chr.X: 136209920-136209920 on Build GRCh38
rs140605116 Chr.X: 136210068-136210068 on Build GRCh38
rs761074507 Chr.X: 136209859-136209863 on Build GRCh38
rs781706497 Chr.X: 136209758-136209758 on Build GRCh38
rs752156219 Chr.X: 136209868-136209869 on Build GRCh38
rs776759929 Chr.X: 136209853-136209853 on Build GRCh38
rs749265329 Chr.X: 136209966-136209966 on Build GRCh38
rs56391414 Chr.X: 136209928-136209928 on Build GRCh38
rs772909951 Chr.X: 136209994-136209994 on Build GRCh38
rs965735796 Chr.X: 136209748-136209748 on Build GRCh38
rs779166852 Chr.X: 136209929-136209929 on Build GRCh38
rs886043219 Chr.X: 136209995-136209995 on Build GRCh38
rs1026588152 Chr.X: 136209779-136209779 on Build GRCh38
rs1004023354 Chr.X: 136209749-136209749 on Build GRCh38
rs778893019 Chr.X: 136210107-136210107 on Build GRCh38
rs886543119 Chr.X: 136209741-136209741 on Build GRCh38
rs747922027 Chr.X: 136209990-136209990 on Build GRCh38
rs774024150 Chr.X: 136209986-136209986 on Build GRCh38
rs751855746 Chr.X: 136210077-136210077 on Build GRCh38
rs893407799 Chr.X: 136209673-136209673 on Build GRCh38
rs192534729 Chr.X: 136209799-136209799 on Build GRCh38
rs758552293 Chr.X: 136209950-136209950 on Build GRCh38
rs775356347 Chr.X: 136210004-136210004 on Build GRCh38
rs779145852 Chr.X: 136209829-136209829 on Build GRCh38
rs772286355 Chr.X: 136209843-136209843 on Build GRCh38
rs771803774 Chr.X: 136209866-136209866 on Build GRCh38
rs981405852 Chr.X: 136209808-136209808 on Build GRCh38
rs184761695 Chr.X: 136209800-136209800 on Build GRCh38
rs869025431 Chr.X: 136209946-136209946 on Build GRCh38
rs2076705 Chr.X: 136209863-136209863 on Build GRCh38
rs774637390 Chr.X: 136209841-136209843 on Build GRCh38
rs775118713 Chr.X: 136209872-136209872 on Build GRCh38
rs757510318 Chr.X: 136210067-136210067 on Build GRCh38
rs780234670 Chr.X: 136209676-136209676 on Build GRCh38
rs771639876 Chr.X: 136209836-136209837 on Build GRCh38
rs1045887959 Chr.X: 136209660-136209660 on Build GRCh38
rs769930579 Chr.X: 136209707-136209707 on Build GRCh38
rs794727853 Chr.X: 136209868-136209868 on Build GRCh38
rs750540064 Chr.X: 136210042-136210042 on Build GRCh38
rs955990553 Chr.X: 136209860-136209860 on Build GRCh38
rs866716263 Chr.X: 136209853-136209853 on Build GRCh38
rs375981682 Chr.X: 136209839-136209839 on Build GRCh38
rs760161304 Chr.X: 136209914-136209914 on Build GRCh38
rs951898374 Chr.X: 136209780-136209780 on Build GRCh38
rs768923941 Chr.X: 136209854-136209854 on Build GRCh38
rs751993620 Chr.X: 136209701-136209701 on Build GRCh38
rs750748012 Chr.X: 136209857-136209858 on Build GRCh38
rs1012775757 Chr.X: 136209787-136209787 on Build GRCh38
rs869025432 Chr.X: 136209947-136209947 on Build GRCh38
rs200510559 Chr.X: 136209968-136209968 on Build GRCh38
rs766836412 Chr.X: 136209858-136209859 on Build GRCh38
rs886043885 Chr.X: 136209862-136209862 on Build GRCh38
rs1049996765 Chr.X: 136209685-136209685 on Build GRCh38
rs773413708 Chr.X: 136209757-136209757 on Build GRCh38
rs914754937 Chr.X: 136209862-136209862 on Build GRCh38
rs754981844 Chr.X: 136210105-136210105 on Build GRCh38
rs758786635 Chr.X: 136210045-136210045 on Build GRCh38
rs62603142 Chr.X: 136209661-136209661 on Build GRCh38
rs190006104 Chr.X: 136209951-136209951 on Build GRCh38
rs767421819 Chr.X: 136209861-136209861 on Build GRCh38
rs774150299 Chr.X: 136209864-136209864 on Build GRCh38
rs1018653824 Chr.X: 136209716-136209716 on Build GRCh38
rs370017771 Chr.X: 136210041-136210041 on Build GRCh38
rs122459148 Chr.X: 136210023-136210023 on Build GRCh38
rs11798700 Chr.X: 136209858-136209858 on Build GRCh38
rs933337742 Chr.X: 136209705-136209705 on Build GRCh38
rs747054561 Chr.X: 136209848-136209848 on Build GRCh38
rs780191041 Chr.X: 136209960-136209960 on Build GRCh38
rs745879108 Chr.X: 136210081-136210081 on Build GRCh38
rs767661713 Chr.X: 136209854-136209855 on Build GRCh38
rs746813826 Chr.X: 136209839-136209843 on Build GRCh38
rs774307953 Chr.X: 136209854-136209858 on Build GRCh38
rs764263558 Chr.X: 136210053-136210053 on Build GRCh38
rs777315661 Chr.X: 136210098-136210098 on Build GRCh38
rs187514969 Chr.X: 136209664-136209664 on Build GRCh38
rs1053251417 Chr.X: 136209668-136209668 on Build GRCh38
rs754308516 Chr.X: 136209921-136209921 on Build GRCh38
rs374867802 Chr.X: 136209853-136209854 on Build GRCh38
rs755327922 Chr.X: 136209820-136209820 on Build GRCh38
rs377071251 Chr.X: 136209970-136209970 on Build GRCh38
rs771676532 Chr.X: 136209993-136209993 on Build GRCh38
rs754404850 Chr.X: 136209860-136209863 on Build GRCh38
rs755471982 Chr.X: 136209723-136209723 on Build GRCh38
rs765851828 Chr.X: 136210001-136210001 on Build GRCh38
rs758031344 Chr.X: 136210032-136210033 on Build GRCh38
rs761406765 Chr.X: 136209865-136209865 on Build GRCh38
rs769912263 Chr.X: 136209857-136209857 on Build GRCh38
rs755385158 Chr.X: 136209926-136209926 on Build GRCh38
rs760214177 Chr.X: 136209997-136209997 on Build GRCh38
rs768705041 Chr.X: 136209943-136209943 on Build GRCh38
rs762027610 Chr.X: 136209854-136209856 on Build GRCh38
rs763065210 Chr.X: 136210061-136210061 on Build GRCh38
rs981817116 Chr.X: 136209798-136209798 on Build GRCh38
rs778432832 Chr.X: 136209862-136209863 on Build GRCh38

View Full Product Details