Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 138711295-138711782 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGF13
Entrez Gene ID
2258
Gene Name
fibroblast growth factor 13
Gene Aliases
FGF-13, FGF2, FHF-2, FHF2
UniGene
Hs.6540
Gene Chromosome Location
Chr.X: 138631573-139222889 on Build GRCh38
Transcripts
NM_001139498.1 NM_001139500.1 NM_001139501.1 NM_001139502.1 NM_004114.3 NM_033642.2 XM_005262399.1

SNP Detail

SNP ID Chromosome Location
rs918980911 Chr.X: 138711513-138711513 on Build GRCh38
rs1017464851 Chr.X: 138711737-138711737 on Build GRCh38
rs931764641 Chr.X: 138711648-138711648 on Build GRCh38
rs12849929 Chr.X: 138711491-138711491 on Build GRCh38
rs75517136 Chr.X: 138711356-138711356 on Build GRCh38
rs1012418783 Chr.X: 138711336-138711336 on Build GRCh38
rs983345647 Chr.X: 138711588-138711588 on Build GRCh38
rs985973409 Chr.X: 138711468-138711468 on Build GRCh38
rs12009215 Chr.X: 138711342-138711342 on Build GRCh38
rs79643183 Chr.X: 138711402-138711402 on Build GRCh38
rs12013797 Chr.X: 138711348-138711348 on Build GRCh38
rs757479747 Chr.X: 138711352-138711353 on Build GRCh38
rs956414917 Chr.X: 138711566-138711566 on Build GRCh38
rs929107503 Chr.X: 138711565-138711565 on Build GRCh38
rs757772435 Chr.X: 138711438-138711438 on Build GRCh38
rs78024675 Chr.X: 138711369-138711369 on Build GRCh38
rs187191248 Chr.X: 138711673-138711673 on Build GRCh38
rs369215573 Chr.X: 138711404-138711404 on Build GRCh38
rs989582305 Chr.X: 138711589-138711589 on Build GRCh38
rs910016508 Chr.X: 138711607-138711607 on Build GRCh38
rs1056033341 Chr.X: 138711650-138711650 on Build GRCh38
rs879092437 Chr.X: 138711486-138711486 on Build GRCh38
rs957210810 Chr.X: 138711461-138711461 on Build GRCh38
rs12851345 Chr.X: 138711385-138711385 on Build GRCh38
rs941512165 Chr.X: 138711609-138711609 on Build GRCh38
rs74469656 Chr.X: 138711366-138711366 on Build GRCh38
rs762860989 Chr.X: 138711379-138711379 on Build GRCh38
rs998917585 Chr.X: 138711693-138711693 on Build GRCh38
rs1023844744 Chr.X: 138711442-138711442 on Build GRCh38
rs75134854 Chr.X: 138711399-138711399 on Build GRCh38
rs766703773 Chr.X: 138711355-138711356 on Build GRCh38
rs76752485 Chr.X: 138711378-138711378 on Build GRCh38
rs780361755 Chr.X: 138711503-138711503 on Build GRCh38
rs916178860 Chr.X: 138711658-138711658 on Build GRCh38
rs768165794 Chr.X: 138711394-138711405 on Build GRCh38
rs944595096 Chr.X: 138711723-138711723 on Build GRCh38
rs79825076 Chr.X: 138711376-138711376 on Build GRCh38
rs76780734 Chr.X: 138711400-138711400 on Build GRCh38
rs190473772 Chr.X: 138711705-138711705 on Build GRCh38
rs982388883 Chr.X: 138711512-138711512 on Build GRCh38
rs11555172 Chr.X: 138711527-138711527 on Build GRCh38
rs971353498 Chr.X: 138711453-138711453 on Build GRCh38
rs999093255 Chr.X: 138711457-138711457 on Build GRCh38
rs367786961 Chr.X: 138711355-138711355 on Build GRCh38
rs965656809 Chr.X: 138711504-138711504 on Build GRCh38
rs77396080 Chr.X: 138711398-138711398 on Build GRCh38
rs764363352 Chr.X: 138711405-138711405 on Build GRCh38
rs779165005 Chr.X: 138711455-138711455 on Build GRCh38
rs765891135 Chr.X: 138711385-138711385 on Build GRCh38
rs903259327 Chr.X: 138711678-138711678 on Build GRCh38
rs950013476 Chr.X: 138711667-138711667 on Build GRCh38
rs1032879439 Chr.X: 138711460-138711460 on Build GRCh38
rs754427469 Chr.X: 138711436-138711436 on Build GRCh38
rs975805242 Chr.X: 138711506-138711506 on Build GRCh38
rs778394309 Chr.X: 138711323-138711323 on Build GRCh38
rs773890668 Chr.X: 138711353-138711354 on Build GRCh38
rs1054952576 Chr.X: 138711697-138711697 on Build GRCh38
rs185599206 Chr.X: 138711742-138711742 on Build GRCh38
rs911727116 Chr.X: 138711712-138711712 on Build GRCh38
rs12849923 Chr.X: 138711480-138711480 on Build GRCh38
rs80315101 Chr.X: 138711361-138711361 on Build GRCh38
rs5012044 Chr.X: 138711315-138711315 on Build GRCh38
rs1031063565 Chr.X: 138711523-138711523 on Build GRCh38
rs183084908 Chr.X: 138711717-138711717 on Build GRCh38
rs773787868 Chr.X: 138711393-138711393 on Build GRCh38

View Full Product Details