Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 186375661-186376186 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf27
Entrez Gene ID
54953
Gene Name
chromosome 1 open reading frame 27
Gene Aliases
ODR4, TTG1, odr-4
UniGene
Hs.371210
Gene Chromosome Location
Chr.1: 186375758-186421375 on Build GRCh38
Transcripts
NM_001164245.1 NM_001164246.1 NM_017847.5 XM_011509660.1 XM_011509661.1 XM_017001543.1

SNP Detail

SNP ID Chromosome Location
rs1011837127 Chr.1: 186375821-186375821 on Build GRCh38
rs775974888 Chr.1: 186375992-186375993 on Build GRCh38
rs936333758 Chr.1: 186376027-186376027 on Build GRCh38
rs760763644 Chr.1: 186375721-186375721 on Build GRCh38
rs879449576 Chr.1: 186375987-186375988 on Build GRCh38
rs775716127 Chr.1: 186376021-186376022 on Build GRCh38
rs970547064 Chr.1: 186375711-186375711 on Build GRCh38
rs765554253 Chr.1: 186375879-186375879 on Build GRCh38
rs975008726 Chr.1: 186375752-186375752 on Build GRCh38
rs1007633069 Chr.1: 186375922-186375922 on Build GRCh38
rs147710547 Chr.1: 186376082-186376082 on Build GRCh38
rs974702821 Chr.1: 186375914-186375914 on Build GRCh38
rs3737937 Chr.1: 186375686-186375686 on Build GRCh38
rs182136141 Chr.1: 186375851-186375851 on Build GRCh38
rs367903899 Chr.1: 186375726-186375726 on Build GRCh38
rs1016325145 Chr.1: 186375895-186375895 on Build GRCh38
rs1052354757 Chr.1: 186375886-186375886 on Build GRCh38
rs9660553 Chr.1: 186376021-186376021 on Build GRCh38
rs892359006 Chr.1: 186375890-186375890 on Build GRCh38
rs936832547 Chr.1: 186375974-186375974 on Build GRCh38
rs1057201918 Chr.1: 186375785-186375785 on Build GRCh38
rs972504699 Chr.1: 186375712-186375712 on Build GRCh38
rs894823769 Chr.1: 186375819-186375819 on Build GRCh38
rs57930082 Chr.1: 186376022-186376023 on Build GRCh38
rs1024854877 Chr.1: 186375861-186375861 on Build GRCh38
rs113247382 Chr.1: 186375993-186375994 on Build GRCh38
rs1285180 Chr.1: 186375881-186375881 on Build GRCh38
rs762174294 Chr.1: 186375870-186375870 on Build GRCh38
rs767991145 Chr.1: 186375987-186375987 on Build GRCh38
rs6425027 Chr.1: 186376034-186376034 on Build GRCh38
rs189291872 Chr.1: 186375776-186375776 on Build GRCh38
rs923484373 Chr.1: 186375772-186375772 on Build GRCh38
rs994039894 Chr.1: 186375874-186375874 on Build GRCh38
rs997368168 Chr.1: 186375898-186375898 on Build GRCh38
rs1012144492 Chr.1: 186375693-186375693 on Build GRCh38
rs3831933 Chr.1: 186375744-186375745 on Build GRCh38
rs184597478 Chr.1: 186376116-186376116 on Build GRCh38
rs184506929 Chr.1: 186375716-186375716 on Build GRCh38
rs114841599 Chr.1: 186375925-186375925 on Build GRCh38
rs116008694 Chr.1: 186375959-186375959 on Build GRCh38
rs551965315 Chr.1: 186375792-186375792 on Build GRCh38
rs6425028 Chr.1: 186376149-186376149 on Build GRCh38
rs376280787 Chr.1: 186375995-186375996 on Build GRCh38
rs933479770 Chr.1: 186375775-186375775 on Build GRCh38
rs894800001 Chr.1: 186375692-186375692 on Build GRCh38
rs1030843847 Chr.1: 186375697-186375697 on Build GRCh38
rs145150452 Chr.1: 186376077-186376077 on Build GRCh38
rs964716527 Chr.1: 186375910-186375910 on Build GRCh38
rs36122553 Chr.1: 186375703-186375703 on Build GRCh38
rs978328227 Chr.1: 186375951-186375951 on Build GRCh38
rs947813776 Chr.1: 186375820-186375820 on Build GRCh38
rs919469560 Chr.1: 186375745-186375746 on Build GRCh38
rs960258167 Chr.1: 186375974-186375974 on Build GRCh38
rs994074125 Chr.1: 186376107-186376107 on Build GRCh38
rs950901587 Chr.1: 186375724-186375724 on Build GRCh38
rs936449289 Chr.1: 186375790-186375790 on Build GRCh38
rs916297318 Chr.1: 186376053-186376053 on Build GRCh38
rs201906770 Chr.1: 186375994-186376001 on Build GRCh38
rs956524920 Chr.1: 186375695-186375695 on Build GRCh38
rs916068593 Chr.1: 186375985-186375985 on Build GRCh38
rs578061133 Chr.1: 186376067-186376067 on Build GRCh38
rs758941016 Chr.1: 186376096-186376096 on Build GRCh38
rs909232814 Chr.1: 186375733-186375733 on Build GRCh38
rs997315573 Chr.1: 186375697-186375697 on Build GRCh38
rs892411485 Chr.1: 186376041-186376041 on Build GRCh38
rs943364654 Chr.1: 186375746-186375746 on Build GRCh38
rs972285048 Chr.1: 186375745-186375745 on Build GRCh38
rs746510487 Chr.1: 186375985-186375986 on Build GRCh38
rs985304262 Chr.1: 186375882-186375882 on Build GRCh38
rs192872862 Chr.1: 186375709-186375709 on Build GRCh38
rs947830485 Chr.1: 186376002-186376003 on Build GRCh38
rs924986562 Chr.1: 186376023-186376023 on Build GRCh38
rs972003503 Chr.1: 186375944-186375947 on Build GRCh38
rs111555426 Chr.1: 186375932-186375932 on Build GRCh38
rs770649531 Chr.1: 186375984-186375985 on Build GRCh38
rs865964837 Chr.1: 186376072-186376072 on Build GRCh38
rs879177779 Chr.1: 186375779-186375779 on Build GRCh38
rs112597707 Chr.1: 186375789-186375789 on Build GRCh38
rs1052064956 Chr.1: 186376102-186376103 on Build GRCh38

View Full Product Details