Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 186390487-186390995 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf27
Entrez Gene ID
54953
Gene Name
chromosome 1 open reading frame 27
Gene Aliases
ODR4, TTG1, odr-4
UniGene
Hs.371210
Gene Chromosome Location
Chr.1: 186375758-186421375 on Build GRCh38
Transcripts
NM_001164245.1 NM_001164246.1 NM_017847.5 XM_011509660.1 XM_011509661.1 XM_017001543.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3369471 c.507A>G p.Q169Q Chr.1: 186390743-186390743 on Build GRCh38
1337223 c.531T>G p.L177L Chr.1: 186390767-186390767 on Build GRCh38
5622530 c.503A>T p.Y168F Chr.1: 186390739-186390739 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs773519415 Chr.1: 186390798-186390798 on Build GRCh38
rs769220502 Chr.1: 186390782-186390782 on Build GRCh38
rs915146564 Chr.1: 186390582-186390582 on Build GRCh38
rs761880073 Chr.1: 186390686-186390686 on Build GRCh38
rs750592224 Chr.1: 186390731-186390731 on Build GRCh38
rs1042034099 Chr.1: 186390592-186390592 on Build GRCh38
rs752364848 Chr.1: 186390832-186390832 on Build GRCh38
rs890129977 Chr.1: 186390605-186390605 on Build GRCh38
rs765089806 Chr.1: 186390810-186390810 on Build GRCh38
rs751520005 Chr.1: 186390885-186390885 on Build GRCh38
rs1027851785 Chr.1: 186390675-186390675 on Build GRCh38
rs890930565 Chr.1: 186390919-186390919 on Build GRCh38
rs764896739 Chr.1: 186390734-186390734 on Build GRCh38
rs750105484 Chr.1: 186390899-186390899 on Build GRCh38
rs781039619 Chr.1: 186390761-186390761 on Build GRCh38
rs367658772 Chr.1: 186390694-186390694 on Build GRCh38
rs1023011137 Chr.1: 186390570-186390570 on Build GRCh38
rs557617777 Chr.1: 186390877-186390877 on Build GRCh38
rs751180543 Chr.1: 186390757-186390757 on Build GRCh38
rs376120716 Chr.1: 186390758-186390758 on Build GRCh38
rs937845802 Chr.1: 186390536-186390536 on Build GRCh38
rs776929590 Chr.1: 186390862-186390862 on Build GRCh38
rs560523977 Chr.1: 186390578-186390578 on Build GRCh38
rs766848929 Chr.1: 186390866-186390867 on Build GRCh38
rs775733948 Chr.1: 186390561-186390561 on Build GRCh38
rs919873302 Chr.1: 186390808-186390808 on Build GRCh38
rs139232938 Chr.1: 186390736-186390736 on Build GRCh38
rs992284252 Chr.1: 186390754-186390754 on Build GRCh38
rs755657501 Chr.1: 186390901-186390901 on Build GRCh38
rs12736676 Chr.1: 186390631-186390631 on Build GRCh38
rs3115752 Chr.1: 186390618-186390618 on Build GRCh38
rs900288805 Chr.1: 186390654-186390654 on Build GRCh38
rs764174056 Chr.1: 186390669-186390669 on Build GRCh38
rs561421993 Chr.1: 186390964-186390964 on Build GRCh38
rs187494832 Chr.1: 186390792-186390792 on Build GRCh38
rs371831284 Chr.1: 186390738-186390738 on Build GRCh38
rs932553165 Chr.1: 186390869-186390869 on Build GRCh38
rs555970257 Chr.1: 186390593-186390593 on Build GRCh38
rs1039195987 Chr.1: 186390621-186390621 on Build GRCh38
rs201865055 Chr.1: 186390839-186390839 on Build GRCh38
rs557173093 Chr.1: 186390543-186390544 on Build GRCh38
rs759419943 Chr.1: 186390733-186390733 on Build GRCh38
rs772111453 Chr.1: 186390797-186390797 on Build GRCh38
rs988415744 Chr.1: 186390511-186390511 on Build GRCh38
rs368373111 Chr.1: 186390780-186390780 on Build GRCh38
rs945320353 Chr.1: 186390590-186390590 on Build GRCh38
rs543347022 Chr.1: 186390963-186390963 on Build GRCh38
rs780226436 Chr.1: 186390778-186390778 on Build GRCh38
rs575735318 Chr.1: 186390937-186390937 on Build GRCh38
rs200057679 Chr.1: 186390897-186390897 on Build GRCh38
rs774829162 Chr.1: 186390789-186390789 on Build GRCh38
rs777423436 Chr.1: 186390743-186390743 on Build GRCh38
rs745478585 Chr.1: 186390762-186390762 on Build GRCh38
rs375472243 Chr.1: 186390809-186390809 on Build GRCh38
rs994993868 Chr.1: 186390667-186390667 on Build GRCh38
rs533549512 Chr.1: 186390683-186390683 on Build GRCh38
rs761129676 Chr.1: 186390786-186390786 on Build GRCh38
rs770217455 Chr.1: 186390768-186390768 on Build GRCh38

View Full Product Details