Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 190097585-190098083 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRINP3
Entrez Gene ID
339479
Gene Name
BMP/retinoic acid inducible neural specific 3
Gene Aliases
DBCCR1L, DBCCR1L1, FAM5C
UniGene
Hs.65765
Gene Chromosome Location
Chr.1: 190097662-190477997 on Build GRCh38
Transcripts
NM_001317188.1 NM_199051.2 XM_011509472.2 XM_011509475.2 XM_011509476.2 XM_017001125.1 XM_017001126.1 XM_017001127.1 XM_017001128.1 XM_017001129.1 XM_017001130.1 XM_017001131.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6060332 c.2294G>T p.C765F Chr.1: 190098025-190098025 on Build GRCh38
6060333 c.2297G>T p.S766I Chr.1: 190098022-190098022 on Build GRCh38
5775963 c.2271G>C p.M757I Chr.1: 190098048-190098048 on Build GRCh38
6060330 c.2261C>A p.P754Q Chr.1: 190098058-190098058 on Build GRCh38
6060331 c.2272G>C p.D758H Chr.1: 190098047-190098047 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs187230047 Chr.1: 190097752-190097752 on Build GRCh38
rs1935880 Chr.1: 190097612-190097612 on Build GRCh38
rs766484744 Chr.1: 190098040-190098040 on Build GRCh38
rs776187925 Chr.1: 190097680-190097680 on Build GRCh38
rs542547026 Chr.1: 190097639-190097639 on Build GRCh38
rs1029952566 Chr.1: 190097721-190097721 on Build GRCh38
rs190588717 Chr.1: 190097797-190097797 on Build GRCh38
rs35296429 Chr.1: 190097780-190097780 on Build GRCh38
rs1053081 Chr.1: 190097855-190097855 on Build GRCh38
rs375495949 Chr.1: 190097990-190097990 on Build GRCh38
rs1006441144 Chr.1: 190097610-190097610 on Build GRCh38
rs762603802 Chr.1: 190097863-190097863 on Build GRCh38
rs928639437 Chr.1: 190097939-190097939 on Build GRCh38
rs536932261 Chr.1: 190097940-190097940 on Build GRCh38
rs781219796 Chr.1: 190097980-190097980 on Build GRCh38
rs567300221 Chr.1: 190097907-190097907 on Build GRCh38
rs990289296 Chr.1: 190097820-190097820 on Build GRCh38
rs900516833 Chr.1: 190097679-190097679 on Build GRCh38
rs550548481 Chr.1: 190097892-190097892 on Build GRCh38
rs762168189 Chr.1: 190098026-190098026 on Build GRCh38
rs377516098 Chr.1: 190097981-190097981 on Build GRCh38
rs768024264 Chr.1: 190098030-190098030 on Build GRCh38
rs1008097714 Chr.1: 190097819-190097819 on Build GRCh38
rs966814399 Chr.1: 190097781-190097781 on Build GRCh38
rs981556457 Chr.1: 190097905-190097905 on Build GRCh38
rs775113006 Chr.1: 190097997-190097997 on Build GRCh38
rs1028267289 Chr.1: 190097728-190097728 on Build GRCh38
rs1032394126 Chr.1: 190097655-190097655 on Build GRCh38
rs542329663 Chr.1: 190098033-190098033 on Build GRCh38
rs141719999 Chr.1: 190098052-190098052 on Build GRCh38
rs768564630 Chr.1: 190097660-190097660 on Build GRCh38
rs900225774 Chr.1: 190097625-190097625 on Build GRCh38
rs751325612 Chr.1: 190098059-190098059 on Build GRCh38
rs769302650 Chr.1: 190097817-190097817 on Build GRCh38
rs761166562 Chr.1: 190098037-190098037 on Build GRCh38
rs996156111 Chr.1: 190097715-190097715 on Build GRCh38
rs1047739324 Chr.1: 190097961-190097961 on Build GRCh38
rs749103035 Chr.1: 190097673-190097674 on Build GRCh38
rs755187296 Chr.1: 190098051-190098051 on Build GRCh38
rs997224454 Chr.1: 190097690-190097690 on Build GRCh38
rs868132084 Chr.1: 190098047-190098047 on Build GRCh38
rs373703733 Chr.1: 190097874-190097874 on Build GRCh38
rs747592391 Chr.1: 190097732-190097732 on Build GRCh38
rs551630764 Chr.1: 190097637-190097638 on Build GRCh38
rs768301367 Chr.1: 190098005-190098005 on Build GRCh38
rs774607581 Chr.1: 190098019-190098019 on Build GRCh38
rs559236851 Chr.1: 190097668-190097668 on Build GRCh38
rs753907285 Chr.1: 190098044-190098044 on Build GRCh38
rs1032363526 Chr.1: 190097803-190097803 on Build GRCh38
rs113262339 Chr.1: 190097898-190097898 on Build GRCh38
rs369279585 Chr.1: 190097707-190097707 on Build GRCh38
rs748939297 Chr.1: 190098003-190098003 on Build GRCh38
rs941963503 Chr.1: 190098021-190098021 on Build GRCh38
rs528261544 Chr.1: 190097744-190097744 on Build GRCh38
rs552205865 Chr.1: 190097846-190097846 on Build GRCh38

View Full Product Details