Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 190226230-190226741 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRINP3
Entrez Gene ID
339479
Gene Name
BMP/retinoic acid inducible neural specific 3
Gene Aliases
DBCCR1L, DBCCR1L1, FAM5C
UniGene
Hs.65765
Gene Chromosome Location
Chr.1: 190097662-190477997 on Build GRCh38
Transcripts
NM_001317188.1 NM_199051.2 XM_011509472.2 XM_011509475.2 XM_011509476.2 XM_017001125.1 XM_017001126.1 XM_017001127.1 XM_017001128.1 XM_017001129.1 XM_017001130.1 XM_017001131.1 XM_017001132.1 XM_017001133.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1295614 c.799T>A p.S267T Chr.1: 190226244-190226244 on Build GRCh38
3480471 c.740T>G p.L247R Chr.1: 190226303-190226303 on Build GRCh38
7357741 c.745G>T p.D249Y Chr.1: 190226298-190226298 on Build GRCh38
4026245 c.737T>G p.L246R Chr.1: 190226306-190226306 on Build GRCh38
5542910 c.763T>A p.F255I Chr.1: 190226280-190226280 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752755498 Chr.1: 190226288-190226288 on Build GRCh38
rs1002833121 Chr.1: 190226560-190226560 on Build GRCh38
rs983244284 Chr.1: 190226596-190226596 on Build GRCh38
rs1054412866 Chr.1: 190226699-190226699 on Build GRCh38
rs539345204 Chr.1: 190226622-190226622 on Build GRCh38
rs758562628 Chr.1: 190226301-190226301 on Build GRCh38
rs781261537 Chr.1: 190226540-190226540 on Build GRCh38
rs900094114 Chr.1: 190226381-190226381 on Build GRCh38
rs879451126 Chr.1: 190226654-190226654 on Build GRCh38
rs959413126 Chr.1: 190226590-190226590 on Build GRCh38
rs747225844 Chr.1: 190226316-190226316 on Build GRCh38
rs148869641 Chr.1: 190226283-190226283 on Build GRCh38
rs768595432 Chr.1: 190226335-190226335 on Build GRCh38
rs908928506 Chr.1: 190226268-190226268 on Build GRCh38
rs1008457604 Chr.1: 190226544-190226544 on Build GRCh38
rs200455431 Chr.1: 190226325-190226325 on Build GRCh38
rs963431031 Chr.1: 190226623-190226623 on Build GRCh38
rs778025234 Chr.1: 190226312-190226312 on Build GRCh38
rs528421572 Chr.1: 190226282-190226282 on Build GRCh38
rs934662507 Chr.1: 190226410-190226410 on Build GRCh38
rs74131321 Chr.1: 190226435-190226435 on Build GRCh38
rs960993311 Chr.1: 190226567-190226568 on Build GRCh38
rs949302237 Chr.1: 190226362-190226362 on Build GRCh38
rs879602676 Chr.1: 190226259-190226259 on Build GRCh38
rs908981602 Chr.1: 190226619-190226619 on Build GRCh38
rs891333973 Chr.1: 190226419-190226419 on Build GRCh38
rs368693065 Chr.1: 190226537-190226537 on Build GRCh38
rs998969256 Chr.1: 190226434-190226434 on Build GRCh38
rs745666527 Chr.1: 190226278-190226278 on Build GRCh38
rs997010008 Chr.1: 190226436-190226436 on Build GRCh38
rs934652440 Chr.1: 190226686-190226686 on Build GRCh38
rs567326372 Chr.1: 190226438-190226438 on Build GRCh38
rs181251228 Chr.1: 190226700-190226700 on Build GRCh38
rs754438802 Chr.1: 190226279-190226279 on Build GRCh38
rs983234241 Chr.1: 190226266-190226266 on Build GRCh38
rs1035600486 Chr.1: 190226572-190226572 on Build GRCh38
rs189688562 Chr.1: 190226576-190226576 on Build GRCh38
rs373471707 Chr.1: 190226703-190226703 on Build GRCh38
rs1011593762 Chr.1: 190226499-190226499 on Build GRCh38
rs773742006 Chr.1: 190226355-190226355 on Build GRCh38
rs974504934 Chr.1: 190226678-190226678 on Build GRCh38
rs566326901 Chr.1: 190226593-190226593 on Build GRCh38
rs575445483 Chr.1: 190226691-190226691 on Build GRCh38
rs370890229 Chr.1: 190226500-190226500 on Build GRCh38
rs1026728274 Chr.1: 190226577-190226577 on Build GRCh38
rs530689944 Chr.1: 190226403-190226403 on Build GRCh38
rs977939403 Chr.1: 190226629-190226629 on Build GRCh38
rs540678302 Chr.1: 190226390-190226390 on Build GRCh38
rs766847160 Chr.1: 190226276-190226276 on Build GRCh38
rs200602238 Chr.1: 190226269-190226269 on Build GRCh38
rs921616074 Chr.1: 190226679-190226679 on Build GRCh38
rs750832980 Chr.1: 190226256-190226256 on Build GRCh38
rs536352080 Chr.1: 190226546-190226546 on Build GRCh38
rs1054465272 Chr.1: 190226431-190226431 on Build GRCh38
rs949229446 Chr.1: 190226629-190226632 on Build GRCh38
rs551365952 Chr.1: 190226332-190226332 on Build GRCh38
rs61819046 Chr.1: 190226387-190226387 on Build GRCh38
rs558955157 Chr.1: 190226684-190226684 on Build GRCh38
rs893169714 Chr.1: 190226493-190226493 on Build GRCh38

View Full Product Details