Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 193135189-193135686 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC73
Entrez Gene ID
79577
Gene Name
cell division cycle 73
Gene Aliases
C1orf28, FIHP, HPTJT, HRPT1, HRPT2, HYX
UniGene
Hs.378996
Gene Chromosome Location
Chr.1: 193121958-193254815 on Build GRCh38
Transcripts
NM_024529.4 XM_006711537.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6917946 c.325G>T p.D109Y Chr.1: 193135408-193135408 on Build GRCh38
5441868 c.415C>G p.R139G Chr.1: 193135581-193135581 on Build GRCh38
7448262 c.312A>G p.T104T Chr.1: 193135395-193135395 on Build GRCh38
4670742 c.383C>T p.A128V Chr.1: 193135549-193135549 on Build GRCh38
331152 c.366_367TC>CT p.T122>? Chr.1: 193135449-193135450 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753791966 Chr.1: 193135369-193135369 on Build GRCh38
rs748367546 Chr.1: 193135618-193135618 on Build GRCh38
rs768985628 Chr.1: 193135505-193135506 on Build GRCh38
rs968260282 Chr.1: 193135532-193135532 on Build GRCh38
rs778469738 Chr.1: 193135371-193135371 on Build GRCh38
rs189907310 Chr.1: 193135665-193135665 on Build GRCh38
rs761807789 Chr.1: 193135431-193135431 on Build GRCh38
rs760583012 Chr.1: 193135470-193135470 on Build GRCh38
rs1046974673 Chr.1: 193135290-193135290 on Build GRCh38
rs886041278 Chr.1: 193135438-193135438 on Build GRCh38
rs773172494 Chr.1: 193135468-193135468 on Build GRCh38
rs768821497 Chr.1: 193135604-193135604 on Build GRCh38
rs563211268 Chr.1: 193135275-193135275 on Build GRCh38
rs775574593 Chr.1: 193135446-193135446 on Build GRCh38
rs745334057 Chr.1: 193135625-193135627 on Build GRCh38
rs952579366 Chr.1: 193135417-193135417 on Build GRCh38
rs965169255 Chr.1: 193135373-193135373 on Build GRCh38
rs374021222 Chr.1: 193135533-193135533 on Build GRCh38
rs747397191 Chr.1: 193135376-193135376 on Build GRCh38
rs1021234265 Chr.1: 193135500-193135500 on Build GRCh38
rs769833729 Chr.1: 193135419-193135419 on Build GRCh38
rs773739963 Chr.1: 193135608-193135613 on Build GRCh38
rs757680253 Chr.1: 193135530-193135530 on Build GRCh38
rs768873520 Chr.1: 193135467-193135467 on Build GRCh38
rs752356591 Chr.1: 193135384-193135384 on Build GRCh38
rs772756239 Chr.1: 193135247-193135247 on Build GRCh38
rs201766013 Chr.1: 193135375-193135375 on Build GRCh38
rs776500275 Chr.1: 193135628-193135628 on Build GRCh38
rs374834265 Chr.1: 193135416-193135416 on Build GRCh38
rs142509713 Chr.1: 193135654-193135654 on Build GRCh38
rs750182992 Chr.1: 193135345-193135345 on Build GRCh38
rs375460045 Chr.1: 193135619-193135619 on Build GRCh38
rs1005361595 Chr.1: 193135328-193135328 on Build GRCh38
rs894581514 Chr.1: 193135221-193135221 on Build GRCh38
rs75227966 Chr.1: 193135385-193135385 on Build GRCh38
rs763780849 Chr.1: 193135520-193135520 on Build GRCh38
rs749344710 Chr.1: 193135595-193135595 on Build GRCh38
rs780017801 Chr.1: 193135560-193135560 on Build GRCh38
rs546046048 Chr.1: 193135267-193135267 on Build GRCh38
rs754491728 Chr.1: 193135370-193135370 on Build GRCh38
rs746368285 Chr.1: 193135538-193135538 on Build GRCh38
rs747337987 Chr.1: 193135395-193135395 on Build GRCh38
rs369542555 Chr.1: 193135578-193135578 on Build GRCh38
rs756584058 Chr.1: 193135548-193135548 on Build GRCh38
rs767218233 Chr.1: 193135341-193135341 on Build GRCh38
rs759510175 Chr.1: 193135497-193135497 on Build GRCh38
rs774763324 Chr.1: 193135615-193135615 on Build GRCh38
rs748880183 Chr.1: 193135523-193135526 on Build GRCh38
rs764670050 Chr.1: 193135498-193135498 on Build GRCh38
rs908205344 Chr.1: 193135624-193135624 on Build GRCh38
rs753881821 Chr.1: 193135495-193135495 on Build GRCh38
rs1011536523 Chr.1: 193135231-193135231 on Build GRCh38
rs770561055 Chr.1: 193135600-193135601 on Build GRCh38
rs765308159 Chr.1: 193135647-193135647 on Build GRCh38
rs758020763 Chr.1: 193135511-193135511 on Build GRCh38
rs369254776 Chr.1: 193135399-193135399 on Build GRCh38
rs760287746 Chr.1: 193135390-193135390 on Build GRCh38
rs547890731 Chr.1: 193135648-193135648 on Build GRCh38
rs766250348 Chr.1: 193135492-193135492 on Build GRCh38
rs777550706 Chr.1: 193135515-193135519 on Build GRCh38
rs751195516 Chr.1: 193135524-193135524 on Build GRCh38
rs923317075 Chr.1: 193135650-193135650 on Build GRCh38
rs901840902 Chr.1: 193135226-193135226 on Build GRCh38
rs763095800 Chr.1: 193135447-193135447 on Build GRCh38
rs1016696958 Chr.1: 193135334-193135334 on Build GRCh38
rs933936281 Chr.1: 193135236-193135236 on Build GRCh38
rs771258964 Chr.1: 193135398-193135398 on Build GRCh38
rs976186333 Chr.1: 193135629-193135629 on Build GRCh38
rs766379008 Chr.1: 193135368-193135368 on Build GRCh38
rs886988441 Chr.1: 193135317-193135317 on Build GRCh38
rs527640228 Chr.1: 193135501-193135501 on Build GRCh38
rs868045207 Chr.1: 193135421-193135421 on Build GRCh38
rs960410437 Chr.1: 193135456-193135456 on Build GRCh38
rs760710336 Chr.1: 193135353-193135353 on Build GRCh38
rs769052041 Chr.1: 193135557-193135557 on Build GRCh38
rs369812032 Chr.1: 193135499-193135499 on Build GRCh38
rs999173509 Chr.1: 193135337-193135337 on Build GRCh38
rs778135669 Chr.1: 193135388-193135388 on Build GRCh38

View Full Product Details