Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 193178607-193179127 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B3GALT2
Entrez Gene ID
8707
Gene Name
beta-1,3-galactosyltransferase 2
Gene Aliases
BETA3GALT2, GLCT2, beta3Gal-T2
UniGene
Hs.518834
Gene Chromosome Location
Chr.1: 193178730-193186613 on Build GRCh38
Transcripts
NM_003783.3 NM_024529.4

Gene Symbol
CDC73
Entrez Gene ID
79577
Gene Name
cell division cycle 73
Gene Aliases
C1orf28, FIHP, HPTJT, HRPT1, HRPT2, HYX
UniGene
Hs.378996
Gene Chromosome Location
Chr.1: 193121958-193254815 on Build GRCh38
Transcripts
NM_003783.3 NM_024529.4

SNP Detail

SNP ID Chromosome Location
rs386638049 Chr.1: 193178685-193178690 on Build GRCh38
rs559355292 Chr.1: 193178637-193178637 on Build GRCh38
rs1046303954 Chr.1: 193178959-193178959 on Build GRCh38
rs907440867 Chr.1: 193178648-193178648 on Build GRCh38
rs892773447 Chr.1: 193178976-193178976 on Build GRCh38
rs183779981 Chr.1: 193178955-193178955 on Build GRCh38
rs576033459 Chr.1: 193178943-193178943 on Build GRCh38
rs903209750 Chr.1: 193178882-193178882 on Build GRCh38
rs139353908 Chr.1: 193178729-193178729 on Build GRCh38
rs144170419 Chr.1: 193178940-193178940 on Build GRCh38
rs563162406 Chr.1: 193178718-193178718 on Build GRCh38
rs1053384247 Chr.1: 193178965-193178965 on Build GRCh38
rs539380780 Chr.1: 193178813-193178813 on Build GRCh38
rs572980874 Chr.1: 193179008-193179011 on Build GRCh38
rs1008008106 Chr.1: 193179025-193179025 on Build GRCh38
rs750808634 Chr.1: 193178884-193178884 on Build GRCh38
rs1055980890 Chr.1: 193179071-193179071 on Build GRCh38
rs975277382 Chr.1: 193178914-193178914 on Build GRCh38
rs191308346 Chr.1: 193178702-193178702 on Build GRCh38
rs1021623916 Chr.1: 193178881-193178881 on Build GRCh38
rs983761749 Chr.1: 193178657-193178657 on Build GRCh38
rs938870943 Chr.1: 193178747-193178747 on Build GRCh38
rs1028085799 Chr.1: 193178735-193178735 on Build GRCh38
rs919237475 Chr.1: 193178930-193178930 on Build GRCh38
rs77971522 Chr.1: 193178945-193178945 on Build GRCh38
rs894583086 Chr.1: 193178764-193178764 on Build GRCh38
rs374173189 Chr.1: 193178961-193178964 on Build GRCh38
rs376457037 Chr.1: 193178936-193178936 on Build GRCh38
rs961255496 Chr.1: 193179068-193179068 on Build GRCh38
rs552475492 Chr.1: 193179035-193179035 on Build GRCh38
rs948837934 Chr.1: 193178957-193178961 on Build GRCh38
rs965555194 Chr.1: 193178899-193178899 on Build GRCh38
rs555301765 Chr.1: 193179044-193179044 on Build GRCh38
rs908212560 Chr.1: 193178964-193178964 on Build GRCh38
rs972690998 Chr.1: 193179048-193179048 on Build GRCh38
rs10921325 Chr.1: 193178690-193178690 on Build GRCh38
rs1028031851 Chr.1: 193179013-193179013 on Build GRCh38
rs10921324 Chr.1: 193178685-193178685 on Build GRCh38
rs189295190 Chr.1: 193179056-193179056 on Build GRCh38
rs35543557 Chr.1: 193178778-193178779 on Build GRCh38
rs919845345 Chr.1: 193179089-193179089 on Build GRCh38
rs758722148 Chr.1: 193178933-193178933 on Build GRCh38
rs972701676 Chr.1: 193178910-193178910 on Build GRCh38
rs939722526 Chr.1: 193179020-193179020 on Build GRCh38
rs762096812 Chr.1: 193178754-193178754 on Build GRCh38
rs765382766 Chr.1: 193178801-193178801 on Build GRCh38
rs928905672 Chr.1: 193179049-193179049 on Build GRCh38
rs953769127 Chr.1: 193179019-193179019 on Build GRCh38
rs961578726 Chr.1: 193178903-193178903 on Build GRCh38
rs1019564226 Chr.1: 193178888-193178888 on Build GRCh38
rs1049093237 Chr.1: 193178940-193178940 on Build GRCh38
rs952448305 Chr.1: 193178937-193178937 on Build GRCh38

View Full Product Details