Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 193181547-193182045 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B3GALT2
Entrez Gene ID
8707
Gene Name
beta-1,3-galactosyltransferase 2
Gene Aliases
BETA3GALT2, GLCT2, beta3Gal-T2
UniGene
Hs.518834
Gene Chromosome Location
Chr.1: 193178730-193186613 on Build GRCh38
Transcripts
NM_003783.3 NM_024529.4

Gene Symbol
CDC73
Entrez Gene ID
79577
Gene Name
cell division cycle 73
Gene Aliases
C1orf28, FIHP, HPTJT, HRPT1, HRPT2, HYX
UniGene
Hs.378996
Gene Chromosome Location
Chr.1: 193121958-193254815 on Build GRCh38
Transcripts
NM_003783.3 NM_024529.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
901397 c.10T>C p.W4R Chr.1: 193181553-193181553 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs772142160 Chr.1: 193181575-193181575 on Build GRCh38
rs529330580 Chr.1: 193181829-193181829 on Build GRCh38
rs551683166 Chr.1: 193181927-193181927 on Build GRCh38
rs879273787 Chr.1: 193181630-193181630 on Build GRCh38
rs960771144 Chr.1: 193181844-193181844 on Build GRCh38
rs901244275 Chr.1: 193181622-193181622 on Build GRCh38
rs540099698 Chr.1: 193181963-193181963 on Build GRCh38
rs1038546040 Chr.1: 193181988-193181988 on Build GRCh38
rs918484629 Chr.1: 193181615-193181615 on Build GRCh38
rs1040096688 Chr.1: 193181882-193181882 on Build GRCh38
rs750379702 Chr.1: 193181970-193181970 on Build GRCh38
rs368420383 Chr.1: 193181874-193181874 on Build GRCh38
rs759771567 Chr.1: 193181794-193181794 on Build GRCh38
rs773217355 Chr.1: 193181577-193181577 on Build GRCh38
rs552103345 Chr.1: 193181625-193181625 on Build GRCh38
rs559252088 Chr.1: 193181669-193181669 on Build GRCh38
rs577961649 Chr.1: 193181701-193181701 on Build GRCh38
rs760629619 Chr.1: 193181619-193181619 on Build GRCh38
rs571192811 Chr.1: 193181973-193181973 on Build GRCh38
rs10494675 Chr.1: 193181743-193181743 on Build GRCh38
rs776696042 Chr.1: 193181590-193181590 on Build GRCh38
rs187364519 Chr.1: 193181846-193181846 on Build GRCh38
rs947874761 Chr.1: 193181631-193181631 on Build GRCh38
rs901198071 Chr.1: 193181884-193181884 on Build GRCh38
rs942697963 Chr.1: 193181585-193181585 on Build GRCh38
rs1026049776 Chr.1: 193181841-193181841 on Build GRCh38
rs763228772 Chr.1: 193181719-193181719 on Build GRCh38
rs796202434 Chr.1: 193181803-193181803 on Build GRCh38
rs998841468 Chr.1: 193181734-193181734 on Build GRCh38
rs774791260 Chr.1: 193181793-193181793 on Build GRCh38
rs979272250 Chr.1: 193181969-193181969 on Build GRCh38
rs943333463 Chr.1: 193181864-193181864 on Build GRCh38
rs913706297 Chr.1: 193181812-193181812 on Build GRCh38
rs762865701 Chr.1: 193181603-193181603 on Build GRCh38
rs901533232 Chr.1: 193181730-193181730 on Build GRCh38
rs979303575 Chr.1: 193181840-193181840 on Build GRCh38
rs756145288 Chr.1: 193181982-193181982 on Build GRCh38
rs969045596 Chr.1: 193181698-193181698 on Build GRCh38
rs765399636 Chr.1: 193181589-193181590 on Build GRCh38
rs887295565 Chr.1: 193181666-193181667 on Build GRCh38
rs771225207 Chr.1: 193181586-193181586 on Build GRCh38
rs563808734 Chr.1: 193181763-193181763 on Build GRCh38
rs962783222 Chr.1: 193181814-193181814 on Build GRCh38
rs375044037 Chr.1: 193181641-193181641 on Build GRCh38
rs747218269 Chr.1: 193181583-193181583 on Build GRCh38
rs750229437 Chr.1: 193181771-193181772 on Build GRCh38
rs935056916 Chr.1: 193182014-193182014 on Build GRCh38
rs1052518422 Chr.1: 193181727-193181727 on Build GRCh38
rs1017151064 Chr.1: 193181691-193181691 on Build GRCh38
rs137952056 Chr.1: 193181667-193181667 on Build GRCh38
rs1047147779 Chr.1: 193181932-193181932 on Build GRCh38
rs889918572 Chr.1: 193181782-193181782 on Build GRCh38
rs528426223 Chr.1: 193181905-193181905 on Build GRCh38
rs773265990 Chr.1: 193181699-193181699 on Build GRCh38
rs994294885 Chr.1: 193181819-193181819 on Build GRCh38
rs559563774 Chr.1: 193181896-193181896 on Build GRCh38

View Full Product Details