Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 193185939-193186448 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B3GALT2
Entrez Gene ID
8707
Gene Name
beta-1,3-galactosyltransferase 2
Gene Aliases
BETA3GALT2, GLCT2, beta3Gal-T2
UniGene
Hs.518834
Gene Chromosome Location
Chr.1: 193178730-193186613 on Build GRCh38
Transcripts
NM_003783.3 NM_024529.4

Gene Symbol
CDC73
Entrez Gene ID
79577
Gene Name
cell division cycle 73
Gene Aliases
C1orf28, FIHP, HPTJT, HRPT1, HRPT2, HYX
UniGene
Hs.378996
Gene Chromosome Location
Chr.1: 193121958-193254815 on Build GRCh38
Transcripts
NM_003783.3 NM_024529.4

SNP Detail

SNP ID Chromosome Location
rs747443534 Chr.1: 193186287-193186287 on Build GRCh38
rs940865600 Chr.1: 193186115-193186115 on Build GRCh38
rs34137906 Chr.1: 193186149-193186150 on Build GRCh38
rs868747050 Chr.1: 193186118-193186118 on Build GRCh38
rs541925867 Chr.1: 193186316-193186316 on Build GRCh38
rs374975381 Chr.1: 193186172-193186172 on Build GRCh38
rs553644947 Chr.1: 193186080-193186080 on Build GRCh38
rs181462524 Chr.1: 193186329-193186329 on Build GRCh38
rs752682761 Chr.1: 193186368-193186368 on Build GRCh38
rs999106392 Chr.1: 193186260-193186260 on Build GRCh38
rs993454276 Chr.1: 193186314-193186314 on Build GRCh38
rs576080026 Chr.1: 193186294-193186294 on Build GRCh38
rs894500950 Chr.1: 193186187-193186187 on Build GRCh38
rs3795616 Chr.1: 193186398-193186398 on Build GRCh38
rs902982050 Chr.1: 193186256-193186256 on Build GRCh38
rs762359755 Chr.1: 193186346-193186346 on Build GRCh38
rs143478275 Chr.1: 193186094-193186094 on Build GRCh38
rs1043369151 Chr.1: 193186254-193186254 on Build GRCh38
rs962213698 Chr.1: 193186351-193186351 on Build GRCh38
rs751377674 Chr.1: 193186010-193186010 on Build GRCh38
rs985487516 Chr.1: 193186069-193186069 on Build GRCh38
rs953648314 Chr.1: 193186049-193186049 on Build GRCh38
rs1052774472 Chr.1: 193186404-193186404 on Build GRCh38
rs545493946 Chr.1: 193186268-193186268 on Build GRCh38
rs376220651 Chr.1: 193186221-193186221 on Build GRCh38
rs972207782 Chr.1: 193186386-193186386 on Build GRCh38
rs116733971 Chr.1: 193186286-193186286 on Build GRCh38
rs11545694 Chr.1: 193186074-193186074 on Build GRCh38
rs1046019459 Chr.1: 193186028-193186028 on Build GRCh38
rs34851524 Chr.1: 193186395-193186396 on Build GRCh38
rs1036420189 Chr.1: 193186116-193186116 on Build GRCh38
rs928723262 Chr.1: 193186137-193186137 on Build GRCh38
rs756068310 Chr.1: 193186380-193186380 on Build GRCh38
rs928818068 Chr.1: 193186390-193186390 on Build GRCh38
rs938773984 Chr.1: 193186146-193186146 on Build GRCh38
rs1055792185 Chr.1: 193186164-193186164 on Build GRCh38
rs147589997 Chr.1: 193186337-193186337 on Build GRCh38
rs906834560 Chr.1: 193186105-193186105 on Build GRCh38
rs916080140 Chr.1: 193185984-193185984 on Build GRCh38
rs947321591 Chr.1: 193186242-193186242 on Build GRCh38
rs754730068 Chr.1: 193186107-193186107 on Build GRCh38
rs892867984 Chr.1: 193186110-193186110 on Build GRCh38
rs948928631 Chr.1: 193185995-193185995 on Build GRCh38
rs1027944036 Chr.1: 193186327-193186327 on Build GRCh38
rs909346377 Chr.1: 193186078-193186078 on Build GRCh38
rs952136907 Chr.1: 193186317-193186317 on Build GRCh38

View Full Product Details