Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 196825335-196825874 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFHR1
Entrez Gene ID
3078
Gene Name
complement factor H related 1
Gene Aliases
CFHL, CFHL1, CFHL1P, CFHR1P, FHR1, H36-1, H36-2, HFL1, HFL2
UniGene
Hs.575869
Gene Chromosome Location
Chr.1: 196819730-196832189 on Build GRCh38
Transcripts
NM_002113.2 XM_011509457.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5528593 c.185C>T p.P62L Chr.1: 196825603-196825603 on Build GRCh38
6627505 c.65T>G p.F22C Chr.1: 196825483-196825483 on Build GRCh38
3480840 c.70G>A p.D24N Chr.1: 196825488-196825488 on Build GRCh38
5440010 c.194C>A p.S65* Chr.1: 196825612-196825612 on Build GRCh38
3480843 c.106G>A p.E36K Chr.1: 196825524-196825524 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764825235 Chr.1: 196825472-196825472 on Build GRCh38
rs760685217 Chr.1: 196825452-196825452 on Build GRCh38
rs373146352 Chr.1: 196825470-196825470 on Build GRCh38
rs771933119 Chr.1: 196825437-196825437 on Build GRCh38
rs764441881 Chr.1: 196825481-196825481 on Build GRCh38
rs409549 Chr.1: 196825779-196825779 on Build GRCh38
rs41310895 Chr.1: 196825517-196825517 on Build GRCh38
rs766844121 Chr.1: 196825671-196825671 on Build GRCh38
rs769231948 Chr.1: 196825682-196825682 on Build GRCh38
rs141741322 Chr.1: 196825735-196825735 on Build GRCh38
rs41305054 Chr.1: 196825613-196825613 on Build GRCh38
rs442759 Chr.1: 196825389-196825389 on Build GRCh38
rs754574977 Chr.1: 196825654-196825654 on Build GRCh38
rs41315553 Chr.1: 196825550-196825550 on Build GRCh38
rs758750227 Chr.1: 196825480-196825480 on Build GRCh38
rs773340802 Chr.1: 196825438-196825438 on Build GRCh38
rs761214027 Chr.1: 196825720-196825720 on Build GRCh38
rs199672226 Chr.1: 196825635-196825635 on Build GRCh38
rs754046162 Chr.1: 196825623-196825623 on Build GRCh38
rs113307454 Chr.1: 196825463-196825463 on Build GRCh38
rs372075 Chr.1: 196825737-196825737 on Build GRCh38
rs767130763 Chr.1: 196825757-196825757 on Build GRCh38
rs41308387 Chr.1: 196825462-196825462 on Build GRCh38
rs750501937 Chr.1: 196825510-196825510 on Build GRCh38
rs770780185 Chr.1: 196825651-196825651 on Build GRCh38
rs766024107 Chr.1: 196825457-196825457 on Build GRCh38
rs41300847 Chr.1: 196825406-196825406 on Build GRCh38
rs762565447 Chr.1: 196825562-196825562 on Build GRCh38
rs534789337 Chr.1: 196825495-196825495 on Build GRCh38
rs376204016 Chr.1: 196825700-196825700 on Build GRCh38
rs41313920 Chr.1: 196825684-196825684 on Build GRCh38
rs777676159 Chr.1: 196825540-196825540 on Build GRCh38
rs959385608 Chr.1: 196825571-196825571 on Build GRCh38
rs147383478 Chr.1: 196825596-196825596 on Build GRCh38
rs768330868 Chr.1: 196825697-196825697 on Build GRCh38
rs775075700 Chr.1: 196825560-196825560 on Build GRCh38
rs750957640 Chr.1: 196825710-196825710 on Build GRCh38
rs41308385 Chr.1: 196825661-196825661 on Build GRCh38
rs374490965 Chr.1: 196825717-196825717 on Build GRCh38
rs138970064 Chr.1: 196825551-196825551 on Build GRCh38
rs531072224 Chr.1: 196825839-196825839 on Build GRCh38
rs576996003 Chr.1: 196825729-196825729 on Build GRCh38
rs772057910 Chr.1: 196825534-196825534 on Build GRCh38
rs41300845 Chr.1: 196825477-196825477 on Build GRCh38
rs780177916 Chr.1: 196825619-196825619 on Build GRCh38
rs773313542 Chr.1: 196825522-196825522 on Build GRCh38
rs776254611 Chr.1: 196825459-196825459 on Build GRCh38
rs548589815 Chr.1: 196825379-196825379 on Build GRCh38
rs41310897 Chr.1: 196825512-196825512 on Build GRCh38
rs146250787 Chr.1: 196825798-196825798 on Build GRCh38
rs576365193 Chr.1: 196825573-196825573 on Build GRCh38
rs758984798 Chr.1: 196825557-196825557 on Build GRCh38
rs146147352 Chr.1: 196825548-196825548 on Build GRCh38
rs747898706 Chr.1: 196825436-196825436 on Build GRCh38
rs371125562 Chr.1: 196825712-196825712 on Build GRCh38
rs762971807 Chr.1: 196825580-196825581 on Build GRCh38
rs557395695 Chr.1: 196825559-196825559 on Build GRCh38
rs41313922 Chr.1: 196825446-196825446 on Build GRCh38
rs775742200 Chr.1: 196825428-196825428 on Build GRCh38
rs76655840 Chr.1: 196825523-196825523 on Build GRCh38
rs762811051 Chr.1: 196825373-196825373 on Build GRCh38
rs552308882 Chr.1: 196825483-196825483 on Build GRCh38
rs148561849 Chr.1: 196825580-196825580 on Build GRCh38
rs780151716 Chr.1: 196825522-196825522 on Build GRCh38
rs537358906 Chr.1: 196825624-196825624 on Build GRCh38
rs766129586 Chr.1: 196825497-196825497 on Build GRCh38
rs773928939 Chr.1: 196825699-196825699 on Build GRCh38
rs541375945 Chr.1: 196825780-196825780 on Build GRCh38
rs865905058 Chr.1: 196825398-196825398 on Build GRCh38
rs745817071 Chr.1: 196825665-196825665 on Build GRCh38
rs764492771 Chr.1: 196825572-196825572 on Build GRCh38
rs200609129 Chr.1: 196825484-196825484 on Build GRCh38
rs41302141 Chr.1: 196825503-196825503 on Build GRCh38
rs762401875 Chr.1: 196825685-196825685 on Build GRCh38

View Full Product Details