Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 197905666-197906177 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf53
Entrez Gene ID
388722
Gene Name
chromosome 1 open reading frame 53
Gene Aliases
-
UniGene
Hs.61329
Gene Chromosome Location
Chr.1: 197902552-197907367 on Build GRCh38
Transcripts
NM_001024594.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1583913 c.356C>T p.A119V Chr.1: 197905887-197905887 on Build GRCh38
4026698 c.301T>C p.Y101H Chr.1: 197905832-197905832 on Build GRCh38
6615281 c.308T>C p.V103A Chr.1: 197905839-197905839 on Build GRCh38
7322616 c.318G>C p.Q106H Chr.1: 197905849-197905849 on Build GRCh38
678291 c.335G>A p.R112K Chr.1: 197905866-197905866 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1038477940 Chr.1: 197906025-197906025 on Build GRCh38
rs920216903 Chr.1: 197905726-197905726 on Build GRCh38
rs377598858 Chr.1: 197905906-197905906 on Build GRCh38
rs771803618 Chr.1: 197905847-197905847 on Build GRCh38
rs556555241 Chr.1: 197905916-197905916 on Build GRCh38
rs759897566 Chr.1: 197905763-197905763 on Build GRCh38
rs749857503 Chr.1: 197906138-197906138 on Build GRCh38
rs755034905 Chr.1: 197905914-197905914 on Build GRCh38
rs570699575 Chr.1: 197905969-197905971 on Build GRCh38
rs760091765 Chr.1: 197905873-197905873 on Build GRCh38
rs183144313 Chr.1: 197906119-197906119 on Build GRCh38
rs756067434 Chr.1: 197905805-197905805 on Build GRCh38
rs747722650 Chr.1: 197905836-197905836 on Build GRCh38
rs747994250 Chr.1: 197905920-197905920 on Build GRCh38
rs138123930 Chr.1: 197905972-197905972 on Build GRCh38
rs981560047 Chr.1: 197905997-197905997 on Build GRCh38
rs765384914 Chr.1: 197905775-197905775 on Build GRCh38
rs1040500033 Chr.1: 197905853-197905853 on Build GRCh38
rs150305479 Chr.1: 197906146-197906146 on Build GRCh38
rs896600824 Chr.1: 197906128-197906128 on Build GRCh38
rs933664130 Chr.1: 197905951-197905951 on Build GRCh38
rs752847353 Chr.1: 197905779-197905779 on Build GRCh38
rs780176821 Chr.1: 197905811-197905811 on Build GRCh38
rs868021267 Chr.1: 197905877-197905877 on Build GRCh38
rs771361126 Chr.1: 197905860-197905860 on Build GRCh38
rs753816568 Chr.1: 197905908-197905908 on Build GRCh38
rs1051224664 Chr.1: 197905961-197905961 on Build GRCh38
rs975162801 Chr.1: 197905934-197905934 on Build GRCh38
rs2270763 Chr.1: 197905819-197905819 on Build GRCh38
rs920079646 Chr.1: 197906136-197906136 on Build GRCh38
rs757944745 Chr.1: 197905921-197905921 on Build GRCh38
rs181264157 Chr.1: 197905746-197905746 on Build GRCh38
rs747046238 Chr.1: 197906139-197906139 on Build GRCh38
rs2270762 Chr.1: 197905752-197905752 on Build GRCh38
rs777481616 Chr.1: 197905924-197905924 on Build GRCh38
rs1011434486 Chr.1: 197905953-197905953 on Build GRCh38
rs766893516 Chr.1: 197905749-197905749 on Build GRCh38
rs527920265 Chr.1: 197906052-197906052 on Build GRCh38
rs772544322 Chr.1: 197905857-197905857 on Build GRCh38
rs757420792 Chr.1: 197905904-197905904 on Build GRCh38
rs967725031 Chr.1: 197905915-197905915 on Build GRCh38
rs186521541 Chr.1: 197905874-197905874 on Build GRCh38
rs777685354 Chr.1: 197905789-197905789 on Build GRCh38
rs372599540 Chr.1: 197905887-197905887 on Build GRCh38
rs545490771 Chr.1: 197905977-197905977 on Build GRCh38
rs755243366 Chr.1: 197905950-197905950 on Build GRCh38
rs369143435 Chr.1: 197905933-197905933 on Build GRCh38
rs536041809 Chr.1: 197905796-197905796 on Build GRCh38
rs763598456 Chr.1: 197905811-197905812 on Build GRCh38
rs200185591 Chr.1: 197905833-197905833 on Build GRCh38
rs758716780 Chr.1: 197905788-197905788 on Build GRCh38
rs777135437 Chr.1: 197905866-197905866 on Build GRCh38
rs771403673 Chr.1: 197905888-197905888 on Build GRCh38
rs992103168 Chr.1: 197906104-197906104 on Build GRCh38
rs374179524 Chr.1: 197906035-197906035 on Build GRCh38
rs775964919 Chr.1: 197905882-197905882 on Build GRCh38
rs1013572215 Chr.1: 197906145-197906145 on Build GRCh38
rs746108983 Chr.1: 197905950-197905950 on Build GRCh38
rs977242433 Chr.1: 197905919-197905919 on Build GRCh38
rs749731186 Chr.1: 197905862-197905862 on Build GRCh38
rs191737701 Chr.1: 197906034-197906034 on Build GRCh38
rs768588490 Chr.1: 197905821-197905821 on Build GRCh38

View Full Product Details