Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 200815443-200815984 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAMSAP2
Entrez Gene ID
23271
Gene Name
calmodulin regulated spectrin associated protein family member 2
Gene Aliases
CAMSAP1L1
UniGene
Hs.23585
Gene Chromosome Location
Chr.1: 200738905-200860707 on Build GRCh38
Transcripts
NM_001297707.1 NM_001297708.1 NM_203459.2 XM_005245041.3 XM_017000799.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7084148 c.622G>A p.V208I Chr.1: 200815621-200815621 on Build GRCh38
4503139 c.631C>T p.P211S Chr.1: 200815630-200815630 on Build GRCh38
678234 c.645G>A p.K215K Chr.1: 200815644-200815644 on Build GRCh38
4503138 c.631C>T p.P211S Chr.1: 200815630-200815630 on Build GRCh38
3400180 c.616C>G p.H206D Chr.1: 200815615-200815615 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs940579871 Chr.1: 200815750-200815750 on Build GRCh38
rs368441570 Chr.1: 200815683-200815683 on Build GRCh38
rs184036699 Chr.1: 200815765-200815765 on Build GRCh38
rs146826954 Chr.1: 200815820-200815820 on Build GRCh38
rs768015579 Chr.1: 200815533-200815533 on Build GRCh38
rs771120105 Chr.1: 200815579-200815579 on Build GRCh38
rs773563102 Chr.1: 200815571-200815571 on Build GRCh38
rs746471163 Chr.1: 200815905-200815905 on Build GRCh38
rs767625939 Chr.1: 200815634-200815634 on Build GRCh38
rs979423075 Chr.1: 200815604-200815604 on Build GRCh38
rs1020846803 Chr.1: 200815843-200815843 on Build GRCh38
rs1018407639 Chr.1: 200815859-200815859 on Build GRCh38
rs748997542 Chr.1: 200815521-200815521 on Build GRCh38
rs540408785 Chr.1: 200815957-200815957 on Build GRCh38
rs751244644 Chr.1: 200815518-200815518 on Build GRCh38
rs143566886 Chr.1: 200815718-200815718 on Build GRCh38
rs191859329 Chr.1: 200815741-200815741 on Build GRCh38
rs71635512 Chr.1: 200815589-200815589 on Build GRCh38
rs199849928 Chr.1: 200815674-200815674 on Build GRCh38
rs758873321 Chr.1: 200815669-200815669 on Build GRCh38
rs923908089 Chr.1: 200815903-200815906 on Build GRCh38
rs936988598 Chr.1: 200815777-200815777 on Build GRCh38
rs758282962 Chr.1: 200815848-200815850 on Build GRCh38
rs756018315 Chr.1: 200815641-200815641 on Build GRCh38
rs772627438 Chr.1: 200815553-200815553 on Build GRCh38
rs968104461 Chr.1: 200815867-200815867 on Build GRCh38
rs780732224 Chr.1: 200815524-200815525 on Build GRCh38
rs759832629 Chr.1: 200815593-200815593 on Build GRCh38
rs1000819677 Chr.1: 200815894-200815894 on Build GRCh38
rs1035969634 Chr.1: 200815763-200815767 on Build GRCh38
rs748055904 Chr.1: 200815812-200815812 on Build GRCh38
rs895541434 Chr.1: 200815809-200815809 on Build GRCh38
rs776667631 Chr.1: 200815582-200815582 on Build GRCh38
rs753368473 Chr.1: 200815656-200815656 on Build GRCh38
rs934117008 Chr.1: 200815725-200815725 on Build GRCh38
rs778275266 Chr.1: 200815667-200815667 on Build GRCh38
rs752833360 Chr.1: 200815618-200815618 on Build GRCh38
rs899114284 Chr.1: 200815776-200815776 on Build GRCh38
rs879090497 Chr.1: 200815734-200815738 on Build GRCh38
rs556114046 Chr.1: 200815734-200815734 on Build GRCh38
rs367624646 Chr.1: 200815543-200815543 on Build GRCh38
rs754595262 Chr.1: 200815523-200815525 on Build GRCh38
rs374737109 Chr.1: 200815637-200815637 on Build GRCh38
rs778494026 Chr.1: 200815534-200815534 on Build GRCh38
rs922688360 Chr.1: 200815686-200815686 on Build GRCh38
rs1055491312 Chr.1: 200815787-200815787 on Build GRCh38
rs777575100 Chr.1: 200815695-200815696 on Build GRCh38
rs573114943 Chr.1: 200815868-200815868 on Build GRCh38
rs994403094 Chr.1: 200815767-200815767 on Build GRCh38
rs34505744 Chr.1: 200815632-200815632 on Build GRCh38
rs946731576 Chr.1: 200815546-200815546 on Build GRCh38
rs754676321 Chr.1: 200815666-200815666 on Build GRCh38
rs1020479077 Chr.1: 200815486-200815486 on Build GRCh38
rs756142462 Chr.1: 200815683-200815684 on Build GRCh38
rs1005882563 Chr.1: 200815738-200815738 on Build GRCh38
rs964304044 Chr.1: 200815926-200815926 on Build GRCh38
rs41308391 Chr.1: 200815565-200815565 on Build GRCh38
rs765162660 Chr.1: 200815599-200815599 on Build GRCh38
rs752133732 Chr.1: 200815646-200815649 on Build GRCh38
rs968004384 Chr.1: 200815490-200815490 on Build GRCh38
rs778302924 Chr.1: 200815672-200815672 on Build GRCh38
rs777057423 Chr.1: 200815692-200815692 on Build GRCh38
rs955530791 Chr.1: 200815830-200815830 on Build GRCh38
rs923876866 Chr.1: 200815622-200815622 on Build GRCh38
rs747762716 Chr.1: 200815668-200815668 on Build GRCh38
rs761068698 Chr.1: 200815575-200815575 on Build GRCh38
rs779933897 Chr.1: 200815650-200815650 on Build GRCh38
rs1033218975 Chr.1: 200815911-200815911 on Build GRCh38
rs188621588 Chr.1: 200815850-200815850 on Build GRCh38

View Full Product Details