Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 200832325-200832841 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAMSAP2
Entrez Gene ID
23271
Gene Name
calmodulin regulated spectrin associated protein family member 2
Gene Aliases
CAMSAP1L1
UniGene
Hs.23585
Gene Chromosome Location
Chr.1: 200738905-200860707 on Build GRCh38
Transcripts
NM_001297707.1 NM_001297708.1 NM_203459.2 XM_005245041.3 XM_017000799.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5770408 c.857G>A p.S286N Chr.1: 200832742-200832742 on Build GRCh38
7171005 c.831G>C p.L277F Chr.1: 200832716-200832716 on Build GRCh38
5770407 c.824G>A p.S275N Chr.1: 200832742-200832742 on Build GRCh38
5735791 c.883G>T p.E295* Chr.1: 200832768-200832768 on Build GRCh38
901989 c.918A>C p.S306S Chr.1: 200832836-200832836 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs897412963 Chr.1: 200832691-200832691 on Build GRCh38
rs374858405 Chr.1: 200832696-200832696 on Build GRCh38
rs897146170 Chr.1: 200832385-200832385 on Build GRCh38
rs1033598274 Chr.1: 200832397-200832397 on Build GRCh38
rs973245116 Chr.1: 200832437-200832438 on Build GRCh38
rs1021020623 Chr.1: 200832353-200832353 on Build GRCh38
rs41306231 Chr.1: 200832807-200832807 on Build GRCh38
rs1041686719 Chr.1: 200832635-200832635 on Build GRCh38
rs780573739 Chr.1: 200832377-200832377 on Build GRCh38
rs770752759 Chr.1: 200832689-200832689 on Build GRCh38
rs559577750 Chr.1: 200832399-200832399 on Build GRCh38
rs769242505 Chr.1: 200832378-200832378 on Build GRCh38
rs983644038 Chr.1: 200832486-200832487 on Build GRCh38
rs1003563360 Chr.1: 200832711-200832711 on Build GRCh38
rs984565069 Chr.1: 200832439-200832439 on Build GRCh38
rs191849409 Chr.1: 200832511-200832511 on Build GRCh38
rs116370820 Chr.1: 200832485-200832485 on Build GRCh38
rs187692622 Chr.1: 200832494-200832494 on Build GRCh38
rs928606900 Chr.1: 200832505-200832505 on Build GRCh38
rs757699429 Chr.1: 200832350-200832350 on Build GRCh38
rs1026991761 Chr.1: 200832434-200832434 on Build GRCh38
rs111264726 Chr.1: 200832488-200832488 on Build GRCh38
rs756447322 Chr.1: 200832362-200832362 on Build GRCh38
rs776583920 Chr.1: 200832690-200832690 on Build GRCh38
rs775277291 Chr.1: 200832728-200832728 on Build GRCh38
rs952800315 Chr.1: 200832438-200832438 on Build GRCh38
rs914302703 Chr.1: 200832555-200832555 on Build GRCh38
rs201192648 Chr.1: 200832352-200832352 on Build GRCh38
rs749466550 Chr.1: 200832380-200832380 on Build GRCh38
rs750727990 Chr.1: 200832356-200832356 on Build GRCh38
rs1049527437 Chr.1: 200832618-200832618 on Build GRCh38
rs375906378 Chr.1: 200832721-200832721 on Build GRCh38
rs1874080 Chr.1: 200832493-200832493 on Build GRCh38
rs776492129 Chr.1: 200832577-200832577 on Build GRCh38
rs779132822 Chr.1: 200832676-200832676 on Build GRCh38
rs932877830 Chr.1: 200832515-200832515 on Build GRCh38
rs748279962 Chr.1: 200832684-200832684 on Build GRCh38
rs184024090 Chr.1: 200832568-200832568 on Build GRCh38
rs1026960748 Chr.1: 200832777-200832777 on Build GRCh38
rs769060648 Chr.1: 200832388-200832388 on Build GRCh38
rs774873897 Chr.1: 200832433-200832433 on Build GRCh38
rs563269466 Chr.1: 200832448-200832449 on Build GRCh38
rs530150498 Chr.1: 200832440-200832440 on Build GRCh38
rs761432779 Chr.1: 200832810-200832810 on Build GRCh38
rs971175459 Chr.1: 200832372-200832372 on Build GRCh38
rs772532652 Chr.1: 200832553-200832553 on Build GRCh38
rs534493672 Chr.1: 200832631-200832631 on Build GRCh38
rs763801333 Chr.1: 200832790-200832790 on Build GRCh38
rs759422790 Chr.1: 200832712-200832712 on Build GRCh38
rs768726638 Chr.1: 200832663-200832667 on Build GRCh38
rs777027508 Chr.1: 200832683-200832683 on Build GRCh38
rs889934106 Chr.1: 200832626-200832626 on Build GRCh38
rs1009735328 Chr.1: 200832597-200832597 on Build GRCh38

View Full Product Details