Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 201496090-201496604 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSRP1
Entrez Gene ID
1465
Gene Name
cysteine and glycine rich protein 1
Gene Aliases
CRP, CRP1, CSRP, CYRP, D1S181E, HEL-141, HEL-S-286
UniGene
Hs.108080
Gene Chromosome Location
Chr.1: 201483530-201507259 on Build GRCh38
Transcripts
NM_001193570.1 NM_001193571.1 NM_001193572.1 NM_004078.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5490809 c.75C>T p.C25C Chr.1: 201496229-201496229 on Build GRCh38
5780898 c.76G>A p.E26K Chr.1: 201496228-201496228 on Build GRCh38
4995352 c.76G>T p.E26* Chr.1: 201496228-201496228 on Build GRCh38
3481569 c.29G>C p.C10S Chr.1: 201496275-201496275 on Build GRCh38
2212465 c.5C>G p.P2R Chr.1: 201496299-201496299 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs370889377 Chr.1: 201496248-201496248 on Build GRCh38
rs936876338 Chr.1: 201496123-201496123 on Build GRCh38
rs781159226 Chr.1: 201496276-201496276 on Build GRCh38
rs767061946 Chr.1: 201496272-201496272 on Build GRCh38
rs771012861 Chr.1: 201496322-201496322 on Build GRCh38
rs943343272 Chr.1: 201496119-201496137 on Build GRCh38
rs906085904 Chr.1: 201496466-201496466 on Build GRCh38
rs756878007 Chr.1: 201496156-201496156 on Build GRCh38
rs181560669 Chr.1: 201496318-201496318 on Build GRCh38
rs868040607 Chr.1: 201496484-201496484 on Build GRCh38
rs747010171 Chr.1: 201496321-201496321 on Build GRCh38
rs374623326 Chr.1: 201496208-201496208 on Build GRCh38
rs964597848 Chr.1: 201496557-201496557 on Build GRCh38
rs975925342 Chr.1: 201496559-201496559 on Build GRCh38
rs118037576 Chr.1: 201496487-201496487 on Build GRCh38
rs779141829 Chr.1: 201496194-201496194 on Build GRCh38
rs558086595 Chr.1: 201496192-201496192 on Build GRCh38
rs1010304090 Chr.1: 201496488-201496488 on Build GRCh38
rs542736703 Chr.1: 201496465-201496465 on Build GRCh38
rs34504522 Chr.1: 201496222-201496222 on Build GRCh38
rs188529073 Chr.1: 201496256-201496256 on Build GRCh38
rs137955502 Chr.1: 201496480-201496480 on Build GRCh38
rs867766397 Chr.1: 201496300-201496300 on Build GRCh38
rs766752796 Chr.1: 201496295-201496295 on Build GRCh38
rs1002963218 Chr.1: 201496209-201496209 on Build GRCh38
rs112968875 Chr.1: 201496190-201496190 on Build GRCh38
rs752982852 Chr.1: 201496317-201496317 on Build GRCh38
rs754308439 Chr.1: 201496298-201496298 on Build GRCh38
rs113037411 Chr.1: 201496191-201496191 on Build GRCh38
rs189276150 Chr.1: 201496145-201496145 on Build GRCh38
rs149370653 Chr.1: 201496229-201496229 on Build GRCh38
rs943137983 Chr.1: 201496133-201496133 on Build GRCh38
rs202032834 Chr.1: 201496184-201496184 on Build GRCh38
rs777921929 Chr.1: 201496319-201496319 on Build GRCh38
rs929041989 Chr.1: 201496370-201496370 on Build GRCh38
rs768417898 Chr.1: 201496478-201496478 on Build GRCh38
rs746890457 Chr.1: 201496228-201496228 on Build GRCh38
rs755381430 Chr.1: 201496299-201496299 on Build GRCh38
rs531966865 Chr.1: 201496423-201496424 on Build GRCh38
rs908858960 Chr.1: 201496400-201496400 on Build GRCh38
rs779233422 Chr.1: 201496308-201496308 on Build GRCh38
rs773159072 Chr.1: 201496217-201496217 on Build GRCh38
rs150789671 Chr.1: 201496212-201496212 on Build GRCh38
rs761543312 Chr.1: 201496354-201496354 on Build GRCh38
rs755219938 Chr.1: 201496193-201496193 on Build GRCh38
rs1047368680 Chr.1: 201496176-201496176 on Build GRCh38
rs768399441 Chr.1: 201496257-201496257 on Build GRCh38
rs200427961 Chr.1: 201496144-201496144 on Build GRCh38
rs145929705 Chr.1: 201496518-201496518 on Build GRCh38
rs749226715 Chr.1: 201496337-201496337 on Build GRCh38
rs565249527 Chr.1: 201496482-201496482 on Build GRCh38
rs185705352 Chr.1: 201496379-201496379 on Build GRCh38
rs192359347 Chr.1: 201496574-201496574 on Build GRCh38
rs180825588 Chr.1: 201496183-201496183 on Build GRCh38
rs527970901 Chr.1: 201496506-201496506 on Build GRCh38
rs928965039 Chr.1: 201496173-201496173 on Build GRCh38
rs868328784 Chr.1: 201496485-201496485 on Build GRCh38
rs375945393 Chr.1: 201496369-201496369 on Build GRCh38
rs774225612 Chr.1: 201496352-201496352 on Build GRCh38
rs914359331 Chr.1: 201496129-201496129 on Build GRCh38
rs1006948788 Chr.1: 201496474-201496474 on Build GRCh38
rs910370652 Chr.1: 201496128-201496128 on Build GRCh38
rs780191040 Chr.1: 201496330-201496330 on Build GRCh38
rs578070214 Chr.1: 201496460-201496460 on Build GRCh38
rs15769 Chr.1: 201496266-201496266 on Build GRCh38
rs149458536 Chr.1: 201496481-201496481 on Build GRCh38
rs931232960 Chr.1: 201496410-201496410 on Build GRCh38
rs768088030 Chr.1: 201496573-201496573 on Build GRCh38
rs942311441 Chr.1: 201496427-201496427 on Build GRCh38
rs979339457 Chr.1: 201496316-201496316 on Build GRCh38
rs142986141 Chr.1: 201496233-201496233 on Build GRCh38
rs995390904 Chr.1: 201496196-201496196 on Build GRCh38
rs368827071 Chr.1: 201496503-201496503 on Build GRCh38
rs751225832 Chr.1: 201496148-201496148 on Build GRCh38
rs545338600 Chr.1: 201496477-201496477 on Build GRCh38
rs766923917 Chr.1: 201496170-201496170 on Build GRCh38
rs745821216 Chr.1: 201496221-201496221 on Build GRCh38
rs904512691 Chr.1: 201496504-201496504 on Build GRCh38
rs768641420 Chr.1: 201496341-201496341 on Build GRCh38
rs761271266 Chr.1: 201496291-201496291 on Build GRCh38
rs566603064 Chr.1: 201496131-201496131 on Build GRCh38
rs979830401 Chr.1: 201496551-201496551 on Build GRCh38
rs763961780 Chr.1: 201496244-201496244 on Build GRCh38

View Full Product Details