Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 202134411-202134856 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL8A
Entrez Gene ID
127829
Gene Name
ADP ribosylation factor like GTPase 8A
Gene Aliases
ARL10B, GIE2
UniGene
Hs.497399
Gene Chromosome Location
Chr.1: 202133404-202144743 on Build GRCh38
Transcripts
NM_001256129.1 NM_138795.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
902292 c.543G>A p.S181S Chr.1: 202134485-202134485 on Build GRCh38
6604849 c.551G>A p.R184Q Chr.1: 202134477-202134477 on Build GRCh38
902293 c.540C>A p.H180Q Chr.1: 202134488-202134488 on Build GRCh38
1245480 c.561A>G p.*187W Chr.1: 202134467-202134467 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs750746128 Chr.1: 202134546-202134546 on Build GRCh38
rs115633954 Chr.1: 202134731-202134731 on Build GRCh38
rs967180720 Chr.1: 202134523-202134523 on Build GRCh38
rs992992046 Chr.1: 202134763-202134763 on Build GRCh38
rs187554537 Chr.1: 202134732-202134732 on Build GRCh38
rs997353413 Chr.1: 202134678-202134678 on Build GRCh38
rs908828138 Chr.1: 202134645-202134645 on Build GRCh38
rs770857905 Chr.1: 202134478-202134478 on Build GRCh38
rs372869400 Chr.1: 202134434-202134434 on Build GRCh38
rs998622672 Chr.1: 202134768-202134768 on Build GRCh38
rs900988137 Chr.1: 202134594-202134594 on Build GRCh38
rs377183598 Chr.1: 202134697-202134697 on Build GRCh38
rs563337857 Chr.1: 202134806-202134806 on Build GRCh38
rs116358781 Chr.1: 202134833-202134833 on Build GRCh38
rs1001718058 Chr.1: 202134700-202134700 on Build GRCh38
rs750091696 Chr.1: 202134668-202134668 on Build GRCh38
rs374485104 Chr.1: 202134450-202134450 on Build GRCh38
rs780423710 Chr.1: 202134685-202134685 on Build GRCh38
rs992521195 Chr.1: 202134514-202134514 on Build GRCh38
rs749361259 Chr.1: 202134456-202134456 on Build GRCh38
rs978929269 Chr.1: 202134812-202134812 on Build GRCh38
rs183146460 Chr.1: 202134570-202134570 on Build GRCh38
rs774904176 Chr.1: 202134493-202134493 on Build GRCh38
rs553549595 Chr.1: 202134540-202134540 on Build GRCh38
rs1008266477 Chr.1: 202134730-202134731 on Build GRCh38
rs1018693351 Chr.1: 202134490-202134490 on Build GRCh38
rs558816143 Chr.1: 202134662-202134662 on Build GRCh38
rs967118932 Chr.1: 202134794-202134794 on Build GRCh38
rs977255644 Chr.1: 202134557-202134557 on Build GRCh38
rs915572957 Chr.1: 202134616-202134616 on Build GRCh38
rs927438452 Chr.1: 202134453-202134453 on Build GRCh38
rs746888089 Chr.1: 202134477-202134477 on Build GRCh38
rs60688550 Chr.1: 202134776-202134776 on Build GRCh38
rs886563677 Chr.1: 202134638-202134638 on Build GRCh38
rs200817993 Chr.1: 202134563-202134563 on Build GRCh38
rs368519424 Chr.1: 202134564-202134564 on Build GRCh38
rs1052272777 Chr.1: 202134611-202134611 on Build GRCh38
rs756337108 Chr.1: 202134552-202134552 on Build GRCh38
rs552398596 Chr.1: 202134466-202134466 on Build GRCh38
rs762448631 Chr.1: 202134521-202134521 on Build GRCh38
rs752073439 Chr.1: 202134528-202134528 on Build GRCh38
rs4342901 Chr.1: 202134674-202134674 on Build GRCh38
rs141580472 Chr.1: 202134485-202134485 on Build GRCh38
rs200958333 Chr.1: 202134550-202134550 on Build GRCh38
rs202209471 Chr.1: 202134537-202134537 on Build GRCh38
rs764641838 Chr.1: 202134526-202134526 on Build GRCh38
rs190151498 Chr.1: 202134556-202134556 on Build GRCh38
rs780288524 Chr.1: 202134561-202134561 on Build GRCh38
rs545321027 Chr.1: 202134599-202134599 on Build GRCh38
rs201262919 Chr.1: 202134543-202134543 on Build GRCh38
rs769466486 Chr.1: 202134486-202134486 on Build GRCh38
rs190515663 Chr.1: 202134761-202134761 on Build GRCh38
rs950293480 Chr.1: 202134631-202134631 on Build GRCh38
rs913011828 Chr.1: 202134533-202134533 on Build GRCh38
rs776450461 Chr.1: 202134479-202134479 on Build GRCh38
rs552435223 Chr.1: 202134834-202134834 on Build GRCh38
rs772172377 Chr.1: 202134462-202134462 on Build GRCh38
rs1005380944 Chr.1: 202134639-202134639 on Build GRCh38

View Full Product Details