Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 202135064-202135564 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL8A
Entrez Gene ID
127829
Gene Name
ADP ribosylation factor like GTPase 8A
Gene Aliases
ARL10B, GIE2
UniGene
Hs.497399
Gene Chromosome Location
Chr.1: 202133404-202144743 on Build GRCh38
Transcripts
NM_001256129.1 NM_138795.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5462116 c.425A>G p.E142G Chr.1: 202135474-202135474 on Build GRCh38
4925236 c.385G>C p.G129R Chr.1: 202135514-202135514 on Build GRCh38
6604848 c.480C>T p.Y160Y Chr.1: 202135181-202135181 on Build GRCh38
6035417 c.511+5G>A p.? Chr.1: 202135145-202135145 on Build GRCh38
7123647 c.496G>C p.E166Q Chr.1: 202135165-202135165 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs903337504 Chr.1: 202135407-202135407 on Build GRCh38
rs750789458 Chr.1: 202135214-202135214 on Build GRCh38
rs745710382 Chr.1: 202135415-202135415 on Build GRCh38
rs982702915 Chr.1: 202135526-202135526 on Build GRCh38
rs978514225 Chr.1: 202135098-202135098 on Build GRCh38
rs202188926 Chr.1: 202135139-202135139 on Build GRCh38
rs769401334 Chr.1: 202135127-202135127 on Build GRCh38
rs780898874 Chr.1: 202135105-202135105 on Build GRCh38
rs549508956 Chr.1: 202135331-202135331 on Build GRCh38
rs764023947 Chr.1: 202135530-202135530 on Build GRCh38
rs779645934 Chr.1: 202135431-202135431 on Build GRCh38
rs1042335412 Chr.1: 202135486-202135486 on Build GRCh38
rs1032758499 Chr.1: 202135413-202135413 on Build GRCh38
rs925722403 Chr.1: 202135116-202135116 on Build GRCh38
rs752803471 Chr.1: 202135409-202135409 on Build GRCh38
rs770947629 Chr.1: 202135102-202135102 on Build GRCh38
rs781186281 Chr.1: 202135412-202135412 on Build GRCh38
rs752893445 Chr.1: 202135497-202135497 on Build GRCh38
rs143346308 Chr.1: 202135195-202135195 on Build GRCh38
rs374952394 Chr.1: 202135437-202135437 on Build GRCh38
rs74745206 Chr.1: 202135202-202135202 on Build GRCh38
rs750377376 Chr.1: 202135531-202135531 on Build GRCh38
rs1055685053 Chr.1: 202135310-202135310 on Build GRCh38
rs768310748 Chr.1: 202135112-202135112 on Build GRCh38
rs755214681 Chr.1: 202135266-202135266 on Build GRCh38
rs758493587 Chr.1: 202135524-202135524 on Build GRCh38
rs757182953 Chr.1: 202135411-202135411 on Build GRCh38
rs188612416 Chr.1: 202135285-202135285 on Build GRCh38
rs776216851 Chr.1: 202135264-202135265 on Build GRCh38
rs932803525 Chr.1: 202135414-202135414 on Build GRCh38
rs184331864 Chr.1: 202135371-202135371 on Build GRCh38
rs371637265 Chr.1: 202135398-202135398 on Build GRCh38
rs559749190 Chr.1: 202135494-202135494 on Build GRCh38
rs745645592 Chr.1: 202135121-202135121 on Build GRCh38
rs1023122402 Chr.1: 202135267-202135267 on Build GRCh38
rs147134888 Chr.1: 202135315-202135315 on Build GRCh38
rs761418805 Chr.1: 202135442-202135443 on Build GRCh38
rs762532680 Chr.1: 202135141-202135141 on Build GRCh38
rs150397242 Chr.1: 202135154-202135154 on Build GRCh38
rs922264836 Chr.1: 202135181-202135181 on Build GRCh38
rs761274233 Chr.1: 202135305-202135305 on Build GRCh38
rs747622594 Chr.1: 202135456-202135456 on Build GRCh38
rs116468762 Chr.1: 202135280-202135280 on Build GRCh38
rs755964614 Chr.1: 202135539-202135539 on Build GRCh38
rs41310917 Chr.1: 202135224-202135224 on Build GRCh38
rs192794747 Chr.1: 202135368-202135368 on Build GRCh38
rs1037780198 Chr.1: 202135283-202135283 on Build GRCh38
rs775197534 Chr.1: 202135140-202135140 on Build GRCh38
rs369366896 Chr.1: 202135447-202135447 on Build GRCh38
rs61758714 Chr.1: 202135488-202135488 on Build GRCh38
rs763974345 Chr.1: 202135441-202135441 on Build GRCh38
rs766800611 Chr.1: 202135490-202135490 on Build GRCh38
rs773896145 Chr.1: 202135444-202135444 on Build GRCh38
rs912614547 Chr.1: 202135440-202135440 on Build GRCh38
rs144483766 Chr.1: 202135487-202135487 on Build GRCh38
rs754120159 Chr.1: 202135261-202135261 on Build GRCh38
rs567207516 Chr.1: 202135095-202135095 on Build GRCh38
rs761093633 Chr.1: 202135223-202135223 on Build GRCh38
rs199641249 Chr.1: 202135495-202135495 on Build GRCh38
rs941215429 Chr.1: 202135282-202135282 on Build GRCh38
rs773323427 Chr.1: 202135143-202135143 on Build GRCh38
rs1014482836 Chr.1: 202135343-202135343 on Build GRCh38
rs140334774 Chr.1: 202135395-202135395 on Build GRCh38
rs558539159 Chr.1: 202135284-202135284 on Build GRCh38
rs920229013 Chr.1: 202135383-202135383 on Build GRCh38
rs974324632 Chr.1: 202135357-202135357 on Build GRCh38

View Full Product Details