Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 203165544-203166052 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADORA1
Entrez Gene ID
134
Gene Name
adenosine A1 receptor
Gene Aliases
RDC7
UniGene
Hs.77867
Gene Chromosome Location
Chr.1: 203127705-203167405 on Build GRCh38
Transcripts
NM_000674.2 NM_001048230.1 XM_005244901.1 XM_005244902.4 XM_017000342.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5353445 c.934G>A p.A312T Chr.1: 203165853-203165853 on Build GRCh38
6595313 c.744G>C p.L248L Chr.1: 203165663-203165663 on Build GRCh38
5451203 c.648G>A p.S216S Chr.1: 203165567-203165567 on Build GRCh38
678082 c.943A>G p.I315V Chr.1: 203165862-203165862 on Build GRCh38
3789350 c.913G>A p.D305N Chr.1: 203165832-203165832 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1003913268 Chr.1: 203165971-203165971 on Build GRCh38
rs200808876 Chr.1: 203165831-203165831 on Build GRCh38
rs141314109 Chr.1: 203165912-203165912 on Build GRCh38
rs756052593 Chr.1: 203165863-203165863 on Build GRCh38
rs766823930 Chr.1: 203165791-203165791 on Build GRCh38
rs537966964 Chr.1: 203165624-203165624 on Build GRCh38
rs201811129 Chr.1: 203165807-203165807 on Build GRCh38
rs780947937 Chr.1: 203165851-203165851 on Build GRCh38
rs200908360 Chr.1: 203165853-203165853 on Build GRCh38
rs201762463 Chr.1: 203165865-203165865 on Build GRCh38
rs748346254 Chr.1: 203165754-203165754 on Build GRCh38
rs750835106 Chr.1: 203165666-203165666 on Build GRCh38
rs201558854 Chr.1: 203165710-203165710 on Build GRCh38
rs769134627 Chr.1: 203165933-203165933 on Build GRCh38
rs762570469 Chr.1: 203165919-203165919 on Build GRCh38
rs746924602 Chr.1: 203165600-203165600 on Build GRCh38
rs201486417 Chr.1: 203165906-203165906 on Build GRCh38
rs146573702 Chr.1: 203165633-203165633 on Build GRCh38
rs141819948 Chr.1: 203165842-203165842 on Build GRCh38
rs199760746 Chr.1: 203165714-203165714 on Build GRCh38
rs543118926 Chr.1: 203165927-203165927 on Build GRCh38
rs796395239 Chr.1: 203165907-203165907 on Build GRCh38
rs748812754 Chr.1: 203165753-203165753 on Build GRCh38
rs761031787 Chr.1: 203165776-203165776 on Build GRCh38
rs201646414 Chr.1: 203165841-203165841 on Build GRCh38
rs200246307 Chr.1: 203165858-203165858 on Build GRCh38
rs777752008 Chr.1: 203165597-203165597 on Build GRCh38
rs775001661 Chr.1: 203165914-203165914 on Build GRCh38
rs202089967 Chr.1: 203165949-203165949 on Build GRCh38
rs202084025 Chr.1: 203165713-203165713 on Build GRCh38
rs200606043 Chr.1: 203165973-203165973 on Build GRCh38
rs200543088 Chr.1: 203165704-203165704 on Build GRCh38
rs920762648 Chr.1: 203165761-203165761 on Build GRCh38
rs575455405 Chr.1: 203165767-203165767 on Build GRCh38
rs199829860 Chr.1: 203165805-203165805 on Build GRCh38
rs201449710 Chr.1: 203165993-203165993 on Build GRCh38
rs763787447 Chr.1: 203165928-203165928 on Build GRCh38
rs775034018 Chr.1: 203165671-203165671 on Build GRCh38
rs750241098 Chr.1: 203165950-203165950 on Build GRCh38
rs376563906 Chr.1: 203165702-203165702 on Build GRCh38
rs202192831 Chr.1: 203165854-203165854 on Build GRCh38
rs749274582 Chr.1: 203165728-203165728 on Build GRCh38
rs200032682 Chr.1: 203165808-203165808 on Build GRCh38
rs200795962 Chr.1: 203165905-203165905 on Build GRCh38
rs778641521 Chr.1: 203165580-203165580 on Build GRCh38
rs539069853 Chr.1: 203165566-203165566 on Build GRCh38
rs772336239 Chr.1: 203165763-203165763 on Build GRCh38
rs199913778 Chr.1: 203165994-203165994 on Build GRCh38
rs752365197 Chr.1: 203165696-203165696 on Build GRCh38
rs1012603464 Chr.1: 203165897-203165897 on Build GRCh38
rs781198573 Chr.1: 203165726-203165726 on Build GRCh38
rs200703683 Chr.1: 203165750-203165750 on Build GRCh38
rs200325525 Chr.1: 203165873-203165873 on Build GRCh38
rs375908464 Chr.1: 203165902-203165902 on Build GRCh38
rs1040627926 Chr.1: 203165796-203165796 on Build GRCh38
rs779515290 Chr.1: 203165570-203165570 on Build GRCh38
rs760187303 Chr.1: 203165712-203165712 on Build GRCh38
rs757137013 Chr.1: 203165845-203165845 on Build GRCh38
rs939953705 Chr.1: 203165673-203165673 on Build GRCh38
rs778391614 Chr.1: 203165688-203165688 on Build GRCh38
rs763843184 Chr.1: 203165882-203165885 on Build GRCh38
rs754539453 Chr.1: 203165685-203165685 on Build GRCh38
rs201016113 Chr.1: 203165576-203165576 on Build GRCh38
rs199516537 Chr.1: 203165661-203165661 on Build GRCh38
rs201482062 Chr.1: 203165681-203165681 on Build GRCh38
rs201703869 Chr.1: 203165790-203165790 on Build GRCh38
rs962249377 Chr.1: 203166030-203166030 on Build GRCh38
rs200239809 Chr.1: 203165567-203165567 on Build GRCh38
rs138495914 Chr.1: 203165885-203165885 on Build GRCh38
rs142252167 Chr.1: 203165806-203165806 on Build GRCh38
rs774651930 Chr.1: 203165628-203165628 on Build GRCh38
rs202104893 Chr.1: 203165867-203165867 on Build GRCh38
rs754581480 Chr.1: 203165577-203165577 on Build GRCh38
rs775712225 Chr.1: 203165619-203165619 on Build GRCh38
rs747849534 Chr.1: 203165584-203165584 on Build GRCh38
rs145245940 Chr.1: 203165674-203165674 on Build GRCh38
rs17852405 Chr.1: 203165701-203165701 on Build GRCh38
rs746871603 Chr.1: 203165703-203165703 on Build GRCh38
rs200117311 Chr.1: 203165964-203165964 on Build GRCh38
rs753610663 Chr.1: 203165885-203165886 on Build GRCh38
rs201849915 Chr.1: 203165585-203165585 on Build GRCh38
rs201317822 Chr.1: 203165874-203165874 on Build GRCh38
rs200564867 Chr.1: 203165675-203165675 on Build GRCh38
rs530740427 Chr.1: 203165952-203165952 on Build GRCh38
rs974004725 Chr.1: 203165963-203165963 on Build GRCh38
rs773446422 Chr.1: 203165888-203165888 on Build GRCh38
rs200653241 Chr.1: 203165913-203165913 on Build GRCh38
rs751284960 Chr.1: 203165931-203165931 on Build GRCh38
rs200281933 Chr.1: 203165615-203165615 on Build GRCh38
rs201083386 Chr.1: 203165623-203165623 on Build GRCh38

View Full Product Details