Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 203178841-203179344 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHI3L1
Entrez Gene ID
1116
Gene Name
chitinase 3 like 1
Gene Aliases
ASRT7, CGP-39, GP-39, GP39, HC-gp39, HCGP-3P, YKL-40, YKL40, YYL-40, hCGP-39
UniGene
Hs.382202
Gene Chromosome Location
Chr.1: 203178931-203186794 on Build GRCh38
Transcripts
NM_001276.2

SNP Detail

SNP ID Chromosome Location
rs571121302 Chr.1: 203179143-203179143 on Build GRCh38
rs895539958 Chr.1: 203179181-203179181 on Build GRCh38
rs192635263 Chr.1: 203179212-203179212 on Build GRCh38
rs933570221 Chr.1: 203178870-203178870 on Build GRCh38
rs892072752 Chr.1: 203179156-203179156 on Build GRCh38
rs796227676 Chr.1: 203178949-203178949 on Build GRCh38
rs1057478544 Chr.1: 203179323-203179323 on Build GRCh38
rs1049452 Chr.1: 203179133-203179133 on Build GRCh38
rs188823413 Chr.1: 203178899-203178899 on Build GRCh38
rs1019281955 Chr.1: 203178942-203178942 on Build GRCh38
rs114943381 Chr.1: 203179040-203179040 on Build GRCh38
rs993815624 Chr.1: 203179185-203179185 on Build GRCh38
rs989201520 Chr.1: 203179165-203179165 on Build GRCh38
rs77111685 Chr.1: 203179231-203179231 on Build GRCh38
rs376137032 Chr.1: 203178984-203178984 on Build GRCh38
rs769420439 Chr.1: 203179038-203179039 on Build GRCh38
rs567550831 Chr.1: 203179169-203179169 on Build GRCh38
rs550138054 Chr.1: 203179005-203179005 on Build GRCh38
rs141536984 Chr.1: 203179166-203179166 on Build GRCh38
rs530085748 Chr.1: 203178992-203178992 on Build GRCh38
rs1049707590 Chr.1: 203179207-203179207 on Build GRCh38
rs184965874 Chr.1: 203179309-203179309 on Build GRCh38
rs888152437 Chr.1: 203179290-203179290 on Build GRCh38
rs1025543021 Chr.1: 203179026-203179026 on Build GRCh38
rs970252131 Chr.1: 203179134-203179134 on Build GRCh38
rs866568105 Chr.1: 203179013-203179013 on Build GRCh38
rs892039816 Chr.1: 203178898-203178898 on Build GRCh38
rs952376641 Chr.1: 203179042-203179042 on Build GRCh38
rs12408583 Chr.1: 203179232-203179232 on Build GRCh38
rs79819896 Chr.1: 203179074-203179074 on Build GRCh38
rs533698461 Chr.1: 203179146-203179146 on Build GRCh38
rs540660421 Chr.1: 203179133-203179133 on Build GRCh38
rs78428658 Chr.1: 203179244-203179244 on Build GRCh38
rs1006500880 Chr.1: 203179058-203179058 on Build GRCh38
rs1053404392 Chr.1: 203178885-203178885 on Build GRCh38
rs112644498 Chr.1: 203178930-203178930 on Build GRCh38
rs905993607 Chr.1: 203179167-203179167 on Build GRCh38
rs557159785 Chr.1: 203178923-203178923 on Build GRCh38
rs1049451 Chr.1: 203179151-203179151 on Build GRCh38
rs868403592 Chr.1: 203179235-203179235 on Build GRCh38
rs55768323 Chr.1: 203179318-203179318 on Build GRCh38
rs923348112 Chr.1: 203179139-203179139 on Build GRCh38
rs943837653 Chr.1: 203179217-203179217 on Build GRCh38
rs913202897 Chr.1: 203179254-203179254 on Build GRCh38
rs2297946 Chr.1: 203178924-203178924 on Build GRCh38
rs1043368621 Chr.1: 203179289-203179289 on Build GRCh38
rs959775713 Chr.1: 203178979-203178979 on Build GRCh38
rs372580019 Chr.1: 203178999-203178999 on Build GRCh38
rs988824824 Chr.1: 203179245-203179245 on Build GRCh38
rs1048521804 Chr.1: 203179264-203179264 on Build GRCh38
rs1025238486 Chr.1: 203179190-203179190 on Build GRCh38
rs1006881968 Chr.1: 203179321-203179321 on Build GRCh38
rs947622493 Chr.1: 203179284-203179284 on Build GRCh38
rs552222824 Chr.1: 203179138-203179138 on Build GRCh38

View Full Product Details