Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 203627100-203627606 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP2B4
Entrez Gene ID
493
Gene Name
ATPase plasma membrane Ca2+ transporting 4
Gene Aliases
ATP2B2, MXRA1, PMCA4, PMCA4b, PMCA4x
UniGene
Hs.343522
Gene Chromosome Location
Chr.1: 203626787-203744081 on Build GRCh38
Transcripts
NM_001001396.2 NM_001684.4

SNP Detail

SNP ID Chromosome Location
rs905026947 Chr.1: 203627509-203627509 on Build GRCh38
rs986227317 Chr.1: 203627292-203627292 on Build GRCh38
rs113901324 Chr.1: 203627318-203627318 on Build GRCh38
rs949191722 Chr.1: 203627468-203627468 on Build GRCh38
rs982001666 Chr.1: 203627540-203627540 on Build GRCh38
rs897770280 Chr.1: 203627566-203627566 on Build GRCh38
rs905017585 Chr.1: 203627184-203627184 on Build GRCh38
rs1046639319 Chr.1: 203627172-203627172 on Build GRCh38
rs570013180 Chr.1: 203627360-203627360 on Build GRCh38
rs1037485373 Chr.1: 203627556-203627556 on Build GRCh38
rs1006025233 Chr.1: 203627532-203627532 on Build GRCh38
rs1036209352 Chr.1: 203627575-203627575 on Build GRCh38
rs371616536 Chr.1: 203627369-203627369 on Build GRCh38
rs910720806 Chr.1: 203627295-203627295 on Build GRCh38
rs954831829 Chr.1: 203627263-203627263 on Build GRCh38
rs17537593 Chr.1: 203627354-203627354 on Build GRCh38
rs995009244 Chr.1: 203627264-203627267 on Build GRCh38
rs1030271788 Chr.1: 203627245-203627245 on Build GRCh38
rs527999930 Chr.1: 203627309-203627309 on Build GRCh38
rs898855511 Chr.1: 203627578-203627578 on Build GRCh38
rs57046878 Chr.1: 203627415-203627415 on Build GRCh38
rs752584688 Chr.1: 203627583-203627583 on Build GRCh38
rs58171074 Chr.1: 203627408-203627408 on Build GRCh38
rs192467816 Chr.1: 203627353-203627353 on Build GRCh38
rs561155740 Chr.1: 203627340-203627340 on Build GRCh38
rs552559558 Chr.1: 203627577-203627577 on Build GRCh38
rs989544569 Chr.1: 203627132-203627132 on Build GRCh38
rs1016142143 Chr.1: 203627191-203627191 on Build GRCh38
rs766495892 Chr.1: 203627474-203627474 on Build GRCh38
rs1023928995 Chr.1: 203627159-203627159 on Build GRCh38
rs536388142 Chr.1: 203627366-203627366 on Build GRCh38
rs1019789002 Chr.1: 203627331-203627331 on Build GRCh38
rs575974752 Chr.1: 203627181-203627181 on Build GRCh38
rs948121518 Chr.1: 203627477-203627477 on Build GRCh38
rs116586407 Chr.1: 203627200-203627200 on Build GRCh38
rs947887428 Chr.1: 203627164-203627164 on Build GRCh38
rs77719238 Chr.1: 203627139-203627139 on Build GRCh38
rs4600103 Chr.1: 203627190-203627190 on Build GRCh38
rs200998597 Chr.1: 203627345-203627346 on Build GRCh38
rs896263140 Chr.1: 203627140-203627140 on Build GRCh38
rs562970343 Chr.1: 203627300-203627300 on Build GRCh38
rs76857273 Chr.1: 203627152-203627152 on Build GRCh38
rs1007089524 Chr.1: 203627324-203627324 on Build GRCh38
rs934883859 Chr.1: 203627355-203627355 on Build GRCh38
rs1030325805 Chr.1: 203627580-203627580 on Build GRCh38
rs927798001 Chr.1: 203627547-203627547 on Build GRCh38
rs767690388 Chr.1: 203627225-203627225 on Build GRCh38
rs917586615 Chr.1: 203627439-203627439 on Build GRCh38
rs76891602 Chr.1: 203627138-203627138 on Build GRCh38

View Full Product Details