Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 204131334-204131844 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ETNK2
Entrez Gene ID
55224
Gene Name
ethanolamine kinase 2
Gene Aliases
EKI2, HMFT1716
UniGene
Hs.497469
Gene Chromosome Location
Chr.1: 204131061-204152182 on Build GRCh38
Transcripts
NM_001297760.1 NM_001297761.1 NM_001297762.1 NM_018208.3 XM_011509714.2 XM_011509715.1 XM_017001632.1

SNP Detail

SNP ID Chromosome Location
rs1036816829 Chr.1: 204131686-204131686 on Build GRCh38
rs967106250 Chr.1: 204131567-204131567 on Build GRCh38
rs529432849 Chr.1: 204131676-204131676 on Build GRCh38
rs76718240 Chr.1: 204131580-204131580 on Build GRCh38
rs79403699 Chr.1: 204131818-204131818 on Build GRCh38
rs777135117 Chr.1: 204131707-204131707 on Build GRCh38
rs564460294 Chr.1: 204131560-204131560 on Build GRCh38
rs893274509 Chr.1: 204131602-204131602 on Build GRCh38
rs967158519 Chr.1: 204131777-204131777 on Build GRCh38
rs1048259842 Chr.1: 204131500-204131500 on Build GRCh38
rs900768992 Chr.1: 204131559-204131559 on Build GRCh38
rs1008763521 Chr.1: 204131759-204131759 on Build GRCh38
rs1004870586 Chr.1: 204131379-204131379 on Build GRCh38
rs916808701 Chr.1: 204131636-204131636 on Build GRCh38
rs181336042 Chr.1: 204131664-204131664 on Build GRCh38
rs983278247 Chr.1: 204131811-204131811 on Build GRCh38
rs920419792 Chr.1: 204131453-204131453 on Build GRCh38
rs746575421 Chr.1: 204131768-204131768 on Build GRCh38
rs948247678 Chr.1: 204131648-204131648 on Build GRCh38
rs778495752 Chr.1: 204131438-204131438 on Build GRCh38
rs998617974 Chr.1: 204131812-204131812 on Build GRCh38
rs995216317 Chr.1: 204131720-204131720 on Build GRCh38
rs959310721 Chr.1: 204131762-204131762 on Build GRCh38
rs746474734 Chr.1: 204131645-204131645 on Build GRCh38
rs1034651757 Chr.1: 204131731-204131731 on Build GRCh38
rs1050349445 Chr.1: 204131740-204131740 on Build GRCh38
rs549500658 Chr.1: 204131788-204131788 on Build GRCh38
rs777254623 Chr.1: 204131671-204131671 on Build GRCh38
rs541550800 Chr.1: 204131455-204131455 on Build GRCh38
rs889016564 Chr.1: 204131750-204131750 on Build GRCh38
rs77936713 Chr.1: 204131822-204131822 on Build GRCh38
rs1046536975 Chr.1: 204131650-204131650 on Build GRCh38
rs989762073 Chr.1: 204131622-204131622 on Build GRCh38
rs935625904 Chr.1: 204131618-204131618 on Build GRCh38
rs574897142 Chr.1: 204131525-204131525 on Build GRCh38
rs75876713 Chr.1: 204131819-204131819 on Build GRCh38
rs966462263 Chr.1: 204131703-204131703 on Build GRCh38
rs772816567 Chr.1: 204131621-204131621 on Build GRCh38
rs990734149 Chr.1: 204131767-204131767 on Build GRCh38
rs533436486 Chr.1: 204131579-204131579 on Build GRCh38
rs944720777 Chr.1: 204131528-204131528 on Build GRCh38
rs543769555 Chr.1: 204131534-204131534 on Build GRCh38
rs1029289603 Chr.1: 204131459-204131459 on Build GRCh38
rs554992422 Chr.1: 204131473-204131473 on Build GRCh38
rs771583361 Chr.1: 204131655-204131655 on Build GRCh38
rs145968121 Chr.1: 204131452-204131452 on Build GRCh38

View Full Product Details