Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 204131721-204132215 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ETNK2
Entrez Gene ID
55224
Gene Name
ethanolamine kinase 2
Gene Aliases
EKI2, HMFT1716
UniGene
Hs.497469
Gene Chromosome Location
Chr.1: 204131061-204152182 on Build GRCh38
Transcripts
NM_001297760.1 NM_001297761.1 NM_001297762.1 NM_018208.3 XM_011509714.2 XM_011509715.1 XM_017001632.1

SNP Detail

SNP ID Chromosome Location
rs1012201125 Chr.1: 204132042-204132042 on Build GRCh38
rs759156287 Chr.1: 204132170-204132170 on Build GRCh38
rs780402256 Chr.1: 204132162-204132163 on Build GRCh38
rs759820778 Chr.1: 204131892-204131892 on Build GRCh38
rs967158519 Chr.1: 204131777-204131777 on Build GRCh38
rs983278247 Chr.1: 204131811-204131811 on Build GRCh38
rs539891992 Chr.1: 204132156-204132156 on Build GRCh38
rs763143837 Chr.1: 204131973-204131973 on Build GRCh38
rs771663723 Chr.1: 204131844-204131844 on Build GRCh38
rs139047077 Chr.1: 204132027-204132027 on Build GRCh38
rs889016564 Chr.1: 204131750-204131750 on Build GRCh38
rs574964789 Chr.1: 204132164-204132164 on Build GRCh38
rs77936713 Chr.1: 204131822-204131822 on Build GRCh38
rs368115670 Chr.1: 204132160-204132160 on Build GRCh38
rs907690738 Chr.1: 204132079-204132079 on Build GRCh38
rs41313900 Chr.1: 204131912-204131912 on Build GRCh38
rs907972385 Chr.1: 204131836-204131836 on Build GRCh38
rs983694989 Chr.1: 204132059-204132059 on Build GRCh38
rs777186551 Chr.1: 204131996-204131996 on Build GRCh38
rs1036459335 Chr.1: 204131966-204131966 on Build GRCh38
rs889070720 Chr.1: 204132002-204132004 on Build GRCh38
rs566547978 Chr.1: 204132035-204132035 on Build GRCh38
rs928140169 Chr.1: 204131914-204131914 on Build GRCh38
rs914810733 Chr.1: 204132172-204132172 on Build GRCh38
rs779185057 Chr.1: 204132142-204132142 on Build GRCh38
rs79403699 Chr.1: 204131818-204131818 on Build GRCh38
rs539339572 Chr.1: 204132010-204132010 on Build GRCh38
rs1027588062 Chr.1: 204132051-204132051 on Build GRCh38
rs1008763521 Chr.1: 204131759-204131759 on Build GRCh38
rs775177450 Chr.1: 204132041-204132041 on Build GRCh38
rs971000432 Chr.1: 204132099-204132099 on Build GRCh38
rs746575421 Chr.1: 204131768-204131768 on Build GRCh38
rs998617974 Chr.1: 204131812-204131812 on Build GRCh38
rs151131812 Chr.1: 204131986-204131986 on Build GRCh38
rs371581071 Chr.1: 204132185-204132185 on Build GRCh38
rs940823018 Chr.1: 204131948-204131948 on Build GRCh38
rs117726554 Chr.1: 204131890-204131890 on Build GRCh38
rs959310721 Chr.1: 204131762-204131762 on Build GRCh38
rs1003286423 Chr.1: 204132033-204132033 on Build GRCh38
rs139891629 Chr.1: 204131908-204131908 on Build GRCh38
rs549500658 Chr.1: 204131788-204131788 on Build GRCh38
rs772509364 Chr.1: 204132150-204132150 on Build GRCh38
rs951976383 Chr.1: 204132055-204132055 on Build GRCh38
rs535590418 Chr.1: 204132116-204132116 on Build GRCh38
rs920650217 Chr.1: 204131967-204131968 on Build GRCh38
rs1040384073 Chr.1: 204131846-204131846 on Build GRCh38
rs1012571119 Chr.1: 204132050-204132050 on Build GRCh38
rs75876713 Chr.1: 204131819-204131819 on Build GRCh38
rs1033148156 Chr.1: 204131833-204131833 on Build GRCh38
rs990734149 Chr.1: 204131767-204131767 on Build GRCh38
rs969607568 Chr.1: 204131909-204131909 on Build GRCh38
rs748662740 Chr.1: 204132147-204132147 on Build GRCh38

View Full Product Details