Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 204137025-204137520 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ETNK2
Entrez Gene ID
55224
Gene Name
ethanolamine kinase 2
Gene Aliases
EKI2, HMFT1716
UniGene
Hs.497469
Gene Chromosome Location
Chr.1: 204131061-204152182 on Build GRCh38
Transcripts
NM_001297760.1 NM_001297761.1 NM_001297762.1 NM_018208.3 XM_011509714.2 XM_011509715.1 XM_017001632.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1205737 c.871G>A p.V291M Chr.1: 204137247-204137247 on Build GRCh38
265656 c.917_918insT p.Q307fs*35 Chr.1: 204137200-204137201 on Build GRCh38
6203083 c.943G>T p.A315S Chr.1: 204137175-204137175 on Build GRCh38
5657004 c.902C>A p.A301E Chr.1: 204137216-204137216 on Build GRCh38
6246751 c.960C>A p.A320A Chr.1: 204137158-204137158 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750862892 Chr.1: 204137125-204137125 on Build GRCh38
rs1047922098 Chr.1: 204137371-204137371 on Build GRCh38
rs149960550 Chr.1: 204137137-204137137 on Build GRCh38
rs150586165 Chr.1: 204137069-204137069 on Build GRCh38
rs764235257 Chr.1: 204137086-204137086 on Build GRCh38
rs751793193 Chr.1: 204137101-204137101 on Build GRCh38
rs959275716 Chr.1: 204137454-204137455 on Build GRCh38
rs542121001 Chr.1: 204137186-204137186 on Build GRCh38
rs767749304 Chr.1: 204137198-204137198 on Build GRCh38
rs770971979 Chr.1: 204137167-204137167 on Build GRCh38
rs958236300 Chr.1: 204137463-204137463 on Build GRCh38
rs200305922 Chr.1: 204137213-204137213 on Build GRCh38
rs754406318 Chr.1: 204137207-204137207 on Build GRCh38
rs1024811494 Chr.1: 204137495-204137495 on Build GRCh38
rs757038635 Chr.1: 204137257-204137257 on Build GRCh38
rs1019324006 Chr.1: 204137406-204137406 on Build GRCh38
rs144520917 Chr.1: 204137254-204137254 on Build GRCh38
rs989390344 Chr.1: 204137468-204137468 on Build GRCh38
rs770142507 Chr.1: 204137213-204137214 on Build GRCh38
rs781006415 Chr.1: 204137258-204137258 on Build GRCh38
rs750856269 Chr.1: 204137204-204137204 on Build GRCh38
rs61825753 Chr.1: 204137188-204137188 on Build GRCh38
rs141356117 Chr.1: 204137445-204137445 on Build GRCh38
rs761926179 Chr.1: 204137219-204137219 on Build GRCh38
rs372782006 Chr.1: 204137218-204137218 on Build GRCh38
rs554916221 Chr.1: 204137327-204137327 on Build GRCh38
rs533254441 Chr.1: 204137247-204137247 on Build GRCh38
rs771127563 Chr.1: 204137097-204137097 on Build GRCh38
rs899392126 Chr.1: 204137091-204137091 on Build GRCh38
rs537270954 Chr.1: 204137387-204137387 on Build GRCh38
rs761470297 Chr.1: 204137215-204137215 on Build GRCh38
rs776082425 Chr.1: 204137396-204137396 on Build GRCh38
rs750538219 Chr.1: 204137223-204137223 on Build GRCh38
rs777234659 Chr.1: 204137239-204137239 on Build GRCh38
rs775624367 Chr.1: 204137164-204137164 on Build GRCh38
rs778646158 Chr.1: 204137149-204137149 on Build GRCh38
rs754536920 Chr.1: 204137140-204137140 on Build GRCh38
rs140658518 Chr.1: 204137209-204137209 on Build GRCh38
rs144439568 Chr.1: 204137163-204137163 on Build GRCh38
rs996928859 Chr.1: 204137444-204137444 on Build GRCh38
rs995081385 Chr.1: 204137337-204137337 on Build GRCh38
rs756691101 Chr.1: 204137127-204137127 on Build GRCh38
rs965130904 Chr.1: 204137419-204137419 on Build GRCh38
rs746909963 Chr.1: 204137166-204137166 on Build GRCh38
rs745785459 Chr.1: 204137259-204137259 on Build GRCh38
rs764023394 Chr.1: 204137189-204137189 on Build GRCh38
rs201036824 Chr.1: 204137242-204137242 on Build GRCh38
rs761038426 Chr.1: 204137225-204137225 on Build GRCh38
rs776441116 Chr.1: 204137176-204137176 on Build GRCh38
rs994241304 Chr.1: 204137281-204137281 on Build GRCh38
rs959224768 Chr.1: 204137158-204137158 on Build GRCh38
rs1034350079 Chr.1: 204137061-204137061 on Build GRCh38
rs375907212 Chr.1: 204137236-204137236 on Build GRCh38
rs202005832 Chr.1: 204137265-204137265 on Build GRCh38
rs146679279 Chr.1: 204137128-204137128 on Build GRCh38
rs753465248 Chr.1: 204137131-204137131 on Build GRCh38
rs913789717 Chr.1: 204137472-204137472 on Build GRCh38
rs372278412 Chr.1: 204137283-204137283 on Build GRCh38
rs570954657 Chr.1: 204137216-204137216 on Build GRCh38
rs566753228 Chr.1: 204137255-204137255 on Build GRCh38
rs1038656090 Chr.1: 204137228-204137228 on Build GRCh38
rs748320572 Chr.1: 204137211-204137212 on Build GRCh38
rs772058483 Chr.1: 204137288-204137288 on Build GRCh38
rs778594983 Chr.1: 204137211-204137211 on Build GRCh38
rs756100601 Chr.1: 204137276-204137276 on Build GRCh38
rs201980742 Chr.1: 204137214-204137214 on Build GRCh38
rs778959019 Chr.1: 204137280-204137280 on Build GRCh38
rs1008317727 Chr.1: 204137362-204137362 on Build GRCh38
rs1032199748 Chr.1: 204137157-204137157 on Build GRCh38
rs899441733 Chr.1: 204137330-204137330 on Build GRCh38
rs891248478 Chr.1: 204137358-204137362 on Build GRCh38
rs760801872 Chr.1: 204137392-204137392 on Build GRCh38
rs745838701 Chr.1: 204137217-204137217 on Build GRCh38
rs747823343 Chr.1: 204137161-204137161 on Build GRCh38
rs1027973860 Chr.1: 204137462-204137462 on Build GRCh38
rs762117921 Chr.1: 204137112-204137112 on Build GRCh38
rs767957800 Chr.1: 204137118-204137118 on Build GRCh38
rs761081739 Chr.1: 204137205-204137205 on Build GRCh38
rs766839183 Chr.1: 204137206-204137206 on Build GRCh38
rs886600906 Chr.1: 204137386-204137386 on Build GRCh38
rs535805165 Chr.1: 204137317-204137317 on Build GRCh38
rs1055105561 Chr.1: 204137325-204137325 on Build GRCh38
rs966646191 Chr.1: 204137076-204137076 on Build GRCh38
rs762064107 Chr.1: 204137190-204137190 on Build GRCh38
rs966538702 Chr.1: 204137483-204137483 on Build GRCh38
rs147815280 Chr.1: 204137382-204137382 on Build GRCh38
rs752325302 Chr.1: 204137248-204137248 on Build GRCh38
rs368335206 Chr.1: 204137252-204137252 on Build GRCh38

View Full Product Details