Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 204141237-204141740 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ETNK2
Entrez Gene ID
55224
Gene Name
ethanolamine kinase 2
Gene Aliases
EKI2, HMFT1716
UniGene
Hs.497469
Gene Chromosome Location
Chr.1: 204131061-204152182 on Build GRCh38
Transcripts
NM_001297760.1 NM_001297761.1 NM_001297762.1 NM_018208.3 XM_011509714.2 XM_011509715.1 XM_017001632.1 NR_123739.1

Gene Symbol
LOC101929441
Entrez Gene ID
101929441
Gene Name
uncharacterized LOC101929441
Gene Aliases
-
UniGene
Hs.736889
Gene Chromosome Location
Chr.1: 204141408-204143009 on Build GRCh38
Transcripts
NM_001297760.1 NM_001297761.1 NM_001297762.1 NM_018208.3 XM_011509714.2 XM_011509715.1 XM_017001632.1 NR_123739.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6285276 c.741C>G p.H247Q Chr.1: 204141358-204141358 on Build GRCh38
5701007 c.761_762insA p.N254fs*5 Chr.1: 204141337-204141338 on Build GRCh38
4764423 c.712C>G p.Q238E Chr.1: 204141387-204141387 on Build GRCh38
4143249 c.671T>A p.V224E Chr.1: 204141428-204141428 on Build GRCh38
4818135 c.784G>C p.G262R Chr.1: 204141315-204141315 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs566648565 Chr.1: 204141632-204141632 on Build GRCh38
rs925670905 Chr.1: 204141585-204141585 on Build GRCh38
rs552476480 Chr.1: 204141646-204141646 on Build GRCh38
rs891736324 Chr.1: 204141631-204141631 on Build GRCh38
rs779236003 Chr.1: 204141312-204141312 on Build GRCh38
rs560866989 Chr.1: 204141533-204141533 on Build GRCh38
rs537211651 Chr.1: 204141460-204141460 on Build GRCh38
rs757371772 Chr.1: 204141280-204141280 on Build GRCh38
rs1005260658 Chr.1: 204141288-204141288 on Build GRCh38
rs770425571 Chr.1: 204141364-204141364 on Build GRCh38
rs775487389 Chr.1: 204141462-204141462 on Build GRCh38
rs180897212 Chr.1: 204141656-204141656 on Build GRCh38
rs373722402 Chr.1: 204141299-204141299 on Build GRCh38
rs780464310 Chr.1: 204141300-204141300 on Build GRCh38
rs963716254 Chr.1: 204141348-204141348 on Build GRCh38
rs1025432358 Chr.1: 204141285-204141285 on Build GRCh38
rs527365491 Chr.1: 204141514-204141514 on Build GRCh38
rs138409813 Chr.1: 204141418-204141418 on Build GRCh38
rs762982783 Chr.1: 204141464-204141464 on Build GRCh38
rs937339161 Chr.1: 204141448-204141448 on Build GRCh38
rs770497691 Chr.1: 204141441-204141441 on Build GRCh38
rs772756403 Chr.1: 204141351-204141351 on Build GRCh38
rs373228420 Chr.1: 204141446-204141446 on Build GRCh38
rs116285702 Chr.1: 204141373-204141373 on Build GRCh38
rs1024538997 Chr.1: 204141356-204141356 on Build GRCh38
rs997850814 Chr.1: 204141670-204141670 on Build GRCh38
rs755276490 Chr.1: 204141481-204141481 on Build GRCh38
rs748681339 Chr.1: 204141328-204141328 on Build GRCh38
rs746649734 Chr.1: 204141357-204141357 on Build GRCh38
rs895675506 Chr.1: 204141264-204141265 on Build GRCh38
rs752705390 Chr.1: 204141276-204141276 on Build GRCh38
rs781466580 Chr.1: 204141287-204141287 on Build GRCh38
rs35235705 Chr.1: 204141415-204141416 on Build GRCh38
rs769075813 Chr.1: 204141307-204141307 on Build GRCh38
rs755254782 Chr.1: 204141423-204141423 on Build GRCh38
rs779374402 Chr.1: 204141426-204141426 on Build GRCh38
rs955799807 Chr.1: 204141524-204141524 on Build GRCh38
rs767389208 Chr.1: 204141469-204141469 on Build GRCh38
rs758185107 Chr.1: 204141534-204141534 on Build GRCh38
rs759383121 Chr.1: 204141272-204141272 on Build GRCh38
rs199805218 Chr.1: 204141271-204141271 on Build GRCh38
rs114517582 Chr.1: 204141630-204141630 on Build GRCh38
rs888192206 Chr.1: 204141286-204141286 on Build GRCh38
rs759163609 Chr.1: 204141391-204141391 on Build GRCh38
rs753971883 Chr.1: 204141477-204141477 on Build GRCh38
rs749545728 Chr.1: 204141305-204141305 on Build GRCh38
rs766508142 Chr.1: 204141475-204141475 on Build GRCh38
rs929834403 Chr.1: 204141564-204141564 on Build GRCh38
rs766171096 Chr.1: 204141262-204141262 on Build GRCh38
rs3737657 Chr.1: 204141419-204141419 on Build GRCh38
rs993987893 Chr.1: 204141274-204141274 on Build GRCh38
rs746465912 Chr.1: 204141435-204141435 on Build GRCh38
rs927300132 Chr.1: 204141438-204141438 on Build GRCh38
rs201765091 Chr.1: 204141295-204141295 on Build GRCh38
rs1014119141 Chr.1: 204141344-204141344 on Build GRCh38
rs988933141 Chr.1: 204141407-204141407 on Build GRCh38
rs1001281648 Chr.1: 204141717-204141717 on Build GRCh38
rs763037814 Chr.1: 204141404-204141404 on Build GRCh38
rs748630621 Chr.1: 204141427-204141427 on Build GRCh38
rs767695300 Chr.1: 204141415-204141415 on Build GRCh38
rs570224117 Chr.1: 204141620-204141620 on Build GRCh38
rs534789862 Chr.1: 204141686-204141686 on Build GRCh38
rs753632325 Chr.1: 204141265-204141265 on Build GRCh38
rs34462438 Chr.1: 204141459-204141460 on Build GRCh38
rs1018363207 Chr.1: 204141293-204141293 on Build GRCh38
rs144487936 Chr.1: 204141573-204141573 on Build GRCh38
rs376360235 Chr.1: 204141289-204141289 on Build GRCh38
rs754108955 Chr.1: 204141420-204141420 on Build GRCh38
rs550461765 Chr.1: 204141586-204141586 on Build GRCh38
rs758961107 Chr.1: 204141428-204141428 on Build GRCh38
rs771539071 Chr.1: 204141331-204141331 on Build GRCh38
rs752049657 Chr.1: 204141625-204141625 on Build GRCh38
rs745574189 Chr.1: 204141449-204141449 on Build GRCh38
rs777258273 Chr.1: 204141434-204141434 on Build GRCh38
rs1030523813 Chr.1: 204141352-204141352 on Build GRCh38
rs200858943 Chr.1: 204141384-204141384 on Build GRCh38
rs942811447 Chr.1: 204141541-204141541 on Build GRCh38
rs756261638 Chr.1: 204141685-204141685 on Build GRCh38
rs776320731 Chr.1: 204141376-204141376 on Build GRCh38
rs3737656 Chr.1: 204141532-204141532 on Build GRCh38
rs760620245 Chr.1: 204141472-204141472 on Build GRCh38
rs775542456 Chr.1: 204141397-204141397 on Build GRCh38
rs762479061 Chr.1: 204141471-204141471 on Build GRCh38

View Full Product Details