Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 204149464-204149958 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ETNK2
Entrez Gene ID
55224
Gene Name
ethanolamine kinase 2
Gene Aliases
EKI2, HMFT1716
UniGene
Hs.497469
Gene Chromosome Location
Chr.1: 204131061-204152182 on Build GRCh38
Transcripts
NM_001297760.1 NM_001297761.1 NM_001297762.1 NM_018208.3 XM_011509715.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4681818 c.336G>A p.R112R Chr.1: 204149885-204149885 on Build GRCh38
5705159 c.479C>A p.A160D Chr.1: 204149742-204149742 on Build GRCh38
1317585 c.507C>T p.P169P Chr.1: 204149714-204149714 on Build GRCh38
6508509 c.327G>T p.V109V Chr.1: 204149894-204149894 on Build GRCh38
2214106 c.510G>A p.R170R Chr.1: 204149711-204149711 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs965435014 Chr.1: 204149552-204149552 on Build GRCh38
rs745615577 Chr.1: 204149805-204149805 on Build GRCh38
rs377403947 Chr.1: 204149763-204149763 on Build GRCh38
rs202185285 Chr.1: 204149871-204149871 on Build GRCh38
rs921274157 Chr.1: 204149865-204149865 on Build GRCh38
rs921572161 Chr.1: 204149574-204149574 on Build GRCh38
rs371666384 Chr.1: 204149539-204149539 on Build GRCh38
rs370190989 Chr.1: 204149862-204149862 on Build GRCh38
rs868307357 Chr.1: 204149660-204149660 on Build GRCh38
rs781729192 Chr.1: 204149798-204149798 on Build GRCh38
rs770133377 Chr.1: 204149818-204149818 on Build GRCh38
rs751819221 Chr.1: 204149886-204149886 on Build GRCh38
rs779042118 Chr.1: 204149755-204149755 on Build GRCh38
rs1050924031 Chr.1: 204149546-204149546 on Build GRCh38
rs758518203 Chr.1: 204149759-204149759 on Build GRCh38
rs774930573 Chr.1: 204149712-204149712 on Build GRCh38
rs763600102 Chr.1: 204149714-204149714 on Build GRCh38
rs372992385 Chr.1: 204149867-204149867 on Build GRCh38
rs370194168 Chr.1: 204149786-204149786 on Build GRCh38
rs528212957 Chr.1: 204149868-204149868 on Build GRCh38
rs746164408 Chr.1: 204149896-204149896 on Build GRCh38
rs370680512 Chr.1: 204149920-204149920 on Build GRCh38
rs773906422 Chr.1: 204149923-204149923 on Build GRCh38
rs776081427 Chr.1: 204149824-204149824 on Build GRCh38
rs373432247 Chr.1: 204149750-204149750 on Build GRCh38
rs1053930428 Chr.1: 204149591-204149591 on Build GRCh38
rs562912778 Chr.1: 204149700-204149700 on Build GRCh38
rs906942816 Chr.1: 204149671-204149671 on Build GRCh38
rs747789899 Chr.1: 204149931-204149931 on Build GRCh38
rs975803045 Chr.1: 204149565-204149565 on Build GRCh38
rs144504466 Chr.1: 204149721-204149721 on Build GRCh38
rs965844176 Chr.1: 204149808-204149808 on Build GRCh38
rs761499775 Chr.1: 204149843-204149843 on Build GRCh38
rs764309147 Chr.1: 204149882-204149882 on Build GRCh38
rs762324880 Chr.1: 204149825-204149825 on Build GRCh38
rs757621574 Chr.1: 204149888-204149888 on Build GRCh38
rs996164674 Chr.1: 204149538-204149538 on Build GRCh38
rs768188076 Chr.1: 204149827-204149827 on Build GRCh38
rs771641428 Chr.1: 204149933-204149933 on Build GRCh38
rs141770072 Chr.1: 204149626-204149626 on Build GRCh38
rs750335710 Chr.1: 204149731-204149731 on Build GRCh38
rs192659299 Chr.1: 204149550-204149550 on Build GRCh38
rs530687373 Chr.1: 204149917-204149917 on Build GRCh38
rs768536552 Chr.1: 204149590-204149590 on Build GRCh38
rs916291525 Chr.1: 204149647-204149647 on Build GRCh38
rs762532276 Chr.1: 204149713-204149713 on Build GRCh38
rs575012577 Chr.1: 204149606-204149606 on Build GRCh38
rs141281450 Chr.1: 204149722-204149722 on Build GRCh38
rs773965589 Chr.1: 204149834-204149834 on Build GRCh38
rs147984605 Chr.1: 204149507-204149507 on Build GRCh38
rs576441238 Chr.1: 204149726-204149726 on Build GRCh38
rs970516355 Chr.1: 204149697-204149697 on Build GRCh38
rs768128714 Chr.1: 204149922-204149922 on Build GRCh38
rs1000394921 Chr.1: 204149910-204149910 on Build GRCh38
rs200236107 Chr.1: 204149897-204149897 on Build GRCh38
rs1033244137 Chr.1: 204149924-204149924 on Build GRCh38
rs200166795 Chr.1: 204149747-204149747 on Build GRCh38
rs555182870 Chr.1: 204149555-204149555 on Build GRCh38
rs914075020 Chr.1: 204149596-204149596 on Build GRCh38
rs1008987944 Chr.1: 204149875-204149875 on Build GRCh38
rs761624943 Chr.1: 204149851-204149851 on Build GRCh38
rs1020461177 Chr.1: 204149571-204149571 on Build GRCh38
rs1041462733 Chr.1: 204149632-204149632 on Build GRCh38
rs936913582 Chr.1: 204149585-204149585 on Build GRCh38
rs188773529 Chr.1: 204149516-204149516 on Build GRCh38
rs559020342 Chr.1: 204149791-204149791 on Build GRCh38
rs982882094 Chr.1: 204149541-204149541 on Build GRCh38
rs201400955 Chr.1: 204149850-204149850 on Build GRCh38

View Full Product Details