Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 205143817-205144318 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DSTYK
Entrez Gene ID
25778
Gene Name
dual serine/threonine and tyrosine protein kinase
Gene Aliases
CAKUT1, DustyPK, HDCMD38P, RIP5, RIPK5
UniGene
Hs.6874
Gene Chromosome Location
Chr.1: 205142497-205211599 on Build GRCh38
Transcripts
NM_015375.2 NM_199462.2 XM_011509392.2 XM_011509393.2 XM_011509394.2

SNP Detail

SNP ID Chromosome Location
rs970374834 Chr.1: 205144289-205144289 on Build GRCh38
rs868170852 Chr.1: 205144055-205144055 on Build GRCh38
rs534885049 Chr.1: 205143953-205143953 on Build GRCh38
rs902888255 Chr.1: 205144103-205144103 on Build GRCh38
rs1009805851 Chr.1: 205144259-205144259 on Build GRCh38
rs917521663 Chr.1: 205143909-205143909 on Build GRCh38
rs565548936 Chr.1: 205143943-205143943 on Build GRCh38
rs991681038 Chr.1: 205144190-205144190 on Build GRCh38
rs1020152311 Chr.1: 205144053-205144053 on Build GRCh38
rs572362427 Chr.1: 205144269-205144269 on Build GRCh38
rs1032744290 Chr.1: 205144180-205144180 on Build GRCh38
rs1001374847 Chr.1: 205144193-205144193 on Build GRCh38
rs144168869 Chr.1: 205143971-205143971 on Build GRCh38
rs980516158 Chr.1: 205143860-205143860 on Build GRCh38
rs899887091 Chr.1: 205144147-205144147 on Build GRCh38
rs971588424 Chr.1: 205144223-205144223 on Build GRCh38
rs771948705 Chr.1: 205143989-205143989 on Build GRCh38
rs950315855 Chr.1: 205143916-205143916 on Build GRCh38
rs558949900 Chr.1: 205144256-205144256 on Build GRCh38
rs1032480780 Chr.1: 205144209-205144209 on Build GRCh38
rs541681829 Chr.1: 205144271-205144271 on Build GRCh38
rs926396738 Chr.1: 205143932-205143932 on Build GRCh38
rs890384146 Chr.1: 205144040-205144040 on Build GRCh38
rs114010349 Chr.1: 205144047-205144047 on Build GRCh38
rs557236533 Chr.1: 205144196-205144196 on Build GRCh38
rs568373768 Chr.1: 205143974-205143974 on Build GRCh38
rs929210565 Chr.1: 205143988-205143988 on Build GRCh38
rs919102921 Chr.1: 205143982-205143982 on Build GRCh38
rs766643516 Chr.1: 205144186-205144186 on Build GRCh38
rs148681906 Chr.1: 205144210-205144210 on Build GRCh38
rs958672159 Chr.1: 205144188-205144188 on Build GRCh38
rs999802433 Chr.1: 205144130-205144130 on Build GRCh38
rs1039686136 Chr.1: 205144066-205144066 on Build GRCh38
rs1050653832 Chr.1: 205144010-205144010 on Build GRCh38
rs899897631 Chr.1: 205143992-205143992 on Build GRCh38
rs891123265 Chr.1: 205144002-205144002 on Build GRCh38
rs561399031 Chr.1: 205144283-205144283 on Build GRCh38
rs539135588 Chr.1: 205144219-205144219 on Build GRCh38

View Full Product Details