Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 205144983-205145495 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DSTYK
Entrez Gene ID
25778
Gene Name
dual serine/threonine and tyrosine protein kinase
Gene Aliases
CAKUT1, DustyPK, HDCMD38P, RIP5, RIPK5
UniGene
Hs.6874
Gene Chromosome Location
Chr.1: 205142497-205211599 on Build GRCh38
Transcripts
NM_015375.2 NM_199462.2 XM_011509392.2 XM_011509393.2 XM_011509394.2

SNP Detail

SNP ID Chromosome Location
rs1046545686 Chr.1: 205145417-205145417 on Build GRCh38
rs372576317 Chr.1: 205145299-205145299 on Build GRCh38
rs966681657 Chr.1: 205145406-205145410 on Build GRCh38
rs1019536611 Chr.1: 205145386-205145386 on Build GRCh38
rs762933508 Chr.1: 205145118-205145118 on Build GRCh38
rs974631390 Chr.1: 205145288-205145288 on Build GRCh38
rs925359608 Chr.1: 205145407-205145407 on Build GRCh38
rs924488217 Chr.1: 205145207-205145207 on Build GRCh38
rs915809948 Chr.1: 205145239-205145239 on Build GRCh38
rs116436604 Chr.1: 205145282-205145282 on Build GRCh38
rs530781873 Chr.1: 205145344-205145344 on Build GRCh38
rs928100280 Chr.1: 205145423-205145423 on Build GRCh38
rs987193775 Chr.1: 205145264-205145264 on Build GRCh38
rs1008653325 Chr.1: 205145366-205145366 on Build GRCh38
rs957319072 Chr.1: 205145311-205145311 on Build GRCh38
rs66966420 Chr.1: 205145366-205145368 on Build GRCh38
rs986795907 Chr.1: 205145064-205145064 on Build GRCh38
rs977229923 Chr.1: 205145181-205145181 on Build GRCh38
rs898698855 Chr.1: 205145336-205145336 on Build GRCh38
rs771578397 Chr.1: 205145365-205145366 on Build GRCh38
rs1025982169 Chr.1: 205145156-205145156 on Build GRCh38
rs112300026 Chr.1: 205145284-205145284 on Build GRCh38
rs950493270 Chr.1: 205145180-205145180 on Build GRCh38
rs183813915 Chr.1: 205145232-205145232 on Build GRCh38
rs1054807860 Chr.1: 205145236-205145236 on Build GRCh38
rs775196353 Chr.1: 205145104-205145104 on Build GRCh38
rs945425913 Chr.1: 205145406-205145406 on Build GRCh38
rs1002443674 Chr.1: 205145065-205145065 on Build GRCh38
rs954210860 Chr.1: 205145014-205145014 on Build GRCh38
rs935829825 Chr.1: 205145226-205145226 on Build GRCh38
rs751829768 Chr.1: 205145270-205145270 on Build GRCh38
rs377096203 Chr.1: 205145352-205145352 on Build GRCh38
rs1029011435 Chr.1: 205145363-205145363 on Build GRCh38
rs948406176 Chr.1: 205145254-205145254 on Build GRCh38
rs757588333 Chr.1: 205145365-205145367 on Build GRCh38
rs745536542 Chr.1: 205145022-205145022 on Build GRCh38
rs995672567 Chr.1: 205145353-205145353 on Build GRCh38
rs989849919 Chr.1: 205145378-205145378 on Build GRCh38
rs913922155 Chr.1: 205145389-205145389 on Build GRCh38
rs973061773 Chr.1: 205145426-205145426 on Build GRCh38
rs769597318 Chr.1: 205145081-205145081 on Build GRCh38
rs36093419 Chr.1: 205145368-205145370 on Build GRCh38

View Full Product Details