Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 205525035-205525468 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK18
Entrez Gene ID
5129
Gene Name
cyclin dependent kinase 18
Gene Aliases
PCTAIRE, PCTAIRE3, PCTK3
UniGene
Hs.445402
Gene Chromosome Location
Chr.1: 205504556-205532793 on Build GRCh38
Transcripts
NM_002596.3 NM_212502.2 NM_212503.2 XM_011509602.2 XM_017001423.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5155169 c.400-7C>A p.? Chr.1: 205525132-205525132 on Build GRCh38
1689711 c.509A>C p.K170T Chr.1: 205525158-205525158 on Build GRCh38
902784 c.446A>G p.K149R Chr.1: 205525185-205525185 on Build GRCh38
3360601 c.491C>T p.S164L Chr.1: 205525140-205525140 on Build GRCh38
3482231 c.417G>A p.G139G Chr.1: 205525156-205525156 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs551228194 Chr.1: 205525286-205525286 on Build GRCh38
rs768293216 Chr.1: 205525195-205525195 on Build GRCh38
rs563457421 Chr.1: 205525069-205525069 on Build GRCh38
rs999703103 Chr.1: 205525400-205525400 on Build GRCh38
rs1019235503 Chr.1: 205525388-205525388 on Build GRCh38
rs916700073 Chr.1: 205525268-205525268 on Build GRCh38
rs528433360 Chr.1: 205525173-205525173 on Build GRCh38
rs763436941 Chr.1: 205525169-205525169 on Build GRCh38
rs779830921 Chr.1: 205525114-205525114 on Build GRCh38
rs907152898 Chr.1: 205525077-205525077 on Build GRCh38
rs757726515 Chr.1: 205525099-205525099 on Build GRCh38
rs765487740 Chr.1: 205525157-205525157 on Build GRCh38
rs982175014 Chr.1: 205525275-205525275 on Build GRCh38
rs12126276 Chr.1: 205525380-205525380 on Build GRCh38
rs773071824 Chr.1: 205525279-205525279 on Build GRCh38
rs745331977 Chr.1: 205525106-205525106 on Build GRCh38
rs1032439041 Chr.1: 205525250-205525250 on Build GRCh38
rs555266035 Chr.1: 205525423-205525423 on Build GRCh38
rs570933501 Chr.1: 205525323-205525323 on Build GRCh38
rs543977031 Chr.1: 205525066-205525066 on Build GRCh38
rs550087546 Chr.1: 205525087-205525087 on Build GRCh38
rs869158349 Chr.1: 205525059-205525059 on Build GRCh38
rs769816139 Chr.1: 205525313-205525313 on Build GRCh38
rs374908323 Chr.1: 205525113-205525113 on Build GRCh38
rs748849535 Chr.1: 205525065-205525065 on Build GRCh38
rs41310919 Chr.1: 205525283-205525283 on Build GRCh38
rs749025147 Chr.1: 205525096-205525096 on Build GRCh38
rs771925553 Chr.1: 205525140-205525140 on Build GRCh38
rs376187770 Chr.1: 205525247-205525247 on Build GRCh38
rs779821232 Chr.1: 205525186-205525186 on Build GRCh38
rs73074319 Chr.1: 205525203-205525203 on Build GRCh38
rs368482220 Chr.1: 205525202-205525202 on Build GRCh38
rs970797698 Chr.1: 205525441-205525441 on Build GRCh38
rs902399893 Chr.1: 205525289-205525289 on Build GRCh38
rs147619411 Chr.1: 205525147-205525147 on Build GRCh38
rs763126471 Chr.1: 205525327-205525327 on Build GRCh38
rs1012986218 Chr.1: 205525339-205525339 on Build GRCh38
rs566473323 Chr.1: 205525391-205525391 on Build GRCh38
rs752861306 Chr.1: 205525161-205525161 on Build GRCh38
rs914892718 Chr.1: 205525257-205525257 on Build GRCh38
rs958187738 Chr.1: 205525407-205525407 on Build GRCh38
rs770683815 Chr.1: 205525144-205525144 on Build GRCh38
rs547469900 Chr.1: 205525345-205525345 on Build GRCh38
rs958212253 Chr.1: 205525255-205525255 on Build GRCh38
rs774401348 Chr.1: 205525124-205525124 on Build GRCh38
rs552356615 Chr.1: 205525440-205525441 on Build GRCh38
rs780855607 Chr.1: 205525100-205525100 on Build GRCh38
rs367572683 Chr.1: 205525146-205525146 on Build GRCh38
rs750000988 Chr.1: 205525182-205525182 on Build GRCh38
rs978315556 Chr.1: 205525240-205525240 on Build GRCh38
rs749146099 Chr.1: 205525122-205525122 on Build GRCh38
rs779145441 Chr.1: 205525121-205525122 on Build GRCh38
rs939846581 Chr.1: 205525090-205525090 on Build GRCh38
rs1008304047 Chr.1: 205525386-205525386 on Build GRCh38
rs76098475 Chr.1: 205525440-205525440 on Build GRCh38
rs1015828322 Chr.1: 205525177-205525177 on Build GRCh38
rs778624604 Chr.1: 205525190-205525190 on Build GRCh38
rs778573693 Chr.1: 205525198-205525198 on Build GRCh38
rs1056293898 Chr.1: 205525309-205525309 on Build GRCh38
rs531865489 Chr.1: 205525172-205525172 on Build GRCh38
rs187930431 Chr.1: 205525405-205525405 on Build GRCh38
rs746097970 Chr.1: 205525161-205525162 on Build GRCh38
rs768151516 Chr.1: 205525393-205525393 on Build GRCh38
rs371259169 Chr.1: 205525181-205525181 on Build GRCh38
rs12122797 Chr.1: 205525384-205525384 on Build GRCh38
rs932408870 Chr.1: 205525221-205525221 on Build GRCh38
rs12126241 Chr.1: 205525269-205525269 on Build GRCh38
rs1029826875 Chr.1: 205525424-205525424 on Build GRCh38
rs939279770 Chr.1: 205525292-205525292 on Build GRCh38
rs80154131 Chr.1: 205525441-205525441 on Build GRCh38
rs577303024 Chr.1: 205525076-205525076 on Build GRCh38
rs1024137899 Chr.1: 205525439-205525439 on Build GRCh38
rs761666709 Chr.1: 205525133-205525133 on Build GRCh38
rs865936804 Chr.1: 205525406-205525406 on Build GRCh38
rs115244221 Chr.1: 205525171-205525171 on Build GRCh38
rs939921504 Chr.1: 205525310-205525310 on Build GRCh38
rs538917059 Chr.1: 205525230-205525230 on Build GRCh38
rs1046048207 Chr.1: 205525068-205525068 on Build GRCh38
rs768330546 Chr.1: 205525123-205525123 on Build GRCh38

View Full Product Details