Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 205526695-205527188 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK18
Entrez Gene ID
5129
Gene Name
cyclin dependent kinase 18
Gene Aliases
PCTAIRE, PCTAIRE3, PCTK3
UniGene
Hs.445402
Gene Chromosome Location
Chr.1: 205504556-205532793 on Build GRCh38
Transcripts
NM_002596.3 NM_212502.2 NM_212503.2 XM_011509602.2 XM_017001423.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5506767 c.759C>T p.D253D Chr.1: 205526777-205526777 on Build GRCh38
677938 c.670A>T p.S224C Chr.1: 205526778-205526778 on Build GRCh38
6625458 c.724G>A p.V242I Chr.1: 205526832-205526832 on Build GRCh38
5506766 c.669C>T p.D223D Chr.1: 205526777-205526777 on Build GRCh38
7097468 c.672T>C p.S224S Chr.1: 205526780-205526780 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs901824960 Chr.1: 205527009-205527009 on Build GRCh38
rs34804445 Chr.1: 205526762-205526763 on Build GRCh38
rs751556589 Chr.1: 205526762-205526762 on Build GRCh38
rs964097055 Chr.1: 205526954-205526954 on Build GRCh38
rs773077844 Chr.1: 205526728-205526728 on Build GRCh38
rs531221690 Chr.1: 205526955-205526955 on Build GRCh38
rs1005983626 Chr.1: 205527098-205527098 on Build GRCh38
rs949552149 Chr.1: 205527045-205527045 on Build GRCh38
rs972014583 Chr.1: 205526853-205526853 on Build GRCh38
rs537824587 Chr.1: 205526813-205526813 on Build GRCh38
rs765039969 Chr.1: 205526857-205526857 on Build GRCh38
rs768728549 Chr.1: 205527157-205527157 on Build GRCh38
rs935585473 Chr.1: 205526986-205526986 on Build GRCh38
rs776504448 Chr.1: 205526851-205526851 on Build GRCh38
rs555855091 Chr.1: 205526729-205526729 on Build GRCh38
rs972290774 Chr.1: 205526904-205526904 on Build GRCh38
rs956528640 Chr.1: 205526795-205526795 on Build GRCh38
rs1544154 Chr.1: 205526848-205526848 on Build GRCh38
rs781249169 Chr.1: 205526781-205526781 on Build GRCh38
rs56085951 Chr.1: 205527156-205527156 on Build GRCh38
rs538954377 Chr.1: 205527091-205527091 on Build GRCh38
rs200375022 Chr.1: 205526831-205526831 on Build GRCh38
rs76552836 Chr.1: 205526760-205526760 on Build GRCh38
rs1037369456 Chr.1: 205527116-205527116 on Build GRCh38
rs77517454 Chr.1: 205527060-205527060 on Build GRCh38
rs542324421 Chr.1: 205526888-205526888 on Build GRCh38
rs191322853 Chr.1: 205526847-205526847 on Build GRCh38
rs772705853 Chr.1: 205526845-205526845 on Build GRCh38
rs779447272 Chr.1: 205527067-205527067 on Build GRCh38
rs562215279 Chr.1: 205526946-205526946 on Build GRCh38
rs993401187 Chr.1: 205527136-205527136 on Build GRCh38
rs751709892 Chr.1: 205526879-205526879 on Build GRCh38
rs200025630 Chr.1: 205526855-205526855 on Build GRCh38
rs1052727788 Chr.1: 205527007-205527007 on Build GRCh38
rs12566558 Chr.1: 205527079-205527079 on Build GRCh38
rs368233715 Chr.1: 205526880-205526880 on Build GRCh38
rs767036197 Chr.1: 205526723-205526723 on Build GRCh38
rs74141244 Chr.1: 205527008-205527008 on Build GRCh38
rs777586549 Chr.1: 205526926-205526926 on Build GRCh38
rs756468597 Chr.1: 205526810-205526810 on Build GRCh38
rs897518856 Chr.1: 205527133-205527133 on Build GRCh38
rs764276589 Chr.1: 205526761-205526761 on Build GRCh38
rs757526160 Chr.1: 205526777-205526777 on Build GRCh38
rs530113323 Chr.1: 205526796-205526796 on Build GRCh38
rs910729768 Chr.1: 205526987-205526987 on Build GRCh38
rs866894634 Chr.1: 205526766-205526766 on Build GRCh38
rs749808161 Chr.1: 205526814-205526814 on Build GRCh38
rs979365300 Chr.1: 205526963-205526963 on Build GRCh38
rs773942682 Chr.1: 205526832-205526832 on Build GRCh38
rs1052298141 Chr.1: 205527046-205527046 on Build GRCh38
rs376270292 Chr.1: 205526843-205526843 on Build GRCh38
rs751336526 Chr.1: 205526951-205526951 on Build GRCh38
rs551124596 Chr.1: 205526978-205526978 on Build GRCh38
rs750055340 Chr.1: 205526906-205526906 on Build GRCh38
rs954596305 Chr.1: 205526856-205526856 on Build GRCh38
rs369627956 Chr.1: 205526741-205526741 on Build GRCh38
rs150207562 Chr.1: 205527027-205527028 on Build GRCh38
rs766456677 Chr.1: 205526730-205526730 on Build GRCh38
rs757249129 Chr.1: 205526984-205526984 on Build GRCh38
rs761531070 Chr.1: 205526719-205526719 on Build GRCh38

View Full Product Details