Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 205600996-205601506 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELK4
Entrez Gene ID
2005
Gene Name
ELK4, ETS transcription factor
Gene Aliases
SAP1
UniGene
Hs.497520
Gene Chromosome Location
Chr.1: 205597774-205634077 on Build GRCh38
Transcripts
NM_181644.4 XM_011509229.2

Gene Symbol
MFSD4A
Entrez Gene ID
148808
Gene Name
major facilitator superfamily domain containing 4A
Gene Aliases
MFSD4, UNQ3064
UniGene
Hs.567714
Gene Chromosome Location
Chr.1: 205568984-205602918 on Build GRCh38
Transcripts
NM_181644.4 XM_011509229.2

SNP Detail

SNP ID Chromosome Location
rs906408212 Chr.1: 205601271-205601271 on Build GRCh38
rs953241649 Chr.1: 205601028-205601028 on Build GRCh38
rs1036085502 Chr.1: 205601299-205601299 on Build GRCh38
rs765664244 Chr.1: 205601199-205601199 on Build GRCh38
rs997552565 Chr.1: 205601349-205601349 on Build GRCh38
rs573965271 Chr.1: 205601424-205601425 on Build GRCh38
rs368756956 Chr.1: 205601249-205601249 on Build GRCh38
rs537046399 Chr.1: 205601470-205601470 on Build GRCh38
rs960960437 Chr.1: 205601172-205601172 on Build GRCh38
rs187936562 Chr.1: 205601080-205601080 on Build GRCh38
rs1021408719 Chr.1: 205601042-205601042 on Build GRCh38
rs113513015 Chr.1: 205601344-205601344 on Build GRCh38
rs7552552 Chr.1: 205601468-205601468 on Build GRCh38
rs924717432 Chr.1: 205601095-205601095 on Build GRCh38
rs68008371 Chr.1: 205601150-205601155 on Build GRCh38
rs1049292265 Chr.1: 205601247-205601247 on Build GRCh38
rs4951245 Chr.1: 205601145-205601145 on Build GRCh38
rs753181664 Chr.1: 205601212-205601212 on Build GRCh38
rs771717157 Chr.1: 205601124-205601124 on Build GRCh38
rs764376710 Chr.1: 205601413-205601413 on Build GRCh38
rs369396782 Chr.1: 205601434-205601434 on Build GRCh38
rs916777037 Chr.1: 205601213-205601213 on Build GRCh38
rs957515418 Chr.1: 205601466-205601466 on Build GRCh38
rs112574875 Chr.1: 205601359-205601359 on Build GRCh38
rs12134829 Chr.1: 205601278-205601278 on Build GRCh38
rs550904376 Chr.1: 205601477-205601477 on Build GRCh38
rs978130806 Chr.1: 205601105-205601105 on Build GRCh38
rs977520927 Chr.1: 205601083-205601083 on Build GRCh38
rs1011437809 Chr.1: 205601395-205601395 on Build GRCh38
rs888824097 Chr.1: 205601375-205601375 on Build GRCh38
rs568118751 Chr.1: 205601464-205601464 on Build GRCh38
rs966672403 Chr.1: 205601416-205601416 on Build GRCh38
rs994616336 Chr.1: 205601338-205601338 on Build GRCh38
rs992429245 Chr.1: 205601182-205601182 on Build GRCh38
rs7552438 Chr.1: 205601308-205601308 on Build GRCh38
rs866612071 Chr.1: 205601292-205601292 on Build GRCh38
rs115314573 Chr.1: 205601244-205601244 on Build GRCh38
rs528604046 Chr.1: 205601392-205601392 on Build GRCh38
rs939198797 Chr.1: 205601273-205601273 on Build GRCh38
rs1019609307 Chr.1: 205601048-205601048 on Build GRCh38
rs897590520 Chr.1: 205601330-205601330 on Build GRCh38
rs769986480 Chr.1: 205601256-205601256 on Build GRCh38
rs953144851 Chr.1: 205601021-205601021 on Build GRCh38
rs977573452 Chr.1: 205601461-205601461 on Build GRCh38
rs763319743 Chr.1: 205601319-205601319 on Build GRCh38
rs76005485 Chr.1: 205601030-205601030 on Build GRCh38

View Full Product Details