Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 205608194-205608701 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELK4
Entrez Gene ID
2005
Gene Name
ELK4, ETS transcription factor
Gene Aliases
SAP1
UniGene
Hs.497520
Gene Chromosome Location
Chr.1: 205597774-205634077 on Build GRCh38
Transcripts
NM_001973.3

SNP Detail

SNP ID Chromosome Location
rs1039195009 Chr.1: 205608281-205608281 on Build GRCh38
rs1024266545 Chr.1: 205608348-205608348 on Build GRCh38
rs914768392 Chr.1: 205608282-205608282 on Build GRCh38
rs12760743 Chr.1: 205608622-205608622 on Build GRCh38
rs535550124 Chr.1: 205608628-205608628 on Build GRCh38
rs943574696 Chr.1: 205608268-205608268 on Build GRCh38
rs938788718 Chr.1: 205608312-205608312 on Build GRCh38
rs1053210916 Chr.1: 205608307-205608307 on Build GRCh38
rs955889446 Chr.1: 205608242-205608242 on Build GRCh38
rs567417427 Chr.1: 205608570-205608570 on Build GRCh38
rs181630769 Chr.1: 205608452-205608452 on Build GRCh38
rs929574983 Chr.1: 205608227-205608227 on Build GRCh38
rs912179808 Chr.1: 205608524-205608524 on Build GRCh38
rs1026282143 Chr.1: 205608463-205608463 on Build GRCh38
rs900339838 Chr.1: 205608509-205608509 on Build GRCh38
rs1030035801 Chr.1: 205608237-205608237 on Build GRCh38
rs968982453 Chr.1: 205608580-205608580 on Build GRCh38
rs935756880 Chr.1: 205608624-205608624 on Build GRCh38
rs117360919 Chr.1: 205608555-205608555 on Build GRCh38
rs1016748173 Chr.1: 205608442-205608442 on Build GRCh38
rs912066847 Chr.1: 205608240-205608240 on Build GRCh38
rs963812462 Chr.1: 205608445-205608445 on Build GRCh38
rs759393647 Chr.1: 205608649-205608649 on Build GRCh38
rs1001800223 Chr.1: 205608365-205608365 on Build GRCh38
rs140225200 Chr.1: 205608302-205608302 on Build GRCh38
rs188826859 Chr.1: 205608285-205608285 on Build GRCh38
rs572111115 Chr.1: 205608477-205608477 on Build GRCh38
rs980295565 Chr.1: 205608300-205608300 on Build GRCh38
rs980347844 Chr.1: 205608583-205608583 on Build GRCh38
rs987651098 Chr.1: 205608239-205608239 on Build GRCh38
rs905768319 Chr.1: 205608353-205608353 on Build GRCh38
rs987432693 Chr.1: 205608500-205608500 on Build GRCh38
rs887669190 Chr.1: 205608473-205608473 on Build GRCh38
rs1021891363 Chr.1: 205608554-205608554 on Build GRCh38
rs145604148 Chr.1: 205608530-205608530 on Build GRCh38
rs749627901 Chr.1: 205608618-205608618 on Build GRCh38
rs992930294 Chr.1: 205608648-205608649 on Build GRCh38
rs919234143 Chr.1: 205608225-205608225 on Build GRCh38
rs989083919 Chr.1: 205608272-205608272 on Build GRCh38
rs530807987 Chr.1: 205608339-205608339 on Build GRCh38
rs988858659 Chr.1: 205608545-205608545 on Build GRCh38
rs1014183651 Chr.1: 205608340-205608340 on Build GRCh38
rs547696431 Chr.1: 205608481-205608481 on Build GRCh38
rs761218075 Chr.1: 205608234-205608234 on Build GRCh38
rs776106096 Chr.1: 205608525-205608525 on Build GRCh38
rs955734841 Chr.1: 205608535-205608535 on Build GRCh38
rs567036153 Chr.1: 205608595-205608595 on Build GRCh38

View Full Product Details