Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 205611086-205611597 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELK4
Entrez Gene ID
2005
Gene Name
ELK4, ETS transcription factor
Gene Aliases
SAP1
UniGene
Hs.497520
Gene Chromosome Location
Chr.1: 205597774-205634077 on Build GRCh38
Transcripts
NM_001973.3

SNP Detail

SNP ID Chromosome Location
rs900911991 Chr.1: 205611395-205611395 on Build GRCh38
rs1042516755 Chr.1: 205611115-205611115 on Build GRCh38
rs529953239 Chr.1: 205611366-205611366 on Build GRCh38
rs893734039 Chr.1: 205611450-205611450 on Build GRCh38
rs1055155221 Chr.1: 205611153-205611153 on Build GRCh38
rs987075422 Chr.1: 205611378-205611378 on Build GRCh38
rs1020810460 Chr.1: 205611501-205611501 on Build GRCh38
rs761164535 Chr.1: 205611210-205611210 on Build GRCh38
rs1027976711 Chr.1: 205611315-205611315 on Build GRCh38
rs1027971897 Chr.1: 205611442-205611442 on Build GRCh38
rs369904737 Chr.1: 205611433-205611433 on Build GRCh38
rs538816886 Chr.1: 205611451-205611451 on Build GRCh38
rs1048555007 Chr.1: 205611322-205611322 on Build GRCh38
rs907053181 Chr.1: 205611190-205611190 on Build GRCh38
rs4573566 Chr.1: 205611188-205611188 on Build GRCh38
rs936727558 Chr.1: 205611150-205611150 on Build GRCh38
rs143311314 Chr.1: 205611530-205611530 on Build GRCh38
rs925424069 Chr.1: 205611489-205611489 on Build GRCh38
rs751949593 Chr.1: 205611119-205611119 on Build GRCh38
rs571746997 Chr.1: 205611473-205611473 on Build GRCh38
rs549998652 Chr.1: 205611412-205611412 on Build GRCh38
rs769416754 Chr.1: 205611307-205611307 on Build GRCh38
rs887243163 Chr.1: 205611331-205611331 on Build GRCh38
rs916170407 Chr.1: 205611304-205611304 on Build GRCh38
rs913714442 Chr.1: 205611124-205611124 on Build GRCh38
rs966497215 Chr.1: 205611512-205611512 on Build GRCh38
rs949390090 Chr.1: 205611510-205611510 on Build GRCh38
rs752544709 Chr.1: 205611369-205611369 on Build GRCh38
rs756791680 Chr.1: 205611120-205611120 on Build GRCh38
rs183927335 Chr.1: 205611248-205611248 on Build GRCh38
rs749113897 Chr.1: 205611413-205611413 on Build GRCh38
rs138606042 Chr.1: 205611223-205611223 on Build GRCh38
rs558004580 Chr.1: 205611468-205611468 on Build GRCh38
rs564624843 Chr.1: 205611161-205611161 on Build GRCh38
rs907141366 Chr.1: 205611529-205611529 on Build GRCh38
rs1004424361 Chr.1: 205611220-205611220 on Build GRCh38
rs1004066483 Chr.1: 205611574-205611574 on Build GRCh38
rs953807484 Chr.1: 205611330-205611330 on Build GRCh38
rs779867264 Chr.1: 205611314-205611314 on Build GRCh38
rs561245393 Chr.1: 205611429-205611429 on Build GRCh38
rs978285712 Chr.1: 205611449-205611449 on Build GRCh38
rs1040811618 Chr.1: 205611388-205611388 on Build GRCh38
rs777208327 Chr.1: 205611557-205611557 on Build GRCh38
rs895147123 Chr.1: 205611159-205611159 on Build GRCh38
rs981940786 Chr.1: 205611543-205611543 on Build GRCh38
rs936779681 Chr.1: 205611496-205611496 on Build GRCh38
rs1010799837 Chr.1: 205611455-205611455 on Build GRCh38
rs973789673 Chr.1: 205611290-205611290 on Build GRCh38
rs1055207460 Chr.1: 205611499-205611499 on Build GRCh38

View Full Product Details