Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 206013985-206013476 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTSE
Entrez Gene ID
1510
Gene Name
cathepsin E
Gene Aliases
CATE
UniGene
Hs.644082
Gene Chromosome Location
Chr.1: 206009264-206023909 on Build GRCh38
Transcripts
NM_001317331.1 NM_001910.3 NM_148964.2 XM_011509244.1 XM_011509245.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6060871 c.776C>T p.A259V Chr.1: 206013781-206013781 on Build GRCh38
4964975 c.680C>T p.A227V Chr.1: 206013877-206013877 on Build GRCh38
902911 c.708C>T p.Y236Y Chr.1: 206013849-206013849 on Build GRCh38
5934275 c.728G>A p.G243E Chr.1: 206013829-206013829 on Build GRCh38
3482389 c.725C>T p.S242F Chr.1: 206013832-206013832 on Build GRCh38
View More

View Full Product Details