Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 206909509-206910017 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCMR
Entrez Gene ID
9214
Gene Name
Fc fragment of IgM receptor
Gene Aliases
FAIM3, TOSO
UniGene
Hs.58831
Gene Chromosome Location
Chr.1: 206903282-206922033 on Build GRCh38
Transcripts
NM_001142473.1 NM_001193338.1 NM_005449.4 XM_005273351.3 XM_005273352.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4831340 c.973G>A p.A325T Chr.1: 206909737-206909737 on Build GRCh38
6306753 c.987C>A p.G329G Chr.1: 206909519-206909519 on Build GRCh38
5580995 c.994G>A p.E332K Chr.1: 206909512-206909512 on Build GRCh38
6369657 c.883G>T p.A295S Chr.1: 206909827-206909827 on Build GRCh38
40713 c.885C>T p.A295A Chr.1: 206909825-206909825 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1039020260 Chr.1: 206909770-206909770 on Build GRCh38
rs35215596 Chr.1: 206909801-206909801 on Build GRCh38
rs539473682 Chr.1: 206909607-206909607 on Build GRCh38
rs779209742 Chr.1: 206909739-206909739 on Build GRCh38
rs773235214 Chr.1: 206909778-206909778 on Build GRCh38
rs1049576905 Chr.1: 206909531-206909531 on Build GRCh38
rs1010553544 Chr.1: 206909626-206909626 on Build GRCh38
rs1035493813 Chr.1: 206909605-206909605 on Build GRCh38
rs746477181 Chr.1: 206909996-206909996 on Build GRCh38
rs1015567819 Chr.1: 206909874-206909874 on Build GRCh38
rs771490277 Chr.1: 206909770-206909772 on Build GRCh38
rs763329858 Chr.1: 206909803-206909803 on Build GRCh38
rs1056457856 Chr.1: 206909793-206909793 on Build GRCh38
rs767870461 Chr.1: 206909884-206909884 on Build GRCh38
rs555432739 Chr.1: 206909697-206909697 on Build GRCh38
rs34609188 Chr.1: 206909790-206909790 on Build GRCh38
rs34917723 Chr.1: 206909747-206909747 on Build GRCh38
rs962332990 Chr.1: 206909885-206909885 on Build GRCh38
rs922316240 Chr.1: 206909703-206909703 on Build GRCh38
rs1039464170 Chr.1: 206909569-206909569 on Build GRCh38
rs887763080 Chr.1: 206909543-206909543 on Build GRCh38
rs902479417 Chr.1: 206909582-206909582 on Build GRCh38
rs756501219 Chr.1: 206909717-206909717 on Build GRCh38
rs528590548 Chr.1: 206909828-206909828 on Build GRCh38
rs952251811 Chr.1: 206909667-206909667 on Build GRCh38
rs34450712 Chr.1: 206909749-206909749 on Build GRCh38
rs918083375 Chr.1: 206909734-206909734 on Build GRCh38
rs1043698268 Chr.1: 206909573-206909573 on Build GRCh38
rs544145639 Chr.1: 206909733-206909733 on Build GRCh38
rs1006346624 Chr.1: 206909855-206909855 on Build GRCh38
rs946541423 Chr.1: 206909751-206909751 on Build GRCh38
rs771946459 Chr.1: 206909758-206909758 on Build GRCh38
rs768080850 Chr.1: 206909889-206909889 on Build GRCh38
rs1023738786 Chr.1: 206909654-206909654 on Build GRCh38
rs1021569785 Chr.1: 206909641-206909641 on Build GRCh38
rs192734594 Chr.1: 206909790-206909790 on Build GRCh38
rs2306711 Chr.1: 206909699-206909699 on Build GRCh38
rs773003045 Chr.1: 206909971-206909971 on Build GRCh38
rs867577370 Chr.1: 206909893-206909893 on Build GRCh38
rs976764669 Chr.1: 206909918-206909918 on Build GRCh38
rs572935194 Chr.1: 206909621-206909621 on Build GRCh38
rs535429372 Chr.1: 206909680-206909680 on Build GRCh38
rs908721369 Chr.1: 206909812-206909813 on Build GRCh38
rs755335834 Chr.1: 206909700-206909700 on Build GRCh38
rs749750564 Chr.1: 206909757-206909757 on Build GRCh38
rs955248741 Chr.1: 206909966-206909966 on Build GRCh38
rs757751118 Chr.1: 206909813-206909813 on Build GRCh38
rs992570170 Chr.1: 206909732-206909732 on Build GRCh38
rs546416340 Chr.1: 206909796-206909796 on Build GRCh38
rs2306712 Chr.1: 206909615-206909615 on Build GRCh38
rs34131092 Chr.1: 206909891-206909891 on Build GRCh38
rs781598082 Chr.1: 206909708-206909708 on Build GRCh38
rs887911195 Chr.1: 206909827-206909827 on Build GRCh38
rs773792868 Chr.1: 206909736-206909736 on Build GRCh38
rs2306713 Chr.1: 206909564-206909564 on Build GRCh38
rs747819766 Chr.1: 206909771-206909771 on Build GRCh38
rs536970105 Chr.1: 206909543-206909543 on Build GRCh38
rs972443465 Chr.1: 206909701-206909701 on Build GRCh38
rs1012771352 Chr.1: 206909623-206909623 on Build GRCh38
rs889647198 Chr.1: 206909535-206909535 on Build GRCh38
rs1043626156 Chr.1: 206909784-206909784 on Build GRCh38
rs910976263 Chr.1: 206909986-206909986 on Build GRCh38
rs570564526 Chr.1: 206909598-206909598 on Build GRCh38
rs746257805 Chr.1: 206909711-206909711 on Build GRCh38
rs559575559 Chr.1: 206909808-206909808 on Build GRCh38
rs566545723 Chr.1: 206909936-206909937 on Build GRCh38
rs969051369 Chr.1: 206909662-206909662 on Build GRCh38
rs909443101 Chr.1: 206909765-206909765 on Build GRCh38
rs760340568 Chr.1: 206909788-206909788 on Build GRCh38
rs182357489 Chr.1: 206909974-206909974 on Build GRCh38
rs748945365 Chr.1: 206909735-206909735 on Build GRCh38
rs1033967565 Chr.1: 206909871-206909871 on Build GRCh38
rs563855490 Chr.1: 206909761-206909761 on Build GRCh38
rs1029328104 Chr.1: 206909955-206909955 on Build GRCh38
rs904399542 Chr.1: 206909825-206909825 on Build GRCh38
rs554628219 Chr.1: 206909572-206909572 on Build GRCh38
rs1033977400 Chr.1: 206909671-206909671 on Build GRCh38
rs779631663 Chr.1: 206909719-206909719 on Build GRCh38
rs867145899 Chr.1: 206909820-206909820 on Build GRCh38

View Full Product Details