Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 206912850-206913345 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCMR
Entrez Gene ID
9214
Gene Name
Fc fragment of IgM receptor
Gene Aliases
FAIM3, TOSO
UniGene
Hs.58831
Gene Chromosome Location
Chr.1: 206903282-206922033 on Build GRCh38
Transcripts
NM_001142473.1 NM_001193338.1 NM_005449.4 XM_005273351.3 XM_005273352.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4832276 c.467C>T p.S156F Chr.1: 206912949-206912949 on Build GRCh38
212961 c.427C>T p.H143Y Chr.1: 206912989-206912989 on Build GRCh38
4950408 c.423G>T p.W141C Chr.1: 206912993-206912993 on Build GRCh38
6349302 c.397C>A p.Q133K Chr.1: 206913019-206913019 on Build GRCh38
232281 c.391G>A p.E131K Chr.1: 206913025-206913025 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1034537017 Chr.1: 206913056-206913056 on Build GRCh38
rs192041098 Chr.1: 206913258-206913258 on Build GRCh38
rs970470470 Chr.1: 206913298-206913298 on Build GRCh38
rs144375619 Chr.1: 206913142-206913142 on Build GRCh38
rs921784343 Chr.1: 206913287-206913287 on Build GRCh38
rs762185043 Chr.1: 206913048-206913051 on Build GRCh38
rs199994920 Chr.1: 206912939-206912939 on Build GRCh38
rs764992422 Chr.1: 206912986-206912986 on Build GRCh38
rs935930226 Chr.1: 206913295-206913295 on Build GRCh38
rs776007536 Chr.1: 206912929-206912929 on Build GRCh38
rs141485983 Chr.1: 206912989-206912989 on Build GRCh38
rs749787467 Chr.1: 206913037-206913037 on Build GRCh38
rs982028429 Chr.1: 206912960-206912961 on Build GRCh38
rs1054812785 Chr.1: 206913300-206913300 on Build GRCh38
rs181999904 Chr.1: 206913170-206913170 on Build GRCh38
rs1010312106 Chr.1: 206912972-206912972 on Build GRCh38
rs542951705 Chr.1: 206913059-206913059 on Build GRCh38
rs944492877 Chr.1: 206913211-206913211 on Build GRCh38
rs767239321 Chr.1: 206912943-206912943 on Build GRCh38
rs757522550 Chr.1: 206912900-206912900 on Build GRCh38
rs578060627 Chr.1: 206912934-206912934 on Build GRCh38
rs150066058 Chr.1: 206912975-206912975 on Build GRCh38
rs749762020 Chr.1: 206912933-206912933 on Build GRCh38
rs753735963 Chr.1: 206912895-206912895 on Build GRCh38
rs529505480 Chr.1: 206913183-206913183 on Build GRCh38
rs776051011 Chr.1: 206913086-206913086 on Build GRCh38
rs974756885 Chr.1: 206913120-206913120 on Build GRCh38
rs139474303 Chr.1: 206912957-206912957 on Build GRCh38
rs769251903 Chr.1: 206913219-206913219 on Build GRCh38
rs753811648 Chr.1: 206912971-206912971 on Build GRCh38
rs1011940016 Chr.1: 206913245-206913245 on Build GRCh38
rs745917338 Chr.1: 206912908-206912908 on Build GRCh38
rs564977515 Chr.1: 206913135-206913135 on Build GRCh38
rs750622888 Chr.1: 206913019-206913019 on Build GRCh38
rs756152640 Chr.1: 206913026-206913026 on Build GRCh38
rs974560422 Chr.1: 206913230-206913230 on Build GRCh38
rs780062992 Chr.1: 206913033-206913033 on Build GRCh38
rs957725683 Chr.1: 206913190-206913190 on Build GRCh38
rs1001678575 Chr.1: 206912994-206912994 on Build GRCh38
rs544915615 Chr.1: 206913122-206913122 on Build GRCh38
rs759433646 Chr.1: 206913087-206913087 on Build GRCh38
rs769784881 Chr.1: 206912911-206912911 on Build GRCh38
rs781213334 Chr.1: 206912904-206912904 on Build GRCh38
rs962921043 Chr.1: 206913072-206913072 on Build GRCh38
rs537908315 Chr.1: 206912896-206912896 on Build GRCh38
rs369159077 Chr.1: 206913013-206913013 on Build GRCh38
rs902392359 Chr.1: 206913169-206913169 on Build GRCh38
rs765816162 Chr.1: 206912970-206912970 on Build GRCh38
rs931367552 Chr.1: 206913100-206913100 on Build GRCh38
rs916059713 Chr.1: 206912953-206912953 on Build GRCh38
rs558220717 Chr.1: 206912898-206912898 on Build GRCh38
rs1032006499 Chr.1: 206913185-206913185 on Build GRCh38
rs760177968 Chr.1: 206912960-206912960 on Build GRCh38
rs990566572 Chr.1: 206912884-206912884 on Build GRCh38
rs538415313 Chr.1: 206913308-206913308 on Build GRCh38
rs546962122 Chr.1: 206913158-206913158 on Build GRCh38
rs748476186 Chr.1: 206913065-206913065 on Build GRCh38
rs754573214 Chr.1: 206913085-206913085 on Build GRCh38
rs774906493 Chr.1: 206912938-206912938 on Build GRCh38
rs186426258 Chr.1: 206913248-206913248 on Build GRCh38
rs1041277549 Chr.1: 206913167-206913167 on Build GRCh38
rs769328235 Chr.1: 206913038-206913038 on Build GRCh38
rs762676860 Chr.1: 206913091-206913091 on Build GRCh38
rs758215939 Chr.1: 206913007-206913007 on Build GRCh38
rs772579482 Chr.1: 206913071-206913071 on Build GRCh38
rs766291673 Chr.1: 206912888-206912888 on Build GRCh38
rs188217894 Chr.1: 206913080-206913080 on Build GRCh38
rs765117250 Chr.1: 206913088-206913088 on Build GRCh38
rs576182203 Chr.1: 206913082-206913082 on Build GRCh38
rs754157576 Chr.1: 206912892-206912892 on Build GRCh38

View Full Product Details