Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 207019249-207019757 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf116
Entrez Gene ID
79098
Gene Name
chromosome 1 open reading frame 116
Gene Aliases
SARG
UniGene
Hs.32417
Gene Chromosome Location
Chr.1: 207018521-207032761 on Build GRCh38
Transcripts
NM_001083924.1 NM_023938.5 XM_005273259.1 XM_006711530.1 XM_011509973.2

SNP Detail

SNP ID Chromosome Location
rs943644780 Chr.1: 207019691-207019691 on Build GRCh38
rs952223910 Chr.1: 207019613-207019613 on Build GRCh38
rs530652867 Chr.1: 207019353-207019353 on Build GRCh38
rs951405184 Chr.1: 207019349-207019349 on Build GRCh38
rs183325011 Chr.1: 207019272-207019272 on Build GRCh38
rs936873587 Chr.1: 207019539-207019539 on Build GRCh38
rs114313797 Chr.1: 207019568-207019568 on Build GRCh38
rs115382268 Chr.1: 207019321-207019321 on Build GRCh38
rs1040799763 Chr.1: 207019393-207019393 on Build GRCh38
rs928318692 Chr.1: 207019275-207019275 on Build GRCh38
rs572532278 Chr.1: 207019655-207019655 on Build GRCh38
rs374046205 Chr.1: 207019540-207019540 on Build GRCh38
rs1019693510 Chr.1: 207019716-207019716 on Build GRCh38
rs961044650 Chr.1: 207019594-207019594 on Build GRCh38
rs892976229 Chr.1: 207019478-207019478 on Build GRCh38
rs565264427 Chr.1: 207019589-207019589 on Build GRCh38
rs754482247 Chr.1: 207019584-207019584 on Build GRCh38
rs561594575 Chr.1: 207019334-207019334 on Build GRCh38
rs544351320 Chr.1: 207019453-207019453 on Build GRCh38
rs943629759 Chr.1: 207019363-207019363 on Build GRCh38
rs930790539 Chr.1: 207019338-207019338 on Build GRCh38
rs773018361 Chr.1: 207019719-207019719 on Build GRCh38
rs995361632 Chr.1: 207019667-207019667 on Build GRCh38
rs771801364 Chr.1: 207019707-207019707 on Build GRCh38
rs976027096 Chr.1: 207019488-207019488 on Build GRCh38
rs961443077 Chr.1: 207019308-207019308 on Build GRCh38
rs188584057 Chr.1: 207019688-207019688 on Build GRCh38
rs910700665 Chr.1: 207019661-207019661 on Build GRCh38
rs962639235 Chr.1: 207019273-207019273 on Build GRCh38
rs1022798029 Chr.1: 207019486-207019486 on Build GRCh38
rs1011379524 Chr.1: 207019479-207019479 on Build GRCh38
rs934754630 Chr.1: 207019438-207019438 on Build GRCh38
rs1025542908 Chr.1: 207019672-207019672 on Build GRCh38
rs986887854 Chr.1: 207019402-207019402 on Build GRCh38
rs34610389 Chr.1: 207019661-207019662 on Build GRCh38
rs1002774570 Chr.1: 207019564-207019564 on Build GRCh38
rs1052700607 Chr.1: 207019560-207019560 on Build GRCh38
rs910836635 Chr.1: 207019354-207019354 on Build GRCh38
rs901828019 Chr.1: 207019428-207019428 on Build GRCh38
rs533748401 Chr.1: 207019581-207019581 on Build GRCh38
rs1026653811 Chr.1: 207019599-207019599 on Build GRCh38
rs532997414 Chr.1: 207019551-207019551 on Build GRCh38
rs548777065 Chr.1: 207019618-207019618 on Build GRCh38
rs925445097 Chr.1: 207019507-207019507 on Build GRCh38
rs564055892 Chr.1: 207019470-207019470 on Build GRCh38
rs912668501 Chr.1: 207019439-207019439 on Build GRCh38
rs542018970 Chr.1: 207019326-207019326 on Build GRCh38
rs891445252 Chr.1: 207019562-207019562 on Build GRCh38
rs951372271 Chr.1: 207019680-207019680 on Build GRCh38
rs568430870 Chr.1: 207019685-207019685 on Build GRCh38
rs972915620 Chr.1: 207019596-207019596 on Build GRCh38
rs566801900 Chr.1: 207019563-207019563 on Build GRCh38
rs559649042 Chr.1: 207019315-207019315 on Build GRCh38
rs557076717 Chr.1: 207019718-207019718 on Build GRCh38
rs778879645 Chr.1: 207019637-207019637 on Build GRCh38

View Full Product Details