Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 207020837-207021348 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf116
Entrez Gene ID
79098
Gene Name
chromosome 1 open reading frame 116
Gene Aliases
SARG
UniGene
Hs.32417
Gene Chromosome Location
Chr.1: 207018521-207032761 on Build GRCh38
Transcripts
NM_001083924.1 NM_023938.5 XM_005273259.1 XM_006711530.1 XM_011509973.2

SNP Detail

SNP ID Chromosome Location
rs983958133 Chr.1: 207020945-207020945 on Build GRCh38
rs772567552 Chr.1: 207021291-207021291 on Build GRCh38
rs946449615 Chr.1: 207021121-207021121 on Build GRCh38
rs989385317 Chr.1: 207021239-207021239 on Build GRCh38
rs749495214 Chr.1: 207021006-207021006 on Build GRCh38
rs796597665 Chr.1: 207020868-207020869 on Build GRCh38
rs1043273379 Chr.1: 207021146-207021146 on Build GRCh38
rs74152403 Chr.1: 207021167-207021167 on Build GRCh38
rs976651310 Chr.1: 207020922-207020922 on Build GRCh38
rs895822799 Chr.1: 207021223-207021223 on Build GRCh38
rs560069877 Chr.1: 207020892-207020892 on Build GRCh38
rs185279571 Chr.1: 207021201-207021201 on Build GRCh38
rs1005579207 Chr.1: 207021260-207021260 on Build GRCh38
rs906659903 Chr.1: 207021058-207021058 on Build GRCh38
rs909699680 Chr.1: 207020959-207020959 on Build GRCh38
rs932489678 Chr.1: 207021226-207021226 on Build GRCh38
rs773588848 Chr.1: 207021319-207021319 on Build GRCh38
rs150602956 Chr.1: 207020926-207020926 on Build GRCh38
rs546814245 Chr.1: 207021117-207021117 on Build GRCh38
rs555579321 Chr.1: 207020891-207020891 on Build GRCh38
rs569182689 Chr.1: 207021277-207021277 on Build GRCh38
rs775908528 Chr.1: 207021003-207021003 on Build GRCh38
rs1025596833 Chr.1: 207021249-207021249 on Build GRCh38
rs1004040818 Chr.1: 207021061-207021061 on Build GRCh38
rs927962343 Chr.1: 207021274-207021274 on Build GRCh38
rs193175566 Chr.1: 207021085-207021085 on Build GRCh38
rs774444261 Chr.1: 207021150-207021150 on Build GRCh38
rs550777902 Chr.1: 207021052-207021052 on Build GRCh38
rs185729431 Chr.1: 207021034-207021034 on Build GRCh38
rs895331103 Chr.1: 207021070-207021070 on Build GRCh38
rs951458633 Chr.1: 207020941-207020941 on Build GRCh38
rs1042355638 Chr.1: 207021255-207021255 on Build GRCh38
rs573597994 Chr.1: 207020903-207020903 on Build GRCh38
rs960288381 Chr.1: 207020908-207020908 on Build GRCh38
rs375888657 Chr.1: 207021188-207021188 on Build GRCh38
rs939422831 Chr.1: 207021287-207021287 on Build GRCh38
rs945359366 Chr.1: 207021251-207021251 on Build GRCh38
rs1034974453 Chr.1: 207021205-207021205 on Build GRCh38
rs190244363 Chr.1: 207021042-207021042 on Build GRCh38
rs751435378 Chr.1: 207021295-207021295 on Build GRCh38
rs558002325 Chr.1: 207021321-207021321 on Build GRCh38
rs1016737942 Chr.1: 207021268-207021268 on Build GRCh38
rs763583918 Chr.1: 207021220-207021220 on Build GRCh38
rs1027886522 Chr.1: 207021111-207021111 on Build GRCh38
rs59099425 Chr.1: 207021067-207021067 on Build GRCh38
rs921097460 Chr.1: 207020925-207020925 on Build GRCh38
rs570890424 Chr.1: 207021168-207021168 on Build GRCh38
rs188877174 Chr.1: 207021312-207021312 on Build GRCh38
rs867359590 Chr.1: 207021270-207021270 on Build GRCh38
rs567210440 Chr.1: 207021241-207021241 on Build GRCh38

View Full Product Details