Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 207123783-207124330 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C4BPA
Entrez Gene ID
722
Gene Name
complement component 4 binding protein alpha
Gene Aliases
C4BP, PRP
UniGene
Hs.1012
Gene Chromosome Location
Chr.1: 207104232-207144972 on Build GRCh38
Transcripts
NM_000715.3 XM_005273251.1 XM_005273252.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
678891 c.612C>T p.D204D Chr.1: 207124272-207124272 on Build GRCh38
6060846 c.520A>T p.K174* Chr.1: 207124180-207124180 on Build GRCh38
6049129 c.521A>T p.K174M Chr.1: 207124181-207124181 on Build GRCh38
3943393 c.574T>A p.F192I Chr.1: 207124234-207124234 on Build GRCh38
1338388 c.459T>C p.R153R Chr.1: 207123952-207123952 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765449419 Chr.1: 207124245-207124245 on Build GRCh38
rs972920982 Chr.1: 207123922-207123922 on Build GRCh38
rs780844313 Chr.1: 207124273-207124273 on Build GRCh38
rs372324807 Chr.1: 207124049-207124049 on Build GRCh38
rs768517294 Chr.1: 207124307-207124307 on Build GRCh38
rs769312663 Chr.1: 207124215-207124215 on Build GRCh38
rs750663585 Chr.1: 207123878-207123878 on Build GRCh38
rs779570908 Chr.1: 207123903-207123903 on Build GRCh38
rs761018226 Chr.1: 207124039-207124039 on Build GRCh38
rs780712968 Chr.1: 207124163-207124163 on Build GRCh38
rs146506452 Chr.1: 207123944-207123944 on Build GRCh38
rs772587638 Chr.1: 207123916-207123916 on Build GRCh38
rs756875201 Chr.1: 207124160-207124160 on Build GRCh38
rs145177697 Chr.1: 207124285-207124285 on Build GRCh38
rs2842706 Chr.1: 207124121-207124121 on Build GRCh38
rs759640687 Chr.1: 207124242-207124242 on Build GRCh38
rs748958102 Chr.1: 207124064-207124064 on Build GRCh38
rs762494849 Chr.1: 207124030-207124033 on Build GRCh38
rs757593380 Chr.1: 207124272-207124272 on Build GRCh38
rs745383796 Chr.1: 207123970-207123970 on Build GRCh38
rs758806306 Chr.1: 207124144-207124144 on Build GRCh38
rs371583733 Chr.1: 207124061-207124061 on Build GRCh38
rs981846973 Chr.1: 207123906-207123906 on Build GRCh38
rs773287684 Chr.1: 207123884-207123885 on Build GRCh38
rs778351463 Chr.1: 207124146-207124146 on Build GRCh38
rs770341708 Chr.1: 207123950-207123950 on Build GRCh38
rs559134560 Chr.1: 207124020-207124020 on Build GRCh38
rs7415033 Chr.1: 207123892-207123892 on Build GRCh38
rs777547401 Chr.1: 207123942-207123942 on Build GRCh38
rs748812319 Chr.1: 207124295-207124295 on Build GRCh38
rs942147808 Chr.1: 207124110-207124110 on Build GRCh38
rs917865398 Chr.1: 207124069-207124069 on Build GRCh38
rs751546788 Chr.1: 207124303-207124303 on Build GRCh38
rs778290166 Chr.1: 207124257-207124257 on Build GRCh38
rs373183962 Chr.1: 207123881-207123881 on Build GRCh38
rs867987239 Chr.1: 207124096-207124096 on Build GRCh38
rs902350759 Chr.1: 207123812-207123812 on Build GRCh38
rs140603272 Chr.1: 207124277-207124277 on Build GRCh38
rs931567260 Chr.1: 207123969-207123969 on Build GRCh38
rs539095228 Chr.1: 207123833-207123833 on Build GRCh38
rs759030264 Chr.1: 207124248-207124248 on Build GRCh38
rs2842707 Chr.1: 207124139-207124139 on Build GRCh38
rs763923960 Chr.1: 207124026-207124026 on Build GRCh38
rs143323893 Chr.1: 207124246-207124246 on Build GRCh38
rs775434720 Chr.1: 207123988-207123988 on Build GRCh38
rs181908416 Chr.1: 207124104-207124104 on Build GRCh38
rs112042443 Chr.1: 207124143-207124143 on Build GRCh38
rs1017914033 Chr.1: 207123929-207123929 on Build GRCh38
rs375366809 Chr.1: 207124054-207124054 on Build GRCh38
rs981734004 Chr.1: 207124131-207124131 on Build GRCh38
rs750807092 Chr.1: 207123860-207123860 on Build GRCh38
rs112839397 Chr.1: 207123824-207123824 on Build GRCh38
rs776261082 Chr.1: 207123951-207123951 on Build GRCh38
rs375865150 Chr.1: 207124154-207124154 on Build GRCh38
rs957979434 Chr.1: 207123856-207123856 on Build GRCh38
rs745339467 Chr.1: 207124185-207124185 on Build GRCh38
rs889373111 Chr.1: 207124202-207124202 on Build GRCh38
rs367547901 Chr.1: 207123911-207123911 on Build GRCh38
rs111846206 Chr.1: 207124008-207124008 on Build GRCh38
rs766587946 Chr.1: 207124240-207124240 on Build GRCh38
rs773389282 Chr.1: 207124036-207124036 on Build GRCh38
rs369304888 Chr.1: 207124066-207124066 on Build GRCh38
rs903492938 Chr.1: 207124263-207124263 on Build GRCh38
rs1032708497 Chr.1: 207123838-207123838 on Build GRCh38
rs372772893 Chr.1: 207123972-207123972 on Build GRCh38
rs368827962 Chr.1: 207124221-207124221 on Build GRCh38
rs376071191 Chr.1: 207124011-207124011 on Build GRCh38
rs773977537 Chr.1: 207124145-207124148 on Build GRCh38
rs369505952 Chr.1: 207124138-207124138 on Build GRCh38
rs780249584 Chr.1: 207123880-207123880 on Build GRCh38
rs967830992 Chr.1: 207123943-207123943 on Build GRCh38
rs755458581 Chr.1: 207123901-207123901 on Build GRCh38
rs774141891 Chr.1: 207124223-207124223 on Build GRCh38
rs770551381 Chr.1: 207124056-207124057 on Build GRCh38
rs1052046836 Chr.1: 207123900-207123900 on Build GRCh38
rs778440020 Chr.1: 207124082-207124082 on Build GRCh38
rs200253338 Chr.1: 207124239-207124239 on Build GRCh38
rs751883518 Chr.1: 207124265-207124265 on Build GRCh38
rs201003117 Chr.1: 207124276-207124276 on Build GRCh38
rs997677116 Chr.1: 207123884-207123884 on Build GRCh38
rs756122597 Chr.1: 207123879-207123879 on Build GRCh38
rs749261139 Chr.1: 207124222-207124222 on Build GRCh38

View Full Product Details