Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 207325618-207326134 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD55
Entrez Gene ID
1604
Gene Name
CD55 molecule (Cromer blood group)
Gene Aliases
CR, CROM, DAF, TC
UniGene
Hs.126517
Gene Chromosome Location
Chr.1: 207321472-207360966 on Build GRCh38
Transcripts
NM_000574.4 NM_001114752.2 NM_001300902.1 NM_001300903.1 NM_001300904.1 NR_125349.1 XM_017000467.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5989681 c.503A>G p.E168G Chr.1: 207325646-207325646 on Build GRCh38
6296824 c.526G>T p.V176L Chr.1: 207325669-207325669 on Build GRCh38
5989205 c.578+1G>T p.? Chr.1: 207325722-207325722 on Build GRCh38
5441030 c.497C>T p.P166L Chr.1: 207325640-207325640 on Build GRCh38
5227932 c.509G>A p.R170Q Chr.1: 207325652-207325652 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs998162792 Chr.1: 207325806-207325806 on Build GRCh38
rs771663673 Chr.1: 207325654-207325654 on Build GRCh38
rs968846289 Chr.1: 207325855-207325855 on Build GRCh38
rs553510719 Chr.1: 207325969-207325969 on Build GRCh38
rs10623899 Chr.1: 207326021-207326022 on Build GRCh38
rs898350735 Chr.1: 207325821-207325821 on Build GRCh38
rs1010991715 Chr.1: 207325841-207325841 on Build GRCh38
rs781529960 Chr.1: 207325734-207325734 on Build GRCh38
rs769110255 Chr.1: 207325745-207325745 on Build GRCh38
rs869160399 Chr.1: 207326022-207326023 on Build GRCh38
rs761966810 Chr.1: 207325729-207325729 on Build GRCh38
rs1008332322 Chr.1: 207326038-207326038 on Build GRCh38
rs571289812 Chr.1: 207325928-207325928 on Build GRCh38
rs992528214 Chr.1: 207326031-207326031 on Build GRCh38
rs760968457 Chr.1: 207325653-207325660 on Build GRCh38
rs752481959 Chr.1: 207325652-207325652 on Build GRCh38
rs915871245 Chr.1: 207325703-207325703 on Build GRCh38
rs774630833 Chr.1: 207325648-207325648 on Build GRCh38
rs201162563 Chr.1: 207326022-207326022 on Build GRCh38
rs151294032 Chr.1: 207325656-207325656 on Build GRCh38
rs954327792 Chr.1: 207325834-207325834 on Build GRCh38
rs779417348 Chr.1: 207325754-207325754 on Build GRCh38
rs769163287 Chr.1: 207325645-207325645 on Build GRCh38
rs767688671 Chr.1: 207325732-207325732 on Build GRCh38
rs568719123 Chr.1: 207325785-207325785 on Build GRCh38
rs915796138 Chr.1: 207326053-207326053 on Build GRCh38
rs754408957 Chr.1: 207325955-207325955 on Build GRCh38
rs551157871 Chr.1: 207325926-207325926 on Build GRCh38
rs750721259 Chr.1: 207325735-207325735 on Build GRCh38
rs28371661 Chr.1: 207326032-207326032 on Build GRCh38
rs368435993 Chr.1: 207326076-207326076 on Build GRCh38
rs977187996 Chr.1: 207325867-207325867 on Build GRCh38
rs374764368 Chr.1: 207325837-207325837 on Build GRCh38
rs548922623 Chr.1: 207325862-207325862 on Build GRCh38
rs995185018 Chr.1: 207325868-207325868 on Build GRCh38
rs938357623 Chr.1: 207325668-207325668 on Build GRCh38
rs1022481303 Chr.1: 207325844-207325844 on Build GRCh38
rs772957291 Chr.1: 207325655-207325655 on Build GRCh38
rs145025194 Chr.1: 207326018-207326019 on Build GRCh38
rs569005343 Chr.1: 207325891-207325891 on Build GRCh38
rs1019181321 Chr.1: 207326048-207326048 on Build GRCh38
rs368905867 Chr.1: 207325716-207325716 on Build GRCh38
rs948620257 Chr.1: 207326075-207326075 on Build GRCh38
rs753303079 Chr.1: 207325673-207325673 on Build GRCh38
rs762195469 Chr.1: 207325651-207325651 on Build GRCh38
rs749330187 Chr.1: 207325742-207325742 on Build GRCh38
rs79108869 Chr.1: 207326020-207326020 on Build GRCh38
rs756237389 Chr.1: 207325919-207325919 on Build GRCh38
rs1057053700 Chr.1: 207325696-207325696 on Build GRCh38
rs537602579 Chr.1: 207325826-207325826 on Build GRCh38
rs1036794642 Chr.1: 207325792-207325792 on Build GRCh38
rs749862284 Chr.1: 207325641-207325641 on Build GRCh38
rs948764533 Chr.1: 207325720-207325720 on Build GRCh38
rs765300495 Chr.1: 207325717-207325717 on Build GRCh38
rs10634414 Chr.1: 207326020-207326021 on Build GRCh38
rs756131028 Chr.1: 207325738-207325738 on Build GRCh38
rs915878687 Chr.1: 207325719-207325719 on Build GRCh38
rs764441759 Chr.1: 207325704-207325706 on Build GRCh38
rs59700450 Chr.1: 207326107-207326107 on Build GRCh38
rs180879554 Chr.1: 207325741-207325741 on Build GRCh38
rs184467616 Chr.1: 207326059-207326059 on Build GRCh38
rs146276485 Chr.1: 207325659-207325659 on Build GRCh38
rs28371660 Chr.1: 207325930-207325930 on Build GRCh38

View Full Product Details