Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 207339087-207339627 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD55
Entrez Gene ID
1604
Gene Name
CD55 molecule (Cromer blood group)
Gene Aliases
CR, CROM, DAF, TC
UniGene
Hs.126517
Gene Chromosome Location
Chr.1: 207321472-207360966 on Build GRCh38
Transcripts
NM_000574.4 NM_001114752.2 NM_001300902.1 NM_001300903.1 NM_001300904.1 NR_125349.1 XM_017000467.1

SNP Detail

SNP ID Chromosome Location
rs758644911 Chr.1: 207339403-207339403 on Build GRCh38
rs897982946 Chr.1: 207339430-207339430 on Build GRCh38
rs746324299 Chr.1: 207339418-207339418 on Build GRCh38
rs1007481120 Chr.1: 207339536-207339536 on Build GRCh38
rs186602607 Chr.1: 207339397-207339397 on Build GRCh38
rs924721150 Chr.1: 207339336-207339336 on Build GRCh38
rs762522400 Chr.1: 207339358-207339358 on Build GRCh38
rs35209555 Chr.1: 207339233-207339234 on Build GRCh38
rs546371980 Chr.1: 207339413-207339413 on Build GRCh38
rs148720717 Chr.1: 207339406-207339406 on Build GRCh38
rs989953729 Chr.1: 207339345-207339345 on Build GRCh38
rs761364842 Chr.1: 207339372-207339372 on Build GRCh38
rs879373916 Chr.1: 207339225-207339225 on Build GRCh38
rs761225241 Chr.1: 207339457-207339457 on Build GRCh38
rs775796455 Chr.1: 207339435-207339435 on Build GRCh38
rs750443090 Chr.1: 207339388-207339388 on Build GRCh38
rs530724306 Chr.1: 207339205-207339205 on Build GRCh38
rs953291023 Chr.1: 207339275-207339275 on Build GRCh38
rs968621175 Chr.1: 207339300-207339300 on Build GRCh38
rs753716038 Chr.1: 207339402-207339402 on Build GRCh38
rs144979738 Chr.1: 207339147-207339147 on Build GRCh38
rs751025150 Chr.1: 207339371-207339371 on Build GRCh38
rs377141848 Chr.1: 207339353-207339353 on Build GRCh38
rs376649492 Chr.1: 207339333-207339333 on Build GRCh38
rs879294236 Chr.1: 207339231-207339232 on Build GRCh38
rs915638234 Chr.1: 207339384-207339384 on Build GRCh38
rs768277801 Chr.1: 207339449-207339449 on Build GRCh38
rs977299319 Chr.1: 207339591-207339591 on Build GRCh38
rs200715817 Chr.1: 207339465-207339465 on Build GRCh38
rs981098394 Chr.1: 207339391-207339391 on Build GRCh38
rs528581736 Chr.1: 207339112-207339112 on Build GRCh38
rs770202881 Chr.1: 207339427-207339427 on Build GRCh38
rs548631858 Chr.1: 207339130-207339130 on Build GRCh38
rs59976929 Chr.1: 207339236-207339237 on Build GRCh38
rs775841683 Chr.1: 207339357-207339357 on Build GRCh38
rs147314732 Chr.1: 207339234-207339235 on Build GRCh38
rs778188153 Chr.1: 207339405-207339405 on Build GRCh38
rs1036527561 Chr.1: 207339116-207339116 on Build GRCh38
rs374419274 Chr.1: 207339492-207339492 on Build GRCh38
rs28371628 Chr.1: 207339389-207339389 on Build GRCh38
rs763770853 Chr.1: 207339365-207339365 on Build GRCh38
rs868355500 Chr.1: 207339404-207339404 on Build GRCh38
rs371872965 Chr.1: 207339500-207339500 on Build GRCh38
rs746475041 Chr.1: 207339294-207339294 on Build GRCh38
rs1031445941 Chr.1: 207339269-207339269 on Build GRCh38
rs777443201 Chr.1: 207339210-207339210 on Build GRCh38
rs766271165 Chr.1: 207339274-207339274 on Build GRCh38
rs555086419 Chr.1: 207339563-207339563 on Build GRCh38
rs930770464 Chr.1: 207339444-207339444 on Build GRCh38
rs72501056 Chr.1: 207339235-207339236 on Build GRCh38
rs1021577982 Chr.1: 207339557-207339557 on Build GRCh38
rs994905240 Chr.1: 207339200-207339200 on Build GRCh38
rs758062911 Chr.1: 207339114-207339114 on Build GRCh38
rs774034891 Chr.1: 207339451-207339451 on Build GRCh38
rs771106629 Chr.1: 207339412-207339412 on Build GRCh38
rs977874707 Chr.1: 207339326-207339326 on Build GRCh38
rs776934951 Chr.1: 207339375-207339375 on Build GRCh38
rs957227933 Chr.1: 207339325-207339325 on Build GRCh38
rs181529094 Chr.1: 207339305-207339305 on Build GRCh38
rs986076006 Chr.1: 207339281-207339281 on Build GRCh38
rs1030232884 Chr.1: 207339169-207339169 on Build GRCh38
rs570039992 Chr.1: 207339493-207339495 on Build GRCh38
rs10641442 Chr.1: 207339232-207339233 on Build GRCh38
rs977347825 Chr.1: 207339237-207339237 on Build GRCh38
rs888808499 Chr.1: 207339523-207339523 on Build GRCh38
rs767328613 Chr.1: 207339385-207339385 on Build GRCh38
rs938661388 Chr.1: 207339346-207339346 on Build GRCh38
rs201307804 Chr.1: 207339445-207339445 on Build GRCh38
rs866503489 Chr.1: 207339520-207339520 on Build GRCh38
rs375092297 Chr.1: 207339232-207339232 on Build GRCh38
rs897594314 Chr.1: 207339433-207339433 on Build GRCh38
rs869302881 Chr.1: 207339237-207339238 on Build GRCh38
rs968916167 Chr.1: 207339566-207339566 on Build GRCh38

View Full Product Details