Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 207495943-207496465 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CR1
Entrez Gene ID
1378
Gene Name
complement component 3b/4b receptor 1 (Knops blood group)
Gene Aliases
C3BR, C4BR, CD35, KN
UniGene
Hs.334019
Gene Chromosome Location
Chr.1: 207496128-207641765 on Build GRCh38
Transcripts
NM_000573.3 NM_000651.4 XM_006711166.2 XM_011509205.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3864326 c.59C>T p.P20L Chr.1: 207496326-207496326 on Build GRCh38
5969113 c.31C>G p.P11A Chr.1: 207496298-207496298 on Build GRCh38
3864324 c.32C>T p.P11L Chr.1: 207496299-207496299 on Build GRCh38
6455587 c.53G>A p.G18D Chr.1: 207496320-207496320 on Build GRCh38
4027610 c.45G>A p.P15P Chr.1: 207496312-207496312 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs61821138 Chr.1: 207495998-207495998 on Build GRCh38
rs772522277 Chr.1: 207496395-207496395 on Build GRCh38
rs370940682 Chr.1: 207496367-207496367 on Build GRCh38
rs568882336 Chr.1: 207496109-207496109 on Build GRCh38
rs752538669 Chr.1: 207496405-207496405 on Build GRCh38
rs913295721 Chr.1: 207496106-207496106 on Build GRCh38
rs1038479607 Chr.1: 207496286-207496286 on Build GRCh38
rs754966774 Chr.1: 207496419-207496419 on Build GRCh38
rs71635143 Chr.1: 207495991-207495991 on Build GRCh38
rs917092678 Chr.1: 207496178-207496178 on Build GRCh38
rs753785825 Chr.1: 207496245-207496245 on Build GRCh38
rs768859252 Chr.1: 207496434-207496434 on Build GRCh38
rs368742318 Chr.1: 207496182-207496182 on Build GRCh38
rs779232378 Chr.1: 207496372-207496372 on Build GRCh38
rs564189415 Chr.1: 207495994-207495994 on Build GRCh38
rs772751212 Chr.1: 207496425-207496425 on Build GRCh38
rs377520654 Chr.1: 207496429-207496429 on Build GRCh38
rs774774612 Chr.1: 207496314-207496314 on Build GRCh38
rs780076848 Chr.1: 207496414-207496414 on Build GRCh38
rs975016241 Chr.1: 207496081-207496081 on Build GRCh38
rs1005290184 Chr.1: 207496267-207496267 on Build GRCh38
rs778265753 Chr.1: 207496428-207496428 on Build GRCh38
rs769828093 Chr.1: 207496308-207496308 on Build GRCh38
rs780093897 Chr.1: 207496366-207496366 on Build GRCh38
rs776088016 Chr.1: 207496438-207496438 on Build GRCh38
rs755629620 Chr.1: 207496239-207496239 on Build GRCh38
rs764815300 Chr.1: 207496403-207496403 on Build GRCh38
rs770765679 Chr.1: 207496399-207496399 on Build GRCh38
rs766008795 Chr.1: 207496241-207496241 on Build GRCh38
rs996388913 Chr.1: 207496303-207496303 on Build GRCh38
rs776199892 Chr.1: 207496400-207496400 on Build GRCh38
rs375945701 Chr.1: 207496312-207496312 on Build GRCh38
rs760378534 Chr.1: 207496398-207496398 on Build GRCh38
rs372551774 Chr.1: 207496301-207496301 on Build GRCh38
rs372830780 Chr.1: 207495990-207495990 on Build GRCh38
rs1041286483 Chr.1: 207496407-207496407 on Build GRCh38
rs778743380 Chr.1: 207496259-207496259 on Build GRCh38
rs2296878 Chr.1: 207496357-207496357 on Build GRCh38
rs374273624 Chr.1: 207496397-207496397 on Build GRCh38
rs758902493 Chr.1: 207496401-207496401 on Build GRCh38
rs772948093 Chr.1: 207496316-207496316 on Build GRCh38
rs1016583160 Chr.1: 207496075-207496075 on Build GRCh38
rs529045474 Chr.1: 207496011-207496011 on Build GRCh38
rs555401223 Chr.1: 207496352-207496352 on Build GRCh38
rs753333659 Chr.1: 207496324-207496324 on Build GRCh38
rs9429782 Chr.1: 207496010-207496010 on Build GRCh38
rs2296879 Chr.1: 207496369-207496369 on Build GRCh38
rs754912397 Chr.1: 207496258-207496258 on Build GRCh38
rs879097166 Chr.1: 207496114-207496115 on Build GRCh38
rs575251255 Chr.1: 207496410-207496410 on Build GRCh38
rs199932175 Chr.1: 207496227-207496227 on Build GRCh38
rs949833518 Chr.1: 207496225-207496225 on Build GRCh38
rs779078873 Chr.1: 207496423-207496423 on Build GRCh38
rs182300322 Chr.1: 207496127-207496127 on Build GRCh38
rs963997425 Chr.1: 207496078-207496078 on Build GRCh38
rs1005608527 Chr.1: 207496052-207496052 on Build GRCh38
rs1055679052 Chr.1: 207496172-207496172 on Build GRCh38
rs775590306 Chr.1: 207496311-207496311 on Build GRCh38
rs113160165 Chr.1: 207496415-207496415 on Build GRCh38
rs1029157595 Chr.1: 207496092-207496092 on Build GRCh38
rs373914260 Chr.1: 207496315-207496315 on Build GRCh38
rs35561698 Chr.1: 207496081-207496082 on Build GRCh38
rs757493323 Chr.1: 207496261-207496261 on Build GRCh38
rs765951976 Chr.1: 207496318-207496318 on Build GRCh38
rs149416512 Chr.1: 207496125-207496125 on Build GRCh38
rs765154374 Chr.1: 207496327-207496327 on Build GRCh38
rs748713759 Chr.1: 207496393-207496393 on Build GRCh38
rs987524422 Chr.1: 207496104-207496104 on Build GRCh38
rs199732885 Chr.1: 207496326-207496326 on Build GRCh38
rs575513177 Chr.1: 207496322-207496322 on Build GRCh38
rs748631518 Chr.1: 207496424-207496424 on Build GRCh38
rs369804986 Chr.1: 207496313-207496313 on Build GRCh38
rs138893073 Chr.1: 207496098-207496098 on Build GRCh38
rs576791246 Chr.1: 207496409-207496409 on Build GRCh38
rs762920284 Chr.1: 207496408-207496408 on Build GRCh38
rs777578334 Chr.1: 207496356-207496356 on Build GRCh38
rs925947189 Chr.1: 207496364-207496364 on Build GRCh38
rs756329514 Chr.1: 207496413-207496413 on Build GRCh38
rs531302285 Chr.1: 207496122-207496122 on Build GRCh38
rs781322624 Chr.1: 207496298-207496298 on Build GRCh38
rs566717082 Chr.1: 207496317-207496317 on Build GRCh38
rs756378518 Chr.1: 207496361-207496361 on Build GRCh38
rs577901202 Chr.1: 207496437-207496437 on Build GRCh38
rs752683886 Chr.1: 207496350-207496350 on Build GRCh38
rs533172421 Chr.1: 207495997-207495997 on Build GRCh38
rs954838156 Chr.1: 207496096-207496096 on Build GRCh38

View Full Product Details