Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 207766922-207767423 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD46
Entrez Gene ID
4179
Gene Name
CD46 molecule
Gene Aliases
AHUS2, MCP, MIC10, TLX, TRA2.10
UniGene
Hs.510402
Gene Chromosome Location
Chr.1: 207752038-207795516 on Build GRCh38
Transcripts
NM_002389.4 NM_153826.3 NM_172350.2 NM_172351.2 NM_172352.2 NM_172353.2 NM_172359.2 NM_172361.2 XM_011509563.1 XM_011509564.1 XM_017001308.1 XM_017001309.1 XM_017001310.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5611892 c.692C>A p.P231Q Chr.1: 207767031-207767031 on Build GRCh38
3671612 c.736T>A p.F246I Chr.1: 207767075-207767075 on Build GRCh38
5379497 c.832C>T p.P278S Chr.1: 207767171-207767171 on Build GRCh38
5611893 c.692C>A p.P231Q Chr.1: 207767031-207767031 on Build GRCh38
5641424 c.700G>A p.E234K Chr.1: 207767039-207767039 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs532074191 Chr.1: 207767287-207767288 on Build GRCh38
rs530080403 Chr.1: 207767331-207767331 on Build GRCh38
rs755419614 Chr.1: 207767138-207767138 on Build GRCh38
rs778743202 Chr.1: 207767229-207767231 on Build GRCh38
rs751844358 Chr.1: 207767107-207767107 on Build GRCh38
rs764916823 Chr.1: 207767098-207767098 on Build GRCh38
rs772672525 Chr.1: 207767083-207767083 on Build GRCh38
rs1020883880 Chr.1: 207767351-207767351 on Build GRCh38
rs41317817 Chr.1: 207767337-207767337 on Build GRCh38
rs759430396 Chr.1: 207767097-207767097 on Build GRCh38
rs750965296 Chr.1: 207767355-207767355 on Build GRCh38
rs1049722180 Chr.1: 207767315-207767315 on Build GRCh38
rs992780212 Chr.1: 207766980-207766980 on Build GRCh38
rs760633093 Chr.1: 207767084-207767084 on Build GRCh38
rs745757525 Chr.1: 207767207-207767207 on Build GRCh38
rs757165980 Chr.1: 207767223-207767223 on Build GRCh38
rs1048263862 Chr.1: 207767074-207767074 on Build GRCh38
rs1022461055 Chr.1: 207767346-207767346 on Build GRCh38
rs1034013756 Chr.1: 207767335-207767335 on Build GRCh38
rs776597190 Chr.1: 207767203-207767203 on Build GRCh38
rs183517108 Chr.1: 207767215-207767215 on Build GRCh38
rs779117047 Chr.1: 207767141-207767141 on Build GRCh38
rs769658537 Chr.1: 207767213-207767213 on Build GRCh38
rs747869014 Chr.1: 207767154-207767154 on Build GRCh38
rs749865187 Chr.1: 207767232-207767234 on Build GRCh38
rs909675840 Chr.1: 207767076-207767076 on Build GRCh38
rs767779192 Chr.1: 207767228-207767228 on Build GRCh38
rs746453295 Chr.1: 207767177-207767177 on Build GRCh38
rs200389060 Chr.1: 207767189-207767189 on Build GRCh38
rs576595023 Chr.1: 207767039-207767039 on Build GRCh38
rs556795257 Chr.1: 207767038-207767038 on Build GRCh38
rs763178656 Chr.1: 207766983-207766983 on Build GRCh38
rs781097294 Chr.1: 207767035-207767035 on Build GRCh38
rs754385690 Chr.1: 207767137-207767137 on Build GRCh38
rs776418162 Chr.1: 207767091-207767091 on Build GRCh38
rs41317819 Chr.1: 207767344-207767344 on Build GRCh38
rs1002049544 Chr.1: 207767317-207767317 on Build GRCh38
rs1037394257 Chr.1: 207767160-207767160 on Build GRCh38
rs750470907 Chr.1: 207767134-207767134 on Build GRCh38
rs121909589 Chr.1: 207767057-207767057 on Build GRCh38
rs766197874 Chr.1: 207766966-207766966 on Build GRCh38
rs757403112 Chr.1: 207767026-207767026 on Build GRCh38
rs374407190 Chr.1: 207767360-207767360 on Build GRCh38
rs780577622 Chr.1: 207767044-207767044 on Build GRCh38
rs913971995 Chr.1: 207766948-207766948 on Build GRCh38
rs201380032 Chr.1: 207767025-207767025 on Build GRCh38
rs17006830 Chr.1: 207767135-207767135 on Build GRCh38
rs115094188 Chr.1: 207767085-207767085 on Build GRCh38
rs374762173 Chr.1: 207767125-207767125 on Build GRCh38
rs761897540 Chr.1: 207767123-207767123 on Build GRCh38
rs141420882 Chr.1: 207767362-207767362 on Build GRCh38
rs749597074 Chr.1: 207767046-207767046 on Build GRCh38
rs768898926 Chr.1: 207767053-207767053 on Build GRCh38
rs966701434 Chr.1: 207767359-207767359 on Build GRCh38
rs932805079 Chr.1: 207767389-207767390 on Build GRCh38
rs187984918 Chr.1: 207767361-207767361 on Build GRCh38
rs557590325 Chr.1: 207767054-207767054 on Build GRCh38
rs1024743006 Chr.1: 207767243-207767243 on Build GRCh38
rs760897738 Chr.1: 207767245-207767245 on Build GRCh38
rs201331374 Chr.1: 207767173-207767173 on Build GRCh38
rs780774047 Chr.1: 207767136-207767136 on Build GRCh38
rs370525888 Chr.1: 207766976-207766976 on Build GRCh38
rs563587992 Chr.1: 207767343-207767343 on Build GRCh38
rs773563367 Chr.1: 207767234-207767234 on Build GRCh38
rs773310797 Chr.1: 207767285-207767286 on Build GRCh38
rs886039868 Chr.1: 207767115-207767115 on Build GRCh38
rs372366355 Chr.1: 207767246-207767246 on Build GRCh38
rs143544996 Chr.1: 207767126-207767126 on Build GRCh38
rs992914728 Chr.1: 207767371-207767371 on Build GRCh38

View Full Product Details