Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 207910924-207911426 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD34
Entrez Gene ID
947
Gene Name
CD34 molecule
Gene Aliases
-
UniGene
Hs.374990
Gene Chromosome Location
Chr.1: 207886538-207911338 on Build GRCh38
Transcripts
NM_001025109.1 NM_001773.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6416416 c.40C>T p.P14S Chr.1: 207911041-207911041 on Build GRCh38
678832 c.18C>T p.G6G Chr.1: 207911063-207911063 on Build GRCh38
6009770 c.50G>A p.W17* Chr.1: 207911031-207911031 on Build GRCh38
4670474 c.20C>T p.A7V Chr.1: 207911061-207911061 on Build GRCh38
3689330 c.76C>T p.L26L Chr.1: 207911005-207911005 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775324439 Chr.1: 207911071-207911071 on Build GRCh38
rs538364094 Chr.1: 207911140-207911140 on Build GRCh38
rs769791456 Chr.1: 207911127-207911127 on Build GRCh38
rs538918329 Chr.1: 207911062-207911062 on Build GRCh38
rs769591700 Chr.1: 207911070-207911070 on Build GRCh38
rs932065541 Chr.1: 207911253-207911253 on Build GRCh38
rs761967848 Chr.1: 207910989-207910989 on Build GRCh38
rs766848253 Chr.1: 207911010-207911010 on Build GRCh38
rs1005564366 Chr.1: 207911295-207911295 on Build GRCh38
rs779936577 Chr.1: 207911129-207911129 on Build GRCh38
rs988703880 Chr.1: 207911119-207911119 on Build GRCh38
rs377589205 Chr.1: 207911027-207911027 on Build GRCh38
rs941027629 Chr.1: 207911164-207911164 on Build GRCh38
rs956827725 Chr.1: 207911038-207911038 on Build GRCh38
rs969831165 Chr.1: 207911363-207911363 on Build GRCh38
rs758179289 Chr.1: 207911029-207911029 on Build GRCh38
rs929739723 Chr.1: 207911211-207911211 on Build GRCh38
rs756747544 Chr.1: 207911110-207911110 on Build GRCh38
rs781190944 Chr.1: 207911008-207911008 on Build GRCh38
rs904138256 Chr.1: 207911366-207911366 on Build GRCh38
rs765792046 Chr.1: 207911254-207911254 on Build GRCh38
rs780684933 Chr.1: 207911114-207911114 on Build GRCh38
rs370283469 Chr.1: 207911079-207911079 on Build GRCh38
rs751175416 Chr.1: 207911107-207911107 on Build GRCh38
rs760695254 Chr.1: 207911085-207911085 on Build GRCh38
rs1007515102 Chr.1: 207911278-207911278 on Build GRCh38
rs1040338284 Chr.1: 207911184-207911184 on Build GRCh38
rs767715110 Chr.1: 207910991-207910991 on Build GRCh38
rs199706543 Chr.1: 207911005-207911005 on Build GRCh38
rs765823561 Chr.1: 207911099-207911099 on Build GRCh38
rs557658156 Chr.1: 207911100-207911100 on Build GRCh38
rs776253136 Chr.1: 207911370-207911370 on Build GRCh38
rs779357137 Chr.1: 207911026-207911026 on Build GRCh38
rs746540053 Chr.1: 207911034-207911034 on Build GRCh38
rs1023977180 Chr.1: 207911352-207911352 on Build GRCh38
rs781011652 Chr.1: 207911058-207911058 on Build GRCh38
rs867164795 Chr.1: 207911041-207911041 on Build GRCh38
rs55864664 Chr.1: 207911167-207911167 on Build GRCh38
rs770836393 Chr.1: 207910957-207910957 on Build GRCh38
rs774585979 Chr.1: 207910986-207910986 on Build GRCh38
rs770798860 Chr.1: 207910999-207911000 on Build GRCh38
rs865977773 Chr.1: 207911367-207911367 on Build GRCh38
rs766444945 Chr.1: 207911092-207911092 on Build GRCh38
rs773464652 Chr.1: 207911082-207911082 on Build GRCh38
rs771789925 Chr.1: 207910958-207910958 on Build GRCh38
rs1015148741 Chr.1: 207911313-207911313 on Build GRCh38
rs1024091453 Chr.1: 207911045-207911045 on Build GRCh38
rs768456579 Chr.1: 207911321-207911321 on Build GRCh38
rs926916557 Chr.1: 207911161-207911161 on Build GRCh38
rs760059829 Chr.1: 207911096-207911096 on Build GRCh38
rs745952179 Chr.1: 207911019-207911019 on Build GRCh38
rs758321887 Chr.1: 207911326-207911329 on Build GRCh38
rs575913414 Chr.1: 207911158-207911158 on Build GRCh38
rs776600352 Chr.1: 207910966-207910966 on Build GRCh38
rs759587686 Chr.1: 207911055-207911056 on Build GRCh38
rs745370982 Chr.1: 207911120-207911120 on Build GRCh38
rs900618301 Chr.1: 207911237-207911237 on Build GRCh38
rs979453471 Chr.1: 207911018-207911018 on Build GRCh38
rs879155336 Chr.1: 207910976-207910976 on Build GRCh38
rs770452625 Chr.1: 207911035-207911035 on Build GRCh38
rs777338501 Chr.1: 207911106-207911106 on Build GRCh38
rs555432955 Chr.1: 207911275-207911275 on Build GRCh38
rs948591775 Chr.1: 207910959-207910977 on Build GRCh38
rs553287118 Chr.1: 207910964-207910964 on Build GRCh38
rs896888813 Chr.1: 207911190-207911190 on Build GRCh38
rs956776331 Chr.1: 207911374-207911374 on Build GRCh38
rs1018537256 Chr.1: 207911348-207911348 on Build GRCh38
rs758800498 Chr.1: 207911102-207911102 on Build GRCh38
rs1018669223 Chr.1: 207910980-207910980 on Build GRCh38
rs774259722 Chr.1: 207911004-207911004 on Build GRCh38
rs774035547 Chr.1: 207911336-207911336 on Build GRCh38
rs1030986603 Chr.1: 207911372-207911372 on Build GRCh38
rs1007156233 Chr.1: 207910975-207910975 on Build GRCh38
rs569400899 Chr.1: 207911097-207911097 on Build GRCh38
rs764961117 Chr.1: 207910979-207910979 on Build GRCh38
rs754325569 Chr.1: 207911016-207911016 on Build GRCh38
rs949982242 Chr.1: 207911112-207911112 on Build GRCh38
rs754309424 Chr.1: 207911093-207911093 on Build GRCh38
rs969778816 Chr.1: 207911078-207911078 on Build GRCh38
rs753411742 Chr.1: 207911137-207911137 on Build GRCh38
rs762732150 Chr.1: 207911075-207911075 on Build GRCh38
rs537224462 Chr.1: 207910974-207910974 on Build GRCh38
rs12135011 Chr.1: 207911222-207911222 on Build GRCh38

View Full Product Details