Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 209675214-209675704 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
G0S2
Entrez Gene ID
50486
Gene Name
G0/G1 switch 2
Gene Aliases
-
UniGene
Hs.432132
Gene Chromosome Location
Chr.1: 209675325-209676390 on Build GRCh38
Transcripts
NM_015714.3 NR_134509.1 NR_134510.1 NR_134511.1

Gene Symbol
LOC101930114
Entrez Gene ID
101930114
Gene Name
uncharacterized LOC101930114
Gene Aliases
-
UniGene
Hs.663934
Gene Chromosome Location
Chr.1: 209661359-209742562 on Build GRCh38
Transcripts
NM_015714.3 NR_134509.1 NR_134510.1 NR_134511.1

SNP Detail

SNP ID Chromosome Location
rs988301174 Chr.1: 209675551-209675551 on Build GRCh38
rs1026309743 Chr.1: 209675616-209675616 on Build GRCh38
rs919494227 Chr.1: 209675386-209675386 on Build GRCh38
rs753948383 Chr.1: 209675390-209675390 on Build GRCh38
rs772800495 Chr.1: 209675676-209675676 on Build GRCh38
rs546567604 Chr.1: 209675370-209675370 on Build GRCh38
rs562119503 Chr.1: 209675637-209675637 on Build GRCh38
rs902422703 Chr.1: 209675487-209675487 on Build GRCh38
rs373701184 Chr.1: 209675311-209675311 on Build GRCh38
rs1002667384 Chr.1: 209675326-209675326 on Build GRCh38
rs902455186 Chr.1: 209675283-209675283 on Build GRCh38
rs914035849 Chr.1: 209675554-209675554 on Build GRCh38
rs79763330 Chr.1: 209675240-209675240 on Build GRCh38
rs1035163665 Chr.1: 209675648-209675648 on Build GRCh38
rs144655136 Chr.1: 209675488-209675488 on Build GRCh38
rs116289160 Chr.1: 209675626-209675626 on Build GRCh38
rs533830875 Chr.1: 209675305-209675305 on Build GRCh38
rs892705077 Chr.1: 209675243-209675243 on Build GRCh38
rs1002313207 Chr.1: 209675450-209675450 on Build GRCh38
rs1815548 Chr.1: 209675255-209675255 on Build GRCh38
rs964495183 Chr.1: 209675397-209675397 on Build GRCh38
rs932375 Chr.1: 209675351-209675351 on Build GRCh38
rs1038164712 Chr.1: 209675472-209675472 on Build GRCh38
rs771564270 Chr.1: 209675667-209675667 on Build GRCh38
rs747878677 Chr.1: 209675659-209675659 on Build GRCh38
rs1016102876 Chr.1: 209675337-209675337 on Build GRCh38
rs3817870 Chr.1: 209675657-209675657 on Build GRCh38
rs925715744 Chr.1: 209675534-209675534 on Build GRCh38
rs954593706 Chr.1: 209675367-209675367 on Build GRCh38
rs905165165 Chr.1: 209675439-209675439 on Build GRCh38
rs978436975 Chr.1: 209675533-209675533 on Build GRCh38
rs982240906 Chr.1: 209675391-209675391 on Build GRCh38
rs954645941 Chr.1: 209675498-209675498 on Build GRCh38
rs532821821 Chr.1: 209675352-209675352 on Build GRCh38
rs77090413 Chr.1: 209675552-209675552 on Build GRCh38
rs955810957 Chr.1: 209675539-209675539 on Build GRCh38
rs1052595709 Chr.1: 209675412-209675412 on Build GRCh38
rs749782145 Chr.1: 209675654-209675654 on Build GRCh38
rs1029944876 Chr.1: 209675328-209675328 on Build GRCh38
rs949642013 Chr.1: 209675565-209675565 on Build GRCh38
rs1042344862 Chr.1: 209675280-209675280 on Build GRCh38
rs866292272 Chr.1: 209675315-209675315 on Build GRCh38
rs190583758 Chr.1: 209675570-209675570 on Build GRCh38
rs1046377150 Chr.1: 209675262-209675262 on Build GRCh38
rs1046609514 Chr.1: 209675574-209675574 on Build GRCh38
rs1007614218 Chr.1: 209675520-209675520 on Build GRCh38
rs535547227 Chr.1: 209675490-209675490 on Build GRCh38
rs112753859 Chr.1: 209675604-209675604 on Build GRCh38
rs11542711 Chr.1: 209675508-209675508 on Build GRCh38
rs1008779613 Chr.1: 209675502-209675502 on Build GRCh38
rs546499183 Chr.1: 209675517-209675517 on Build GRCh38
rs769193505 Chr.1: 209675655-209675655 on Build GRCh38
rs763214260 Chr.1: 209675287-209675287 on Build GRCh38
rs905318926 Chr.1: 209675270-209675270 on Build GRCh38
rs759418977 Chr.1: 209675437-209675437 on Build GRCh38
rs756670467 Chr.1: 209675647-209675647 on Build GRCh38
rs543874312 Chr.1: 209675341-209675346 on Build GRCh38
rs780389970 Chr.1: 209675650-209675650 on Build GRCh38
rs552079737 Chr.1: 209675341-209675341 on Build GRCh38
rs370602402 Chr.1: 209675401-209675401 on Build GRCh38
rs371085022 Chr.1: 209675413-209675413 on Build GRCh38
rs982777181 Chr.1: 209675627-209675627 on Build GRCh38
rs987175516 Chr.1: 209675372-209675372 on Build GRCh38
rs566701786 Chr.1: 209675473-209675473 on Build GRCh38
rs949629653 Chr.1: 209675419-209675419 on Build GRCh38
rs978894905 Chr.1: 209675398-209675398 on Build GRCh38
rs954815326 Chr.1: 209675335-209675335 on Build GRCh38
rs148090379 Chr.1: 209675494-209675494 on Build GRCh38
rs936862292 Chr.1: 209675402-209675402 on Build GRCh38
rs1053086605 Chr.1: 209675242-209675242 on Build GRCh38

View Full Product Details