Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 209781964-209782468 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf74
Entrez Gene ID
148304
Gene Name
chromosome 1 open reading frame 74
Gene Aliases
URLC4
UniGene
Hs.497642
Gene Chromosome Location
Chr.1: 209779208-209784730 on Build GRCh38
Transcripts
NM_152485.3 NM_001320143.1 NM_001320144.1 NM_025228.3 XM_005273279.4 XM_005273280.2 XM_017002399.1

Gene Symbol
TRAF3IP3
Entrez Gene ID
80342
Gene Name
TRAF3 interacting protein 3
Gene Aliases
T3JAM
UniGene
Hs.147434
Gene Chromosome Location
Chr.1: 209756032-209782323 on Build GRCh38
Transcripts
NM_152485.3 NM_001320143.1 NM_001320144.1 NM_025228.3 XM_005273279.4 XM_005273280.2 XM_017002399.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6340565 c.1606G>T p.A536S Chr.1: 209782098-209782098 on Build GRCh38
3482911 c.1520G>A p.W507* Chr.1: 209782072-209782072 on Build GRCh38
1668438 c.1521G>A p.W507* Chr.1: 209782073-209782073 on Build GRCh38
32286 c.1525C>T p.P509S Chr.1: 209782077-209782077 on Build GRCh38
5502393 c.1564-7C>A p.? Chr.1: 209782049-209782049 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs568607543 Chr.1: 209782396-209782396 on Build GRCh38
rs11553707 Chr.1: 209782365-209782365 on Build GRCh38
rs546229553 Chr.1: 209782066-209782066 on Build GRCh38
rs1056198749 Chr.1: 209782159-209782159 on Build GRCh38
rs773715802 Chr.1: 209782088-209782088 on Build GRCh38
rs1008504437 Chr.1: 209782383-209782383 on Build GRCh38
rs775186506 Chr.1: 209782132-209782134 on Build GRCh38
rs749519656 Chr.1: 209782050-209782050 on Build GRCh38
rs779234473 Chr.1: 209782186-209782186 on Build GRCh38
rs745855726 Chr.1: 209782022-209782022 on Build GRCh38
rs916819024 Chr.1: 209782377-209782377 on Build GRCh38
rs780432245 Chr.1: 209782046-209782046 on Build GRCh38
rs751333631 Chr.1: 209782012-209782012 on Build GRCh38
rs527257150 Chr.1: 209782233-209782233 on Build GRCh38
rs751451429 Chr.1: 209782166-209782166 on Build GRCh38
rs763981196 Chr.1: 209782213-209782213 on Build GRCh38
rs564537532 Chr.1: 209782069-209782069 on Build GRCh38
rs748195490 Chr.1: 209782194-209782194 on Build GRCh38
rs994833789 Chr.1: 209782008-209782008 on Build GRCh38
rs913024091 Chr.1: 209782244-209782244 on Build GRCh38
rs903524786 Chr.1: 209782287-209782287 on Build GRCh38
rs138738510 Chr.1: 209782096-209782096 on Build GRCh38
rs920841438 Chr.1: 209782064-209782064 on Build GRCh38
rs144054907 Chr.1: 209782139-209782139 on Build GRCh38
rs140967802 Chr.1: 209782000-209782000 on Build GRCh38
rs772483060 Chr.1: 209782073-209782073 on Build GRCh38
rs115797133 Chr.1: 209782107-209782107 on Build GRCh38
rs537582332 Chr.1: 209782360-209782360 on Build GRCh38
rs764768391 Chr.1: 209782126-209782126 on Build GRCh38
rs772132832 Chr.1: 209782195-209782195 on Build GRCh38
rs757059821 Chr.1: 209782258-209782258 on Build GRCh38
rs141902362 Chr.1: 209782089-209782089 on Build GRCh38
rs771675397 Chr.1: 209782123-209782123 on Build GRCh38
rs748726048 Chr.1: 209782072-209782072 on Build GRCh38
rs757070403 Chr.1: 209782167-209782167 on Build GRCh38
rs781123450 Chr.1: 209782172-209782172 on Build GRCh38
rs943367789 Chr.1: 209782034-209782034 on Build GRCh38
rs755699106 Chr.1: 209782092-209782092 on Build GRCh38
rs1028883305 Chr.1: 209782209-209782209 on Build GRCh38
rs967093654 Chr.1: 209782248-209782248 on Build GRCh38
rs551776018 Chr.1: 209782234-209782234 on Build GRCh38
rs928140909 Chr.1: 209782276-209782276 on Build GRCh38
rs747546306 Chr.1: 209782197-209782197 on Build GRCh38
rs1019017139 Chr.1: 209782391-209782391 on Build GRCh38
rs372222220 Chr.1: 209782198-209782198 on Build GRCh38
rs757022735 Chr.1: 209782021-209782021 on Build GRCh38
rs751458311 Chr.1: 209782227-209782227 on Build GRCh38
rs759173782 Chr.1: 209782111-209782111 on Build GRCh38
rs544376202 Chr.1: 209782118-209782118 on Build GRCh38
rs182448330 Chr.1: 209782174-209782174 on Build GRCh38
rs529994882 Chr.1: 209782156-209782156 on Build GRCh38
rs548665844 Chr.1: 209782328-209782329 on Build GRCh38
rs1055358360 Chr.1: 209781989-209781989 on Build GRCh38
rs750059595 Chr.1: 209782409-209782409 on Build GRCh38
rs536050310 Chr.1: 209782419-209782419 on Build GRCh38
rs569357008 Chr.1: 209782147-209782147 on Build GRCh38
rs939506134 Chr.1: 209782344-209782344 on Build GRCh38
rs755216807 Chr.1: 209782184-209782184 on Build GRCh38
rs144888845 Chr.1: 209782392-209782392 on Build GRCh38
rs753849030 Chr.1: 209782394-209782394 on Build GRCh38
rs1040460852 Chr.1: 209782426-209782426 on Build GRCh38
rs886403612 Chr.1: 209782413-209782413 on Build GRCh38
rs570190103 Chr.1: 209782311-209782311 on Build GRCh38
rs374407402 Chr.1: 209782199-209782199 on Build GRCh38

View Full Product Details